Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84850441-84850714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD2
Entrez Gene ID
83716
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Gene Aliases
CRISP11, LCRISP2
UniGene
Hs.513779
Gene Chromosome Location
Chr.16: 84819981-84909510 on Build GRCh38
Transcripts
NM_031476.3 XM_005256190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4912153 c.533A>G p.N178S Chr.16: 84850608-84850608 on Build GRCh38
6876387 c.583T>C p.Y195H Chr.16: 84850658-84850658 on Build GRCh38
2154841 c.504C>A p.A168A Chr.16: 84850579-84850579 on Build GRCh38
6876386 c.583T>C p.Y195H Chr.16: 84850658-84850658 on Build GRCh38
974474 c.545A>T p.K182M Chr.16: 84850620-84850620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4783090 Chr.16: 84850510-84850510 on Build GRCh38
rs1013203732 Chr.16: 84850483-84850483 on Build GRCh38
rs771198486 Chr.16: 84850548-84850548 on Build GRCh38
rs765288617 Chr.16: 84850519-84850519 on Build GRCh38
rs779069297 Chr.16: 84850657-84850657 on Build GRCh38
rs762978773 Chr.16: 84850579-84850579 on Build GRCh38
rs539176962 Chr.16: 84850642-84850642 on Build GRCh38
rs764695065 Chr.16: 84850691-84850691 on Build GRCh38
rs751235634 Chr.16: 84850532-84850532 on Build GRCh38
rs757246124 Chr.16: 84850624-84850624 on Build GRCh38
rs1023218425 Chr.16: 84850488-84850488 on Build GRCh38
rs370001556 Chr.16: 84850527-84850527 on Build GRCh38
rs769688566 Chr.16: 84850576-84850576 on Build GRCh38
rs192130840 Chr.16: 84850540-84850540 on Build GRCh38
rs775626498 Chr.16: 84850577-84850577 on Build GRCh38
rs903327531 Chr.16: 84850468-84850468 on Build GRCh38
rs750016537 Chr.16: 84850599-84850599 on Build GRCh38
rs200807269 Chr.16: 84850511-84850511 on Build GRCh38
rs199497599 Chr.16: 84850575-84850575 on Build GRCh38
rs896175382 Chr.16: 84850481-84850481 on Build GRCh38
rs768656898 Chr.16: 84850652-84850652 on Build GRCh38
rs867736433 Chr.16: 84850479-84850479 on Build GRCh38
rs756246534 Chr.16: 84850585-84850587 on Build GRCh38
rs77992357 Chr.16: 84850537-84850537 on Build GRCh38
rs759021868 Chr.16: 84850678-84850678 on Build GRCh38
rs143085896 Chr.16: 84850611-84850611 on Build GRCh38
rs903378757 Chr.16: 84850602-84850602 on Build GRCh38
rs754247892 Chr.16: 84850534-84850534 on Build GRCh38
rs747136317 Chr.16: 84850623-84850623 on Build GRCh38
rs368719942 Chr.16: 84850617-84850617 on Build GRCh38
rs763681555 Chr.16: 84850530-84850530 on Build GRCh38
rs147475865 Chr.16: 84850648-84850648 on Build GRCh38
rs112959518 Chr.16: 84850463-84850463 on Build GRCh38
rs753074972 Chr.16: 84850682-84850682 on Build GRCh38
rs373699416 Chr.16: 84850654-84850654 on Build GRCh38
rs187545358 Chr.16: 84850467-84850467 on Build GRCh38
rs572744640 Chr.16: 84850651-84850651 on Build GRCh38
rs34925006 Chr.16: 84850594-84850594 on Build GRCh38
rs553335360 Chr.16: 84850470-84850470 on Build GRCh38
rs764053728 Chr.16: 84850582-84850582 on Build GRCh38
rs746995803 Chr.16: 84850542-84850542 on Build GRCh38
rs745715018 Chr.16: 84850573-84850573 on Build GRCh38
rs760320435 Chr.16: 84850671-84850671 on Build GRCh38
rs773922119 Chr.16: 84850587-84850587 on Build GRCh38
rs775536619 Chr.16: 84850522-84850522 on Build GRCh38
rs200065021 Chr.16: 84850664-84850664 on Build GRCh38
rs781524453 Chr.16: 84850628-84850628 on Build GRCh38
rs910526303 Chr.16: 84850566-84850566 on Build GRCh38
rs4782673 Chr.16: 84850466-84850466 on Build GRCh38
rs1023355831 Chr.16: 84850471-84850471 on Build GRCh38
rs969286559 Chr.16: 84850503-84850503 on Build GRCh38
rs1005694320 Chr.16: 84850637-84850637 on Build GRCh38
rs934837634 Chr.16: 84850603-84850603 on Build GRCh38
rs377535486 Chr.16: 84850560-84850560 on Build GRCh38
rs756936476 Chr.16: 84850533-84850533 on Build GRCh38
rs942782457 Chr.16: 84850559-84850559 on Build GRCh38
rs778486821 Chr.16: 84850626-84850629 on Build GRCh38
rs765577582 Chr.16: 84850605-84850605 on Build GRCh38
rs999705333 Chr.16: 84850473-84850473 on Build GRCh38
rs1041555756 Chr.16: 84850588-84850588 on Build GRCh38
rs541718336 Chr.16: 84850653-84850653 on Build GRCh38
rs372647970 Chr.16: 84850536-84850536 on Build GRCh38
rs753278157 Chr.16: 84850677-84850677 on Build GRCh38
rs751986671 Chr.16: 84850694-84850694 on Build GRCh38
rs927632583 Chr.16: 84850482-84850482 on Build GRCh38
rs371054674 Chr.16: 84850600-84850600 on Build GRCh38
rs767227669 Chr.16: 84850595-84850595 on Build GRCh38
rs530977189 Chr.16: 84850514-84850514 on Build GRCh38
rs185114443 Chr.16: 84850555-84850555 on Build GRCh38

View Full Product Details