Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
Chromosome Location
Chr.4: 87975650 - 87983411 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040058.1 NP_001035147.1 4 - 5 340 66
NM_001251830.1 NP_001238759.1 5 - 6 529 66
XM_017008564.1 XP_016864053.1 2 - 3 766 66
GenBank mRNA AK057738.1 - 4 - 5 276 66
AK075463.1 - 4 - 5 276 66
AK290104.1 - 4 - 5 277 66
AK295491.1 - 5 - 6 464 66
AY956318.1 - 4 - 5 242 66
BC017387.1 - 4 - 5 280 66
D28759.1 - 3 - 4 175 66
DQ839491.1 - 3 - 4 175 66
DQ846870.1 - 3 - 4 175 66
DQ846871.1 - 3 - 4 175 66
JF412666.1 - 3 - 4 177 66
KR062183.1 - 5 - 6 464 66
M83248.1 - 4 - 5 272 66
X13694.1 - 4 - 5 242 66

View Full Product Details