The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317184.1 | NP_001304113.1 | 12 - 13 | 2101 | 92 |
NM_001317185.1 | NP_001304114.1 | 13 - 14 | 2351 | 92 | |
NM_001317186.1 | NP_001304115.1 | 12 - 13 | 2138 | 92 | |
NM_004360.4 | NP_004351.1 | 13 - 14 | 2284 | 92 | |
XM_011523488.1 | XP_011521790.1 | 12 - 13 | 2148 | 92 | |
XM_011523489.1 | XP_011521791.1 | 12 - 13 | 2435 | 92 | |
GenBank mRNA | AB025105.1 | - | 12 - 13 | 2039 | 92 |
AB025106.1 | - | 13 - 14 | 2279 | 92 | |
AK290012.1 | - | 13 - 14 | 2256 | 92 | |
AK309703.1 | - | 12 - 13 | 2230 | 92 | |
AK312551.1 | - | 13 - 14 | 2284 | 92 | |
EU709494.1 | - | 13 - 14 | 2282 | 92 | |
L08599.1 | - | 13 - 14 | 2254 | 92 | |
Z13009.1 | - | 13 - 14 | 2252 | 92 | |
Z18923.1 | - | 13 - 14 | 2158 | 92 |