Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53038957-53039478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD1
Entrez Gene ID
80821
Gene Name
DDHD domain containing 1
Gene Aliases
PA-PLA1, PAPLA1, SPG28
UniGene
Hs.125525
Gene Chromosome Location
Chr.14: 53036740-53153328 on Build GRCh38
Transcripts
NM_001160147.1 NM_001160148.1 NM_030637.2 XM_011537188.2

SNP Detail

SNP ID Chromosome Location
rs912150645 Chr.14: 53039148-53039148 on Build GRCh38
rs942265003 Chr.14: 53039106-53039106 on Build GRCh38
rs937506606 Chr.14: 53039147-53039147 on Build GRCh38
rs374445469 Chr.14: 53039361-53039361 on Build GRCh38
rs904997813 Chr.14: 53039186-53039186 on Build GRCh38
rs954640544 Chr.14: 53039347-53039347 on Build GRCh38
rs1030070150 Chr.14: 53039323-53039323 on Build GRCh38
rs1004120509 Chr.14: 53039252-53039252 on Build GRCh38
rs141477612 Chr.14: 53039141-53039141 on Build GRCh38
rs1056272574 Chr.14: 53039213-53039213 on Build GRCh38
rs973583784 Chr.14: 53039450-53039450 on Build GRCh38
rs112569744 Chr.14: 53039155-53039155 on Build GRCh38
rs753420626 Chr.14: 53039307-53039307 on Build GRCh38
rs539891293 Chr.14: 53039390-53039390 on Build GRCh38
rs561579840 Chr.14: 53039351-53039351 on Build GRCh38
rs192562810 Chr.14: 53039226-53039226 on Build GRCh38
rs912096696 Chr.14: 53039359-53039359 on Build GRCh38
rs369579072 Chr.14: 53039298-53039298 on Build GRCh38
rs146858302 Chr.14: 53039446-53039446 on Build GRCh38
rs964573120 Chr.14: 53039272-53039272 on Build GRCh38
rs1037933519 Chr.14: 53039109-53039109 on Build GRCh38
rs1010226952 Chr.14: 53039214-53039214 on Build GRCh38
rs548407416 Chr.14: 53039145-53039145 on Build GRCh38
rs996448181 Chr.14: 53039282-53039282 on Build GRCh38
rs577176294 Chr.14: 53039345-53039345 on Build GRCh38
rs781075098 Chr.14: 53039161-53039161 on Build GRCh38
rs1003341123 Chr.14: 53039211-53039211 on Build GRCh38
rs749233070 Chr.14: 53039268-53039268 on Build GRCh38
rs1000754443 Chr.14: 53039257-53039257 on Build GRCh38
rs145410773 Chr.14: 53039086-53039086 on Build GRCh38
rs757367065 Chr.14: 53038988-53038988 on Build GRCh38
rs914869813 Chr.14: 53039445-53039445 on Build GRCh38
rs746156492 Chr.14: 53039023-53039023 on Build GRCh38
rs1029086191 Chr.14: 53039344-53039344 on Build GRCh38
rs2150541 Chr.14: 53039173-53039173 on Build GRCh38
rs150051791 Chr.14: 53039070-53039070 on Build GRCh38
rs747905472 Chr.14: 53039162-53039162 on Build GRCh38
rs182883863 Chr.14: 53039438-53039438 on Build GRCh38
rs73292092 Chr.14: 53039020-53039020 on Build GRCh38
rs1044840596 Chr.14: 53039185-53039185 on Build GRCh38
rs371806361 Chr.14: 53039300-53039300 on Build GRCh38
rs995301486 Chr.14: 53039285-53039285 on Build GRCh38
rs954396893 Chr.14: 53039036-53039036 on Build GRCh38
rs758489038 Chr.14: 53039095-53039095 on Build GRCh38
rs1041761690 Chr.14: 53039220-53039220 on Build GRCh38

View Full Product Details