Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50105229-50105755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDK2
Entrez Gene ID
5164
Gene Name
pyruvate dehydrogenase kinase 2
Gene Aliases
PDHK2, PDKII
UniGene
Hs.256667
Gene Chromosome Location
Chr.17: 50094737-50111369 on Build GRCh38
Transcripts
NM_001199898.1 NM_001199899.1 NM_001199900.1 NM_002611.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2698837 c.320G>A p.R107H Chr.17: 50105430-50105430 on Build GRCh38
5772122 c.308C>G p.P103R Chr.17: 50105418-50105418 on Build GRCh38
3889866 c.294C>T p.F98F Chr.17: 50105404-50105404 on Build GRCh38
314062 c.271A>T p.S91C Chr.17: 50105381-50105381 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760452103 Chr.17: 50105340-50105340 on Build GRCh38
rs765380884 Chr.17: 50105533-50105533 on Build GRCh38
rs184266557 Chr.17: 50105604-50105604 on Build GRCh38
rs754149853 Chr.17: 50105404-50105404 on Build GRCh38
rs1044947569 Chr.17: 50105626-50105626 on Build GRCh38
rs939178526 Chr.17: 50105651-50105651 on Build GRCh38
rs144791334 Chr.17: 50105453-50105453 on Build GRCh38
rs202103578 Chr.17: 50105430-50105430 on Build GRCh38
rs755327965 Chr.17: 50105397-50105397 on Build GRCh38
rs983760135 Chr.17: 50105592-50105592 on Build GRCh38
rs546121252 Chr.17: 50105649-50105649 on Build GRCh38
rs1054753871 Chr.17: 50105654-50105654 on Build GRCh38
rs574520904 Chr.17: 50105577-50105577 on Build GRCh38
rs753013740 Chr.17: 50105488-50105488 on Build GRCh38
rs753175183 Chr.17: 50105408-50105408 on Build GRCh38
rs957470246 Chr.17: 50105485-50105485 on Build GRCh38
rs74416879 Chr.17: 50105704-50105704 on Build GRCh38
rs847696 Chr.17: 50105720-50105720 on Build GRCh38
rs139747313 Chr.17: 50105613-50105613 on Build GRCh38
rs761730316 Chr.17: 50105459-50105459 on Build GRCh38
rs755173011 Chr.17: 50105468-50105468 on Build GRCh38
rs894808719 Chr.17: 50105699-50105699 on Build GRCh38
rs771488396 Chr.17: 50105429-50105429 on Build GRCh38
rs1007845391 Chr.17: 50105714-50105714 on Build GRCh38
rs892549838 Chr.17: 50105670-50105670 on Build GRCh38
rs112116459 Chr.17: 50105703-50105703 on Build GRCh38
rs750474509 Chr.17: 50105367-50105367 on Build GRCh38
rs759797419 Chr.17: 50105487-50105487 on Build GRCh38
rs140445730 Chr.17: 50105406-50105406 on Build GRCh38
rs953611731 Chr.17: 50105501-50105501 on Build GRCh38
rs534376153 Chr.17: 50105495-50105495 on Build GRCh38
rs554665103 Chr.17: 50105508-50105508 on Build GRCh38
rs545485591 Chr.17: 50105719-50105719 on Build GRCh38
rs556540017 Chr.17: 50105427-50105427 on Build GRCh38
rs756486870 Chr.17: 50105489-50105489 on Build GRCh38
rs1131996 Chr.17: 50105419-50105419 on Build GRCh38
rs766431765 Chr.17: 50105478-50105478 on Build GRCh38
rs766366400 Chr.17: 50105366-50105366 on Build GRCh38
rs1048346506 Chr.17: 50105265-50105265 on Build GRCh38
rs373389971 Chr.17: 50105462-50105462 on Build GRCh38
rs776607192 Chr.17: 50105454-50105454 on Build GRCh38
rs768639647 Chr.17: 50105347-50105347 on Build GRCh38
rs556557602 Chr.17: 50105273-50105273 on Build GRCh38
rs775436657 Chr.17: 50105251-50105251 on Build GRCh38
rs202164922 Chr.17: 50105482-50105482 on Build GRCh38
rs544637875 Chr.17: 50105543-50105543 on Build GRCh38
rs376116940 Chr.17: 50105368-50105368 on Build GRCh38
rs766561596 Chr.17: 50105374-50105374 on Build GRCh38
rs986360241 Chr.17: 50105513-50105513 on Build GRCh38
rs563421437 Chr.17: 50105606-50105606 on Build GRCh38
rs776696533 Chr.17: 50105353-50105353 on Build GRCh38
rs372205207 Chr.17: 50105358-50105358 on Build GRCh38
rs9902933 Chr.17: 50105295-50105295 on Build GRCh38
rs138234460 Chr.17: 50105377-50105377 on Build GRCh38
rs1054409379 Chr.17: 50105271-50105271 on Build GRCh38
rs929615008 Chr.17: 50105250-50105250 on Build GRCh38
rs966600691 Chr.17: 50105531-50105531 on Build GRCh38
rs12943522 Chr.17: 50105652-50105652 on Build GRCh38
rs891293519 Chr.17: 50105314-50105314 on Build GRCh38
rs1031484313 Chr.17: 50105360-50105360 on Build GRCh38
rs769804058 Chr.17: 50105460-50105460 on Build GRCh38
rs775020449 Chr.17: 50105327-50105327 on Build GRCh38
rs778166941 Chr.17: 50105420-50105420 on Build GRCh38
rs1019138825 Chr.17: 50105526-50105526 on Build GRCh38
rs375938799 Chr.17: 50105334-50105334 on Build GRCh38
rs374556397 Chr.17: 50105359-50105359 on Build GRCh38
rs947940078 Chr.17: 50105280-50105280 on Build GRCh38
rs987063915 Chr.17: 50105546-50105546 on Build GRCh38
rs918330085 Chr.17: 50105548-50105548 on Build GRCh38
rs115305261 Chr.17: 50105710-50105710 on Build GRCh38
rs112518125 Chr.17: 50105369-50105369 on Build GRCh38
rs1051661684 Chr.17: 50105313-50105313 on Build GRCh38
rs906389883 Chr.17: 50105640-50105640 on Build GRCh38
rs746716387 Chr.17: 50105451-50105451 on Build GRCh38
rs774409789 Chr.17: 50105328-50105328 on Build GRCh38

View Full Product Details