Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3630506-3631016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP5K1C
Entrez Gene ID
23396
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
Gene Aliases
LCCS3, PIP5K-GAMMA, PIP5K1-gamma, PIP5Kgamma
UniGene
Hs.282177
Gene Chromosome Location
Chr.19: 3630181-3700492 on Build GRCh38
Transcripts
NM_001195733.1 NM_012398.2 XM_017026540.1

SNP Detail

SNP ID Chromosome Location
rs914965217 Chr.19: 3630598-3630598 on Build GRCh38
rs1030719646 Chr.19: 3630977-3630977 on Build GRCh38
rs1025686388 Chr.19: 3630635-3630648 on Build GRCh38
rs543817499 Chr.19: 3630639-3630639 on Build GRCh38
rs1035388029 Chr.19: 3630553-3630553 on Build GRCh38
rs185865770 Chr.19: 3630897-3630897 on Build GRCh38
rs757169133 Chr.19: 3630949-3630949 on Build GRCh38
rs150799442 Chr.19: 3630768-3630768 on Build GRCh38
rs527814833 Chr.19: 3630950-3630950 on Build GRCh38
rs995173167 Chr.19: 3630616-3630616 on Build GRCh38
rs1027177351 Chr.19: 3630619-3630619 on Build GRCh38
rs574508045 Chr.19: 3630574-3630574 on Build GRCh38
rs930995934 Chr.19: 3630902-3630902 on Build GRCh38
rs974349425 Chr.19: 3630927-3630927 on Build GRCh38
rs990149637 Chr.19: 3630734-3630734 on Build GRCh38
rs1047624139 Chr.19: 3630708-3630708 on Build GRCh38
rs1006560450 Chr.19: 3630939-3630939 on Build GRCh38
rs965271711 Chr.19: 3630679-3630679 on Build GRCh38
rs896855907 Chr.19: 3630584-3630584 on Build GRCh38
rs753150388 Chr.19: 3630774-3630774 on Build GRCh38
rs576930255 Chr.19: 3630715-3630715 on Build GRCh38
rs928751069 Chr.19: 3630877-3630877 on Build GRCh38
rs889544703 Chr.19: 3630928-3630928 on Build GRCh38
rs367949600 Chr.19: 3630716-3630716 on Build GRCh38
rs938882775 Chr.19: 3630557-3630557 on Build GRCh38
rs1022067492 Chr.19: 3630796-3630796 on Build GRCh38
rs1006694843 Chr.19: 3630624-3630624 on Build GRCh38
rs139128069 Chr.19: 3630974-3630974 on Build GRCh38
rs576403122 Chr.19: 3630772-3630772 on Build GRCh38
rs1037725592 Chr.19: 3630891-3630891 on Build GRCh38
rs895081029 Chr.19: 3630729-3630729 on Build GRCh38
rs745337186 Chr.19: 3630978-3630978 on Build GRCh38
rs1048033692 Chr.19: 3630920-3630920 on Build GRCh38
rs938862776 Chr.19: 3630878-3630878 on Build GRCh38
rs967791353 Chr.19: 3630615-3630615 on Build GRCh38
rs550137010 Chr.19: 3630903-3630903 on Build GRCh38
rs55774465 Chr.19: 3630540-3630540 on Build GRCh38
rs923619860 Chr.19: 3630632-3630632 on Build GRCh38
rs758672648 Chr.19: 3630942-3630942 on Build GRCh38
rs980275977 Chr.19: 3630851-3630851 on Build GRCh38
rs1044560403 Chr.19: 3630534-3630534 on Build GRCh38
rs866757913 Chr.19: 3630963-3630963 on Build GRCh38
rs911451301 Chr.19: 3630908-3630908 on Build GRCh38
rs914649886 Chr.19: 3630526-3630526 on Build GRCh38
rs112547299 Chr.19: 3630692-3630692 on Build GRCh38
rs990622157 Chr.19: 3630596-3630596 on Build GRCh38
rs977862738 Chr.19: 3630876-3630876 on Build GRCh38
rs942814741 Chr.19: 3630643-3630643 on Build GRCh38
rs1056219881 Chr.19: 3630952-3630953 on Build GRCh38
rs1051312749 Chr.19: 3630638-3630638 on Build GRCh38
rs1036704815 Chr.19: 3630567-3630567 on Build GRCh38
rs376582556 Chr.19: 3630943-3630943 on Build GRCh38
rs948890969 Chr.19: 3630533-3630533 on Build GRCh38
rs920082826 Chr.19: 3630932-3630932 on Build GRCh38
rs117279098 Chr.19: 3630969-3630969 on Build GRCh38
rs537494558 Chr.19: 3630531-3630531 on Build GRCh38
rs1012012209 Chr.19: 3630767-3630767 on Build GRCh38
rs1012108391 Chr.19: 3630966-3630966 on Build GRCh38
rs755829405 Chr.19: 3630610-3630610 on Build GRCh38
rs1038565203 Chr.19: 3630677-3630678 on Build GRCh38
rs754416569 Chr.19: 3630652-3630652 on Build GRCh38
rs1034910516 Chr.19: 3630726-3630726 on Build GRCh38
rs779528081 Chr.19: 3630706-3630710 on Build GRCh38
rs994239680 Chr.19: 3630693-3630693 on Build GRCh38
rs930175634 Chr.19: 3630940-3630940 on Build GRCh38
rs967886179 Chr.19: 3630875-3630875 on Build GRCh38
rs955031736 Chr.19: 3630899-3630899 on Build GRCh38
rs753712935 Chr.19: 3630757-3630757 on Build GRCh38
rs889513505 Chr.19: 3630621-3630621 on Build GRCh38

View Full Product Details