Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108635016-108635520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102B
Entrez Gene ID
284611
Gene Name
family with sequence similarity 102 member B
Gene Aliases
SYM-3B
UniGene
Hs.200230
Gene Chromosome Location
Chr.1: 108560349-108639327 on Build GRCh38
Transcripts
NM_001010883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205998 c.1041-1G>T p.? Chr.1: 108635107-108635107 on Build GRCh38
4682332 c.1066G>A p.V356I Chr.1: 108635133-108635133 on Build GRCh38
5052352 c.979A>G p.I327V Chr.1: 108635139-108635139 on Build GRCh38
6658027 c.981C>A p.I327I Chr.1: 108635141-108635141 on Build GRCh38
6343131 c.954G>T p.G318G Chr.1: 108635114-108635114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754559561 Chr.1: 108635161-108635161 on Build GRCh38
rs558099174 Chr.1: 108635069-108635069 on Build GRCh38
rs755839292 Chr.1: 108635153-108635153 on Build GRCh38
rs770267981 Chr.1: 108635364-108635379 on Build GRCh38
rs200629120 Chr.1: 108635126-108635126 on Build GRCh38
rs774851579 Chr.1: 108635105-108635105 on Build GRCh38
rs753502382 Chr.1: 108635160-108635160 on Build GRCh38
rs879176637 Chr.1: 108635114-108635114 on Build GRCh38
rs983010053 Chr.1: 108635052-108635052 on Build GRCh38
rs895815897 Chr.1: 108635233-108635233 on Build GRCh38
rs546002868 Chr.1: 108635466-108635466 on Build GRCh38
rs969719488 Chr.1: 108635322-108635322 on Build GRCh38
rs535665754 Chr.1: 108635375-108635379 on Build GRCh38
rs76503936 Chr.1: 108635454-108635454 on Build GRCh38
rs970199790 Chr.1: 108635413-108635413 on Build GRCh38
rs41278478 Chr.1: 108635180-108635180 on Build GRCh38
rs201749748 Chr.1: 108635158-108635158 on Build GRCh38
rs950336504 Chr.1: 108635465-108635465 on Build GRCh38
rs561666388 Chr.1: 108635086-108635086 on Build GRCh38
rs540375337 Chr.1: 108635340-108635340 on Build GRCh38
rs987881678 Chr.1: 108635366-108635366 on Build GRCh38
rs377241428 Chr.1: 108635197-108635197 on Build GRCh38
rs747321671 Chr.1: 108635102-108635102 on Build GRCh38
rs574362687 Chr.1: 108635379-108635379 on Build GRCh38
rs908808832 Chr.1: 108635064-108635064 on Build GRCh38
rs551897160 Chr.1: 108635298-108635298 on Build GRCh38
rs771737802 Chr.1: 108635181-108635181 on Build GRCh38
rs767990815 Chr.1: 108635146-108635146 on Build GRCh38
rs910774073 Chr.1: 108635442-108635442 on Build GRCh38
rs1032571607 Chr.1: 108635361-108635361 on Build GRCh38
rs769899084 Chr.1: 108635059-108635059 on Build GRCh38
rs775392277 Chr.1: 108635192-108635192 on Build GRCh38
rs776972439 Chr.1: 108635165-108635166 on Build GRCh38
rs1052933573 Chr.1: 108635212-108635212 on Build GRCh38
rs750740240 Chr.1: 108635148-108635148 on Build GRCh38
rs769233263 Chr.1: 108635149-108635150 on Build GRCh38
rs770790903 Chr.1: 108635061-108635061 on Build GRCh38
rs748241550 Chr.1: 108635362-108635369 on Build GRCh38
rs921339480 Chr.1: 108635155-108635155 on Build GRCh38
rs879676180 Chr.1: 108635362-108635362 on Build GRCh38
rs531355070 Chr.1: 108635309-108635309 on Build GRCh38
rs188604153 Chr.1: 108635301-108635301 on Build GRCh38
rs752689265 Chr.1: 108635187-108635187 on Build GRCh38
rs954320441 Chr.1: 108635418-108635418 on Build GRCh38
rs562996338 Chr.1: 108635488-108635488 on Build GRCh38
rs560098170 Chr.1: 108635441-108635441 on Build GRCh38
rs764108204 Chr.1: 108635101-108635101 on Build GRCh38
rs893056497 Chr.1: 108635244-108635244 on Build GRCh38
rs1014248259 Chr.1: 108635293-108635293 on Build GRCh38
rs370345928 Chr.1: 108635362-108635378 on Build GRCh38
rs201335946 Chr.1: 108635164-108635164 on Build GRCh38
rs763047452 Chr.1: 108635070-108635070 on Build GRCh38
rs746746296 Chr.1: 108635202-108635202 on Build GRCh38
rs146864319 Chr.1: 108635088-108635088 on Build GRCh38
rs530664464 Chr.1: 108635362-108635373 on Build GRCh38
rs200324562 Chr.1: 108635203-108635203 on Build GRCh38
rs1002018662 Chr.1: 108635330-108635330 on Build GRCh38
rs372276354 Chr.1: 108635259-108635259 on Build GRCh38
rs776792497 Chr.1: 108635447-108635447 on Build GRCh38
rs943515008 Chr.1: 108635076-108635076 on Build GRCh38
rs748877332 Chr.1: 108635168-108635171 on Build GRCh38

View Full Product Details