Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185914489-185914987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRA2B
Entrez Gene ID
6434
Gene Name
transformer 2 beta homolog (Drosophila)
Gene Aliases
Htra2-beta, PPP1R156, SFRS10, SRFS10, TRA2-BETA, TRAN2B
UniGene
Hs.533122
Gene Chromosome Location
Chr.3: 185914568-185938136 on Build GRCh38
Transcripts
NM_001243879.1 NM_004593.2

SNP Detail

SNP ID Chromosome Location
rs1014526781 Chr.3: 185914881-185914881 on Build GRCh38
rs1009897998 Chr.3: 185914537-185914537 on Build GRCh38
rs1018786736 Chr.3: 185914539-185914539 on Build GRCh38
rs999424753 Chr.3: 185914804-185914804 on Build GRCh38
rs532143295 Chr.3: 185914665-185914665 on Build GRCh38
rs960717209 Chr.3: 185914558-185914560 on Build GRCh38
rs140868850 Chr.3: 185914615-185914615 on Build GRCh38
rs1037159684 Chr.3: 185914940-185914940 on Build GRCh38
rs1043637904 Chr.3: 185914674-185914674 on Build GRCh38
rs984987908 Chr.3: 185914875-185914875 on Build GRCh38
rs755772548 Chr.3: 185914857-185914857 on Build GRCh38
rs151014364 Chr.3: 185914542-185914542 on Build GRCh38
rs373472956 Chr.3: 185914753-185914753 on Build GRCh38
rs74542454 Chr.3: 185914750-185914750 on Build GRCh38
rs1043724057 Chr.3: 185914757-185914757 on Build GRCh38
rs757814561 Chr.3: 185914526-185914526 on Build GRCh38
rs376257973 Chr.3: 185914640-185914644 on Build GRCh38
rs563413121 Chr.3: 185914723-185914723 on Build GRCh38
rs989270442 Chr.3: 185914810-185914810 on Build GRCh38
rs72003092 Chr.3: 185914765-185914767 on Build GRCh38
rs949087907 Chr.3: 185914935-185914935 on Build GRCh38
rs928194060 Chr.3: 185914927-185914927 on Build GRCh38
rs1013748189 Chr.3: 185914743-185914743 on Build GRCh38
rs189663792 Chr.3: 185914582-185914582 on Build GRCh38
rs577661293 Chr.3: 185914883-185914883 on Build GRCh38
rs1001885442 Chr.3: 185914547-185914547 on Build GRCh38
rs944113476 Chr.3: 185914634-185914634 on Build GRCh38
rs537960808 Chr.3: 185914856-185914856 on Build GRCh38
rs901780020 Chr.3: 185914838-185914838 on Build GRCh38
rs903488864 Chr.3: 185914546-185914546 on Build GRCh38
rs897281280 Chr.3: 185914758-185914758 on Build GRCh38
rs995644271 Chr.3: 185914761-185914761 on Build GRCh38
rs954283567 Chr.3: 185914786-185914786 on Build GRCh38
rs1013363540 Chr.3: 185914568-185914569 on Build GRCh38
rs569795985 Chr.3: 185914807-185914807 on Build GRCh38
rs761355152 Chr.3: 185914622-185914622 on Build GRCh38
rs939491065 Chr.3: 185914706-185914706 on Build GRCh38
rs543745729 Chr.3: 185914557-185914557 on Build GRCh38
rs75907905 Chr.3: 185914524-185914524 on Build GRCh38
rs750060215 Chr.3: 185914751-185914752 on Build GRCh38
rs944161704 Chr.3: 185914517-185914517 on Build GRCh38
rs746073690 Chr.3: 185914602-185914602 on Build GRCh38
rs541074274 Chr.3: 185914914-185914914 on Build GRCh38
rs138994557 Chr.3: 185914790-185914790 on Build GRCh38
rs538637719 Chr.3: 185914958-185914958 on Build GRCh38
rs58388826 Chr.3: 185914767-185914769 on Build GRCh38
rs187122656 Chr.3: 185914847-185914847 on Build GRCh38
rs571268128 Chr.3: 185914633-185914633 on Build GRCh38
rs921678420 Chr.3: 185914564-185914564 on Build GRCh38
rs937901642 Chr.3: 185914892-185914892 on Build GRCh38
rs192025176 Chr.3: 185914871-185914871 on Build GRCh38
rs920184798 Chr.3: 185914603-185914605 on Build GRCh38
rs903791648 Chr.3: 185914677-185914677 on Build GRCh38
rs767343847 Chr.3: 185914677-185914677 on Build GRCh38
rs562306179 Chr.3: 185914532-185914532 on Build GRCh38
rs896920014 Chr.3: 185914956-185914956 on Build GRCh38
rs1037271527 Chr.3: 185914667-185914667 on Build GRCh38
rs937934188 Chr.3: 185914684-185914684 on Build GRCh38
rs896099953 Chr.3: 185914931-185914931 on Build GRCh38
rs1055083485 Chr.3: 185914912-185914912 on Build GRCh38
rs939607084 Chr.3: 185914937-185914937 on Build GRCh38
rs1054781159 Chr.3: 185914688-185914688 on Build GRCh38
rs984440320 Chr.3: 185914590-185914590 on Build GRCh38
rs1053247221 Chr.3: 185914636-185914640 on Build GRCh38
rs532363779 Chr.3: 185914612-185914613 on Build GRCh38
rs542836825 Chr.3: 185914536-185914536 on Build GRCh38
rs912592397 Chr.3: 185914627-185914629 on Build GRCh38
rs981083921 Chr.3: 185914922-185914922 on Build GRCh38
rs548104077 Chr.3: 185914796-185914796 on Build GRCh38
rs182835373 Chr.3: 185914729-185914729 on Build GRCh38
rs1011414548 Chr.3: 185914755-185914755 on Build GRCh38
rs1016167451 Chr.3: 185914578-185914582 on Build GRCh38
rs779718282 Chr.3: 185914779-185914779 on Build GRCh38

View Full Product Details