Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75314310-75314818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4900326 c.1141G>A p.E381K Chr.6: 75314691-75314691 on Build GRCh38
7000283 c.1230G>T p.K410N Chr.6: 75314602-75314602 on Build GRCh38
3922174 c.1224G>A p.R408R Chr.6: 75314608-75314608 on Build GRCh38
3629962 c.1253G>A p.S418N Chr.6: 75314579-75314579 on Build GRCh38
4522789 c.1180G>A p.D394N Chr.6: 75314652-75314652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113401431 Chr.6: 75314408-75314408 on Build GRCh38
rs764647058 Chr.6: 75314351-75314351 on Build GRCh38
rs147080592 Chr.6: 75314681-75314681 on Build GRCh38
rs749141864 Chr.6: 75314607-75314607 on Build GRCh38
rs950813994 Chr.6: 75314350-75314350 on Build GRCh38
rs756290344 Chr.6: 75314767-75314767 on Build GRCh38
rs771543313 Chr.6: 75314785-75314785 on Build GRCh38
rs377467412 Chr.6: 75314638-75314638 on Build GRCh38
rs759161187 Chr.6: 75314484-75314484 on Build GRCh38
rs778800256 Chr.6: 75314406-75314406 on Build GRCh38
rs770746648 Chr.6: 75314609-75314609 on Build GRCh38
rs148809043 Chr.6: 75314617-75314617 on Build GRCh38
rs750470097 Chr.6: 75314575-75314575 on Build GRCh38
rs369160081 Chr.6: 75314642-75314642 on Build GRCh38
rs780250278 Chr.6: 75314582-75314582 on Build GRCh38
rs778985483 Chr.6: 75314694-75314694 on Build GRCh38
rs770926736 Chr.6: 75314462-75314462 on Build GRCh38
rs753083664 Chr.6: 75314341-75314341 on Build GRCh38
rs747926737 Chr.6: 75314427-75314427 on Build GRCh38
rs745729808 Chr.6: 75314696-75314696 on Build GRCh38
rs1050867202 Chr.6: 75314534-75314534 on Build GRCh38
rs919662056 Chr.6: 75314436-75314436 on Build GRCh38
rs777628730 Chr.6: 75314432-75314432 on Build GRCh38
rs757783831 Chr.6: 75314405-75314405 on Build GRCh38
rs747756770 Chr.6: 75314378-75314381 on Build GRCh38
rs766905802 Chr.6: 75314740-75314740 on Build GRCh38
rs746478676 Chr.6: 75314791-75314791 on Build GRCh38
rs749124321 Chr.6: 75314434-75314434 on Build GRCh38
rs141211664 Chr.6: 75314729-75314729 on Build GRCh38
rs112573013 Chr.6: 75314412-75314412 on Build GRCh38
rs759714559 Chr.6: 75314746-75314746 on Build GRCh38
rs767624596 Chr.6: 75314750-75314750 on Build GRCh38
rs768376745 Chr.6: 75314641-75314641 on Build GRCh38
rs751662162 Chr.6: 75314680-75314680 on Build GRCh38
rs768162714 Chr.6: 75314715-75314715 on Build GRCh38
rs1040617206 Chr.6: 75314586-75314586 on Build GRCh38
rs557428315 Chr.6: 75314512-75314512 on Build GRCh38
rs376668841 Chr.6: 75314361-75314361 on Build GRCh38
rs972588280 Chr.6: 75314344-75314344 on Build GRCh38
rs769646566 Chr.6: 75314431-75314431 on Build GRCh38
rs901845283 Chr.6: 75314602-75314602 on Build GRCh38
rs780178951 Chr.6: 75314788-75314788 on Build GRCh38
rs149447209 Chr.6: 75314786-75314786 on Build GRCh38
rs746559220 Chr.6: 75314706-75314706 on Build GRCh38
rs181966825 Chr.6: 75314716-75314716 on Build GRCh38
rs931076119 Chr.6: 75314762-75314762 on Build GRCh38
rs866515747 Chr.6: 75314514-75314514 on Build GRCh38
rs773696946 Chr.6: 75314482-75314482 on Build GRCh38
rs891538655 Chr.6: 75314522-75314522 on Build GRCh38
rs777385545 Chr.6: 75314605-75314605 on Build GRCh38
rs373125676 Chr.6: 75314627-75314627 on Build GRCh38
rs919711527 Chr.6: 75314745-75314745 on Build GRCh38
rs151315040 Chr.6: 75314653-75314653 on Build GRCh38
rs763403214 Chr.6: 75314739-75314739 on Build GRCh38
rs751715503 Chr.6: 75314589-75314589 on Build GRCh38
rs765805359 Chr.6: 75314569-75314569 on Build GRCh38
rs753025702 Chr.6: 75314760-75314760 on Build GRCh38
rs773581684 Chr.6: 75314733-75314733 on Build GRCh38
rs746426776 Chr.6: 75314580-75314580 on Build GRCh38
rs866728885 Chr.6: 75314667-75314667 on Build GRCh38
rs41269319 Chr.6: 75314504-75314504 on Build GRCh38
rs1025520310 Chr.6: 75314657-75314657 on Build GRCh38
rs765063750 Chr.6: 75314773-75314773 on Build GRCh38
rs771831041 Chr.6: 75314488-75314488 on Build GRCh38
rs1005153142 Chr.6: 75314566-75314566 on Build GRCh38
rs750262495 Chr.6: 75314785-75314785 on Build GRCh38
rs758421123 Chr.6: 75314702-75314702 on Build GRCh38
rs1053240402 Chr.6: 75314521-75314521 on Build GRCh38
rs55780050 Chr.6: 75314644-75314644 on Build GRCh38
rs61736188 Chr.6: 75314703-75314703 on Build GRCh38
rs753556981 Chr.6: 75314682-75314682 on Build GRCh38
rs758577044 Chr.6: 75314414-75314414 on Build GRCh38
rs141766419 Chr.6: 75314676-75314676 on Build GRCh38
rs770077609 Chr.6: 75314732-75314732 on Build GRCh38
rs755243579 Chr.6: 75314593-75314593 on Build GRCh38
rs765638620 Chr.6: 75314646-75314646 on Build GRCh38
rs144226005 Chr.6: 75314526-75314526 on Build GRCh38
rs145185243 Chr.6: 75314613-75314613 on Build GRCh38
rs758532905 Chr.6: 75314581-75314581 on Build GRCh38
rs867364294 Chr.6: 75314512-75314513 on Build GRCh38
rs780293127 Chr.6: 75314416-75314416 on Build GRCh38
rs889426006 Chr.6: 75314572-75314572 on Build GRCh38
rs369951203 Chr.6: 75314520-75314520 on Build GRCh38
rs762175679 Chr.6: 75314558-75314558 on Build GRCh38
rs754365039 Chr.6: 75314378-75314378 on Build GRCh38
rs202178049 Chr.6: 75314675-75314675 on Build GRCh38
rs960539845 Chr.6: 75314658-75314658 on Build GRCh38
rs774386197 Chr.6: 75314744-75314744 on Build GRCh38
rs35464830 Chr.6: 75314349-75314349 on Build GRCh38
rs757137460 Chr.6: 75314692-75314692 on Build GRCh38

View Full Product Details