Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102318883-102318336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXF2
Entrez Gene ID
56001
Gene Name
nuclear RNA export factor 2
Gene Aliases
CT39, TAPL-2, TCP11X2
UniGene
Hs.530358
Gene Chromosome Location
Chr.X: 102247100-102326720 on Build GRCh38
Transcripts

View Full Product Details