Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56137026-56137523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8J3
Entrez Gene ID
81168
Gene Name
olfactory receptor family 8 subfamily J member 3
Gene Aliases
OR11-173
UniGene
Hs.554523
Gene Chromosome Location
Chr.11: 56136771-56137718 on Build GRCh38
Transcripts
NM_001004064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676010 c.595G>C p.V199L Chr.11: 56137124-56137124 on Build GRCh38
2150239 c.364C>T p.R122C Chr.11: 56137355-56137355 on Build GRCh38
5968362 c.386C>A p.P129H Chr.11: 56137333-56137333 on Build GRCh38
4994194 c.549T>A p.P183P Chr.11: 56137170-56137170 on Build GRCh38
313790 c.256T>A p.L86I Chr.11: 56137463-56137463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758931012 Chr.11: 56137152-56137152 on Build GRCh38
rs749528854 Chr.11: 56137292-56137292 on Build GRCh38
rs752356052 Chr.11: 56137135-56137135 on Build GRCh38
rs377708276 Chr.11: 56137374-56137374 on Build GRCh38
rs940375714 Chr.11: 56137242-56137242 on Build GRCh38
rs756078581 Chr.11: 56137333-56137333 on Build GRCh38
rs1947923 Chr.11: 56137275-56137275 on Build GRCh38
rs756342663 Chr.11: 56137286-56137286 on Build GRCh38
rs930491972 Chr.11: 56137138-56137138 on Build GRCh38
rs772204754 Chr.11: 56137296-56137296 on Build GRCh38
rs764058463 Chr.11: 56137384-56137384 on Build GRCh38
rs753990594 Chr.11: 56137386-56137386 on Build GRCh38
rs768687027 Chr.11: 56137215-56137215 on Build GRCh38
rs775970814 Chr.11: 56137461-56137461 on Build GRCh38
rs7936390 Chr.11: 56137356-56137356 on Build GRCh38
rs770427966 Chr.11: 56137310-56137311 on Build GRCh38
rs189377785 Chr.11: 56137338-56137338 on Build GRCh38
rs770591214 Chr.11: 56137293-56137293 on Build GRCh38
rs759834099 Chr.11: 56137376-56137376 on Build GRCh38
rs11227321 Chr.11: 56137352-56137352 on Build GRCh38
rs776110652 Chr.11: 56137305-56137305 on Build GRCh38
rs778817921 Chr.11: 56137350-56137350 on Build GRCh38
rs780178067 Chr.11: 56137059-56137059 on Build GRCh38
rs775501990 Chr.11: 56137354-56137354 on Build GRCh38
rs866900448 Chr.11: 56137473-56137473 on Build GRCh38
rs766290415 Chr.11: 56137107-56137107 on Build GRCh38
rs767085939 Chr.11: 56137249-56137249 on Build GRCh38
rs533143740 Chr.11: 56137389-56137389 on Build GRCh38
rs747711861 Chr.11: 56137430-56137430 on Build GRCh38
rs759577103 Chr.11: 56137244-56137244 on Build GRCh38
rs758228543 Chr.11: 56137403-56137403 on Build GRCh38
rs767141122 Chr.11: 56137115-56137115 on Build GRCh38
rs762621665 Chr.11: 56137450-56137450 on Build GRCh38
rs373232843 Chr.11: 56137303-56137303 on Build GRCh38
rs556840660 Chr.11: 56137220-56137220 on Build GRCh38
rs373304736 Chr.11: 56137392-56137392 on Build GRCh38
rs75655853 Chr.11: 56137435-56137435 on Build GRCh38
rs200183320 Chr.11: 56137360-56137360 on Build GRCh38
rs763712468 Chr.11: 56137251-56137251 on Build GRCh38
rs141817251 Chr.11: 56137055-56137055 on Build GRCh38
rs761345367 Chr.11: 56137365-56137365 on Build GRCh38
rs772372821 Chr.11: 56137453-56137453 on Build GRCh38
rs750628245 Chr.11: 56137499-56137499 on Build GRCh38
rs777795903 Chr.11: 56137290-56137290 on Build GRCh38
rs200020512 Chr.11: 56137218-56137218 on Build GRCh38
rs768683469 Chr.11: 56137416-56137416 on Build GRCh38
rs749422716 Chr.11: 56137335-56137335 on Build GRCh38
rs755470525 Chr.11: 56137314-56137314 on Build GRCh38
rs533978590 Chr.11: 56137353-56137353 on Build GRCh38
rs748240455 Chr.11: 56137285-56137285 on Build GRCh38
rs780566048 Chr.11: 56137419-56137419 on Build GRCh38
rs796250073 Chr.11: 56137219-56137219 on Build GRCh38
rs374675673 Chr.11: 56137243-56137243 on Build GRCh38
rs184855718 Chr.11: 56137112-56137112 on Build GRCh38
rs780367713 Chr.11: 56137171-56137171 on Build GRCh38
rs201535279 Chr.11: 56137355-56137355 on Build GRCh38
rs752016036 Chr.11: 56137283-56137283 on Build GRCh38
rs142082643 Chr.11: 56137470-56137470 on Build GRCh38
rs146251646 Chr.11: 56137306-56137306 on Build GRCh38
rs767975410 Chr.11: 56137136-56137137 on Build GRCh38
rs761055298 Chr.11: 56137466-56137466 on Build GRCh38
rs768740274 Chr.11: 56137069-56137069 on Build GRCh38
rs1008231775 Chr.11: 56137309-56137309 on Build GRCh38
rs755909549 Chr.11: 56137079-56137079 on Build GRCh38
rs1384094 Chr.11: 56137097-56137097 on Build GRCh38
rs747150200 Chr.11: 56137066-56137066 on Build GRCh38
rs200978969 Chr.11: 56137380-56137380 on Build GRCh38
rs146502389 Chr.11: 56137390-56137390 on Build GRCh38
rs746550275 Chr.11: 56137090-56137091 on Build GRCh38
rs148614083 Chr.11: 56137410-56137410 on Build GRCh38
rs760372350 Chr.11: 56137250-56137250 on Build GRCh38
rs112783093 Chr.11: 56137105-56137105 on Build GRCh38
rs765441051 Chr.11: 56137495-56137495 on Build GRCh38
rs755743354 Chr.11: 56137139-56137139 on Build GRCh38
rs750681288 Chr.11: 56137394-56137394 on Build GRCh38
rs750821999 Chr.11: 56137146-56137146 on Build GRCh38
rs774307758 Chr.11: 56137294-56137294 on Build GRCh38
rs754315706 Chr.11: 56137476-56137476 on Build GRCh38
rs768067505 Chr.11: 56137318-56137318 on Build GRCh38
rs751097194 Chr.11: 56137111-56137111 on Build GRCh38
rs374711816 Chr.11: 56137480-56137480 on Build GRCh38
rs939073787 Chr.11: 56137377-56137377 on Build GRCh38
rs779306688 Chr.11: 56137142-56137142 on Build GRCh38
rs778973470 Chr.11: 56137304-56137304 on Build GRCh38
rs7937461 Chr.11: 56137460-56137460 on Build GRCh38
rs139290067 Chr.11: 56137307-56137307 on Build GRCh38
rs201307710 Chr.11: 56137232-56137232 on Build GRCh38
rs758627520 Chr.11: 56137264-56137264 on Build GRCh38
rs749250964 Chr.11: 56137248-56137248 on Build GRCh38
rs377352527 Chr.11: 56137412-56137412 on Build GRCh38
rs773053114 Chr.11: 56137227-56137227 on Build GRCh38
rs376757102 Chr.11: 56137326-56137326 on Build GRCh38
rs779864478 Chr.11: 56137088-56137088 on Build GRCh38
rs769302466 Chr.11: 56137432-56137432 on Build GRCh38
rs772497322 Chr.11: 56137057-56137057 on Build GRCh38
rs538117651 Chr.11: 56137237-56137237 on Build GRCh38
rs747522678 Chr.11: 56137212-56137212 on Build GRCh38
rs751964519 Chr.11: 56137311-56137311 on Build GRCh38
rs755531760 Chr.11: 56137284-56137284 on Build GRCh38
rs748350508 Chr.11: 56137217-56137217 on Build GRCh38
rs753102892 Chr.11: 56137381-56137381 on Build GRCh38
rs868279382 Chr.11: 56137422-56137422 on Build GRCh38
rs369307213 Chr.11: 56137302-56137302 on Build GRCh38
rs753542656 Chr.11: 56137252-56137252 on Build GRCh38
rs777940073 Chr.11: 56137334-56137334 on Build GRCh38
rs540423866 Chr.11: 56137364-56137364 on Build GRCh38
rs772782648 Chr.11: 56137448-56137448 on Build GRCh38

View Full Product Details