Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16799585-16800110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XKR3
Entrez Gene ID
150165
Gene Name
XK related 3
Gene Aliases
XRG3, XTES
UniGene
Hs.356069
Gene Chromosome Location
Chr.22: 16783412-16825412 on Build GRCh38
Transcripts
NM_001318251.1 NM_175878.4 XM_017028600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013664 c.440G>T p.R147M Chr.22: 16799920-16799920 on Build GRCh38
4973528 c.525T>C p.S175S Chr.22: 16799835-16799835 on Build GRCh38
3964054 c.368G>C p.W123S Chr.22: 16799992-16799992 on Build GRCh38
5243682 c.561C>T p.L187L Chr.22: 16799799-16799799 on Build GRCh38
3842128 c.353G>T p.R118I Chr.22: 16800007-16800007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577441042 Chr.22: 16799796-16799796 on Build GRCh38
rs769391090 Chr.22: 16799941-16799941 on Build GRCh38
rs374448009 Chr.22: 16799941-16799941 on Build GRCh38
rs766347796 Chr.22: 16799747-16799747 on Build GRCh38
rs775657898 Chr.22: 16799900-16799900 on Build GRCh38
rs146283339 Chr.22: 16799884-16799884 on Build GRCh38
rs745513001 Chr.22: 16799897-16799897 on Build GRCh38
rs766355071 Chr.22: 16799920-16799920 on Build GRCh38
rs778984865 Chr.22: 16799617-16799617 on Build GRCh38
rs963231162 Chr.22: 16799661-16799661 on Build GRCh38
rs764828366 Chr.22: 16800031-16800031 on Build GRCh38
rs375886528 Chr.22: 16799740-16799740 on Build GRCh38
rs781686360 Chr.22: 16799967-16799967 on Build GRCh38
rs748539508 Chr.22: 16799734-16799734 on Build GRCh38
rs772412143 Chr.22: 16799843-16799843 on Build GRCh38
rs112619670 Chr.22: 16799888-16799888 on Build GRCh38
rs747406232 Chr.22: 16799957-16799957 on Build GRCh38
rs757924257 Chr.22: 16800034-16800034 on Build GRCh38
rs200760603 Chr.22: 16799898-16799898 on Build GRCh38
rs753406079 Chr.22: 16799830-16799830 on Build GRCh38
rs148299520 Chr.22: 16799992-16799992 on Build GRCh38
rs181203356 Chr.22: 16800057-16800057 on Build GRCh38
rs750295947 Chr.22: 16799903-16799903 on Build GRCh38
rs5748648 Chr.22: 16799932-16799932 on Build GRCh38
rs756130513 Chr.22: 16799793-16799793 on Build GRCh38
rs771292132 Chr.22: 16799961-16799961 on Build GRCh38
rs753352907 Chr.22: 16799923-16799923 on Build GRCh38
rs111932609 Chr.22: 16799999-16799999 on Build GRCh38
rs868615932 Chr.22: 16799770-16799770 on Build GRCh38
rs752323399 Chr.22: 16800032-16800032 on Build GRCh38
rs372497479 Chr.22: 16799856-16799856 on Build GRCh38
rs370231145 Chr.22: 16799758-16799758 on Build GRCh38
rs1028323967 Chr.22: 16799681-16799681 on Build GRCh38
rs368535963 Chr.22: 16799979-16799979 on Build GRCh38
rs1046083860 Chr.22: 16799940-16799940 on Build GRCh38
rs763828328 Chr.22: 16800039-16800039 on Build GRCh38
rs189983916 Chr.22: 16799905-16799905 on Build GRCh38
rs775600496 Chr.22: 16799991-16799991 on Build GRCh38
rs777996720 Chr.22: 16799950-16799950 on Build GRCh38
rs996083932 Chr.22: 16799663-16799663 on Build GRCh38
rs1004597366 Chr.22: 16799976-16799976 on Build GRCh38
rs562118958 Chr.22: 16799792-16799792 on Build GRCh38
rs373390623 Chr.22: 16799733-16799733 on Build GRCh38
rs141561342 Chr.22: 16800051-16800051 on Build GRCh38
rs559105450 Chr.22: 16799785-16799785 on Build GRCh38
rs766382936 Chr.22: 16800002-16800002 on Build GRCh38
rs769842461 Chr.22: 16799990-16799990 on Build GRCh38
rs760543897 Chr.22: 16800001-16800001 on Build GRCh38
rs1029356049 Chr.22: 16800018-16800018 on Build GRCh38
rs954211130 Chr.22: 16799694-16799694 on Build GRCh38
rs768486952 Chr.22: 16799729-16799729 on Build GRCh38
rs183667204 Chr.22: 16799842-16799842 on Build GRCh38
rs767385121 Chr.22: 16799790-16799790 on Build GRCh38
rs759752138 Chr.22: 16799784-16799784 on Build GRCh38
rs760460948 Chr.22: 16799908-16799908 on Build GRCh38
rs763207211 Chr.22: 16799901-16799901 on Build GRCh38
rs563205767 Chr.22: 16799862-16799862 on Build GRCh38
rs754569809 Chr.22: 16799930-16799930 on Build GRCh38
rs776831836 Chr.22: 16799761-16799761 on Build GRCh38
rs754622915 Chr.22: 16799841-16799841 on Build GRCh38
rs777221397 Chr.22: 16799761-16799762 on Build GRCh38
rs781586047 Chr.22: 16800061-16800061 on Build GRCh38
rs761626594 Chr.22: 16799789-16799789 on Build GRCh38
rs761536069 Chr.22: 16799742-16799742 on Build GRCh38
rs779535698 Chr.22: 16799994-16799994 on Build GRCh38
rs200976040 Chr.22: 16800015-16800015 on Build GRCh38
rs748721423 Chr.22: 16799654-16799655 on Build GRCh38
rs540965162 Chr.22: 16799688-16799688 on Build GRCh38
rs551959412 Chr.22: 16800028-16800028 on Build GRCh38
rs752376401 Chr.22: 16799935-16799935 on Build GRCh38
rs367950897 Chr.22: 16800068-16800068 on Build GRCh38
rs776632289 Chr.22: 16799886-16799886 on Build GRCh38
rs267606170 Chr.22: 16799801-16799801 on Build GRCh38
rs748488241 Chr.22: 16799851-16799851 on Build GRCh38

View Full Product Details