Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 109018106-109018656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM245
Entrez Gene ID
23731
Gene Name
transmembrane protein 245
Gene Aliases
C9orf5, CG-2, CG2
UniGene
Hs.308074
Gene Chromosome Location
Chr.9: 109015135-109119963 on Build GRCh38
Transcripts
NM_032012.3 XM_011518446.2 XM_011518447.1 XM_011518449.1 XM_011518451.1 XM_011518452.1 XM_017014571.1 XM_017014572.1

SNP Detail

SNP ID Chromosome Location
rs959889019 Chr.9: 109018515-109018515 on Build GRCh38
rs561037237 Chr.9: 109018180-109018180 on Build GRCh38
rs995002657 Chr.9: 109018149-109018149 on Build GRCh38
rs536425350 Chr.9: 109018290-109018290 on Build GRCh38
rs895771700 Chr.9: 109018174-109018174 on Build GRCh38
rs879804391 Chr.9: 109018578-109018578 on Build GRCh38
rs749909869 Chr.9: 109018243-109018243 on Build GRCh38
rs947594371 Chr.9: 109018461-109018461 on Build GRCh38
rs986404082 Chr.9: 109018389-109018389 on Build GRCh38
rs1005405259 Chr.9: 109018604-109018604 on Build GRCh38
rs961011428 Chr.9: 109018329-109018329 on Build GRCh38
rs1004558161 Chr.9: 109018492-109018492 on Build GRCh38
rs1133868 Chr.9: 109018381-109018381 on Build GRCh38
rs952581967 Chr.9: 109018221-109018221 on Build GRCh38
rs911735800 Chr.9: 109018586-109018586 on Build GRCh38
rs1049185944 Chr.9: 109018472-109018472 on Build GRCh38
rs767051090 Chr.9: 109018138-109018138 on Build GRCh38
rs966797680 Chr.9: 109018548-109018548 on Build GRCh38
rs989881089 Chr.9: 109018250-109018255 on Build GRCh38
rs534869394 Chr.9: 109018428-109018428 on Build GRCh38
rs910678233 Chr.9: 109018460-109018460 on Build GRCh38
rs575797695 Chr.9: 109018617-109018617 on Build GRCh38
rs539373025 Chr.9: 109018547-109018547 on Build GRCh38
rs978148146 Chr.9: 109018566-109018566 on Build GRCh38
rs998831896 Chr.9: 109018136-109018136 on Build GRCh38
rs137858607 Chr.9: 109018357-109018357 on Build GRCh38
rs114531063 Chr.9: 109018613-109018613 on Build GRCh38
rs1043099103 Chr.9: 109018498-109018498 on Build GRCh38
rs528118580 Chr.9: 109018191-109018191 on Build GRCh38
rs1003760659 Chr.9: 109018512-109018512 on Build GRCh38
rs926630889 Chr.9: 109018231-109018231 on Build GRCh38
rs1027439521 Chr.9: 109018151-109018151 on Build GRCh38
rs1057100031 Chr.9: 109018551-109018551 on Build GRCh38
rs190299919 Chr.9: 109018240-109018240 on Build GRCh38
rs1004359834 Chr.9: 109018156-109018156 on Build GRCh38
rs368773555 Chr.9: 109018192-109018192 on Build GRCh38
rs970964528 Chr.9: 109018195-109018196 on Build GRCh38
rs757336149 Chr.9: 109018382-109018382 on Build GRCh38
rs1022892449 Chr.9: 109018599-109018599 on Build GRCh38
rs915369773 Chr.9: 109018306-109018306 on Build GRCh38
rs895818087 Chr.9: 109018557-109018557 on Build GRCh38
rs989520516 Chr.9: 109018527-109018527 on Build GRCh38
rs900126833 Chr.9: 109018144-109018144 on Build GRCh38
rs185440827 Chr.9: 109018225-109018225 on Build GRCh38
rs567710938 Chr.9: 109018317-109018317 on Build GRCh38
rs532247302 Chr.9: 109018210-109018210 on Build GRCh38
rs1013214310 Chr.9: 109018573-109018573 on Build GRCh38
rs903300757 Chr.9: 109018522-109018522 on Build GRCh38
rs371312177 Chr.9: 109018304-109018304 on Build GRCh38
rs564841305 Chr.9: 109018208-109018208 on Build GRCh38
rs994551380 Chr.9: 109018407-109018407 on Build GRCh38
rs185718293 Chr.9: 109018499-109018499 on Build GRCh38
rs748835370 Chr.9: 109018193-109018196 on Build GRCh38
rs888251659 Chr.9: 109018600-109018600 on Build GRCh38
rs777996131 Chr.9: 109018330-109018330 on Build GRCh38
rs1033971775 Chr.9: 109018513-109018513 on Build GRCh38
rs181220845 Chr.9: 109018301-109018301 on Build GRCh38
rs78397412 Chr.9: 109018458-109018458 on Build GRCh38
rs886216919 Chr.9: 109018480-109018480 on Build GRCh38
rs570103853 Chr.9: 109018176-109018176 on Build GRCh38
rs950516722 Chr.9: 109018155-109018155 on Build GRCh38

View Full Product Details