Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39064876-39065375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100131347
Entrez Gene ID
100131347
Gene Name
RAD52 motif containing 1 pseudogene
Gene Aliases
-
UniGene
Hs.676510
Gene Chromosome Location
Chr.17: 39057019-39081451 on Build GRCh38
Transcripts
NR_036551.1 NM_020405.4

Gene Symbol
PLXDC1
Entrez Gene ID
57125
Gene Name
plexin domain containing 1
Gene Aliases
TEM3, TEM7
UniGene
Hs.125036
Gene Chromosome Location
Chr.17: 39063303-39151649 on Build GRCh38
Transcripts
NR_036551.1 NM_020405.4

SNP Detail

SNP ID Chromosome Location
rs777495332 Chr.17: 39064974-39064974 on Build GRCh38
rs894225925 Chr.17: 39065223-39065223 on Build GRCh38
rs1003058270 Chr.17: 39064995-39064995 on Build GRCh38
rs536378885 Chr.17: 39064947-39064947 on Build GRCh38
rs920959341 Chr.17: 39065228-39065229 on Build GRCh38
rs542580024 Chr.17: 39065131-39065131 on Build GRCh38
rs756588705 Chr.17: 39064950-39064950 on Build GRCh38
rs887317872 Chr.17: 39064988-39064988 on Build GRCh38
rs770185009 Chr.17: 39065236-39065236 on Build GRCh38
rs117460627 Chr.17: 39065065-39065065 on Build GRCh38
rs541894114 Chr.17: 39065148-39065148 on Build GRCh38
rs748735773 Chr.17: 39065221-39065221 on Build GRCh38
rs117169947 Chr.17: 39065027-39065027 on Build GRCh38
rs1019426701 Chr.17: 39065066-39065066 on Build GRCh38
rs116524631 Chr.17: 39065111-39065111 on Build GRCh38
rs75586806 Chr.17: 39064911-39064911 on Build GRCh38
rs574572748 Chr.17: 39065147-39065147 on Build GRCh38
rs1039314959 Chr.17: 39064907-39064907 on Build GRCh38
rs531084810 Chr.17: 39065159-39065159 on Build GRCh38
rs76079214 Chr.17: 39065258-39065258 on Build GRCh38
rs569831440 Chr.17: 39064994-39064994 on Build GRCh38
rs1048181700 Chr.17: 39065305-39065305 on Build GRCh38
rs894165535 Chr.17: 39065016-39065016 on Build GRCh38
rs547402728 Chr.17: 39065302-39065302 on Build GRCh38
rs965224295 Chr.17: 39065076-39065076 on Build GRCh38
rs565646917 Chr.17: 39065314-39065314 on Build GRCh38
rs931085868 Chr.17: 39064980-39064980 on Build GRCh38
rs1048710602 Chr.17: 39064903-39064903 on Build GRCh38
rs148226547 Chr.17: 39065151-39065151 on Build GRCh38
rs940273154 Chr.17: 39065149-39065149 on Build GRCh38
rs938183926 Chr.17: 39065179-39065179 on Build GRCh38
rs546087311 Chr.17: 39065215-39065215 on Build GRCh38
rs748956667 Chr.17: 39064989-39064989 on Build GRCh38
rs967277432 Chr.17: 39065041-39065041 on Build GRCh38
rs940220846 Chr.17: 39064935-39064935 on Build GRCh38
rs756907082 Chr.17: 39065040-39065040 on Build GRCh38
rs778453281 Chr.17: 39065095-39065095 on Build GRCh38
rs529737039 Chr.17: 39065343-39065343 on Build GRCh38
rs143969163 Chr.17: 39065145-39065145 on Build GRCh38
rs149611241 Chr.17: 39064986-39064986 on Build GRCh38
rs527572739 Chr.17: 39065013-39065013 on Build GRCh38
rs148203398 Chr.17: 39065175-39065175 on Build GRCh38
rs902834541 Chr.17: 39065347-39065347 on Build GRCh38
rs928150644 Chr.17: 39065173-39065173 on Build GRCh38
rs553387057 Chr.17: 39065032-39065032 on Build GRCh38
rs1004786415 Chr.17: 39065002-39065002 on Build GRCh38
rs189076220 Chr.17: 39065216-39065216 on Build GRCh38
rs1046645463 Chr.17: 39065012-39065012 on Build GRCh38

View Full Product Details