Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156069625-156070124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB25
Entrez Gene ID
57111
Gene Name
RAB25, member RAS oncogene family
Gene Aliases
CATX-8, RAB11C
UniGene
Hs.632469
Gene Chromosome Location
Chr.1: 156061149-156070514 on Build GRCh38
Transcripts
NM_020387.3 NR_133653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7120855 c.476C>G p.S159C Chr.1: 156069713-156069713 on Build GRCh38
4939897 c.492C>G p.T164T Chr.1: 156069717-156069717 on Build GRCh38
6459395 c.522G>C p.L174L Chr.1: 156069747-156069747 on Build GRCh38
6121403 c.510G>A p.L170L Chr.1: 156069747-156069747 on Build GRCh38
6262732 c.514+7G>T p.? Chr.1: 156069758-156069758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200597559 Chr.1: 156069782-156069782 on Build GRCh38
rs571452256 Chr.1: 156069716-156069716 on Build GRCh38
rs769419178 Chr.1: 156069878-156069878 on Build GRCh38
rs535972958 Chr.1: 156069837-156069837 on Build GRCh38
rs931879171 Chr.1: 156069731-156069731 on Build GRCh38
rs767167776 Chr.1: 156069678-156069678 on Build GRCh38
rs765028300 Chr.1: 156069710-156069710 on Build GRCh38
rs771260017 Chr.1: 156069669-156069669 on Build GRCh38
rs771365149 Chr.1: 156069650-156069651 on Build GRCh38
rs371193816 Chr.1: 156069802-156069802 on Build GRCh38
rs1024298259 Chr.1: 156070002-156070002 on Build GRCh38
rs758368794 Chr.1: 156069715-156069715 on Build GRCh38
rs760040854 Chr.1: 156069676-156069676 on Build GRCh38
rs777973695 Chr.1: 156069738-156069738 on Build GRCh38
rs1007322003 Chr.1: 156070058-156070058 on Build GRCh38
rs982095690 Chr.1: 156069776-156069787 on Build GRCh38
rs538502117 Chr.1: 156069719-156069719 on Build GRCh38
rs771222468 Chr.1: 156069741-156069741 on Build GRCh38
rs772757207 Chr.1: 156069769-156069769 on Build GRCh38
rs796066801 Chr.1: 156069824-156069827 on Build GRCh38
rs746229301 Chr.1: 156069791-156069791 on Build GRCh38
rs749680413 Chr.1: 156069653-156069653 on Build GRCh38
rs554230917 Chr.1: 156069842-156069842 on Build GRCh38
rs994340609 Chr.1: 156069841-156069841 on Build GRCh38
rs151082047 Chr.1: 156070095-156070095 on Build GRCh38
rs760331746 Chr.1: 156069780-156069780 on Build GRCh38
rs749469341 Chr.1: 156069739-156069739 on Build GRCh38
rs975824650 Chr.1: 156070099-156070099 on Build GRCh38
rs773407896 Chr.1: 156069652-156069652 on Build GRCh38
rs767962154 Chr.1: 156069788-156069788 on Build GRCh38
rs28534050 Chr.1: 156070097-156070097 on Build GRCh38
rs746188197 Chr.1: 156069765-156069765 on Build GRCh38
rs902564259 Chr.1: 156069819-156069819 on Build GRCh38
rs368834458 Chr.1: 156069688-156069688 on Build GRCh38
rs965463223 Chr.1: 156070084-156070084 on Build GRCh38
rs776165422 Chr.1: 156069788-156069788 on Build GRCh38
rs540494183 Chr.1: 156069988-156069988 on Build GRCh38
rs960844785 Chr.1: 156069950-156069950 on Build GRCh38
rs375123831 Chr.1: 156069686-156069686 on Build GRCh38
rs897289927 Chr.1: 156069839-156069839 on Build GRCh38
rs1054149434 Chr.1: 156069702-156069702 on Build GRCh38
rs774732367 Chr.1: 156069673-156069673 on Build GRCh38
rs757011349 Chr.1: 156069707-156069707 on Build GRCh38
rs377745191 Chr.1: 156069792-156069792 on Build GRCh38
rs1018261266 Chr.1: 156070074-156070074 on Build GRCh38
rs142878365 Chr.1: 156069903-156069903 on Build GRCh38
rs1022296710 Chr.1: 156069812-156069812 on Build GRCh38
rs565431691 Chr.1: 156070086-156070086 on Build GRCh38
rs759775661 Chr.1: 156069742-156069751 on Build GRCh38
rs893082251 Chr.1: 156069784-156069784 on Build GRCh38
rs984825819 Chr.1: 156070091-156070091 on Build GRCh38
rs567486849 Chr.1: 156069684-156069684 on Build GRCh38
rs757389803 Chr.1: 156069850-156069850 on Build GRCh38
rs940418176 Chr.1: 156070096-156070096 on Build GRCh38
rs776129273 Chr.1: 156069778-156069778 on Build GRCh38
rs750355500 Chr.1: 156069711-156069711 on Build GRCh38
rs756180463 Chr.1: 156069728-156069728 on Build GRCh38
rs1027677740 Chr.1: 156069848-156069848 on Build GRCh38
rs577477547 Chr.1: 156070092-156070092 on Build GRCh38
rs200069825 Chr.1: 156069742-156069742 on Build GRCh38
rs558136209 Chr.1: 156069657-156069658 on Build GRCh38
rs534531639 Chr.1: 156069687-156069687 on Build GRCh38

View Full Product Details