Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34985217-34985486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM35B
Entrez Gene ID
100506144
Gene Name
transmembrane protein 35B
Gene Aliases
ZMYM6NB
UniGene
Hs.533986
Gene Chromosome Location
Chr.1: 34981526-34985347 on Build GRCh38
Transcripts
NM_001195156.1

SNP Detail

SNP ID Chromosome Location
rs756368285 Chr.1: 34985310-34985311 on Build GRCh38
rs754523731 Chr.1: 34985238-34985238 on Build GRCh38
rs948937146 Chr.1: 34985253-34985253 on Build GRCh38
rs780739916 Chr.1: 34985298-34985298 on Build GRCh38
rs796327202 Chr.1: 34985387-34985387 on Build GRCh38
rs1032125251 Chr.1: 34985255-34985255 on Build GRCh38
rs774465256 Chr.1: 34985354-34985354 on Build GRCh38
rs902207446 Chr.1: 34985236-34985236 on Build GRCh38
rs866326031 Chr.1: 34985242-34985242 on Build GRCh38
rs915953575 Chr.1: 34985249-34985249 on Build GRCh38
rs770825052 Chr.1: 34985344-34985344 on Build GRCh38
rs570099897 Chr.1: 34985418-34985418 on Build GRCh38
rs116315555 Chr.1: 34985460-34985460 on Build GRCh38
rs1042332519 Chr.1: 34985377-34985377 on Build GRCh38
rs1043299997 Chr.1: 34985305-34985305 on Build GRCh38
rs566317990 Chr.1: 34985320-34985320 on Build GRCh38
rs902168394 Chr.1: 34985392-34985392 on Build GRCh38
rs900177169 Chr.1: 34985390-34985390 on Build GRCh38
rs72907569 Chr.1: 34985416-34985416 on Build GRCh38
rs1019756886 Chr.1: 34985287-34985287 on Build GRCh38
rs1008681961 Chr.1: 34985351-34985351 on Build GRCh38
rs999904887 Chr.1: 34985314-34985314 on Build GRCh38
rs918563824 Chr.1: 34985302-34985302 on Build GRCh38
rs952781789 Chr.1: 34985304-34985304 on Build GRCh38
rs1053315098 Chr.1: 34985335-34985335 on Build GRCh38
rs965571451 Chr.1: 34985291-34985291 on Build GRCh38
rs747553730 Chr.1: 34985319-34985319 on Build GRCh38
rs1038679698 Chr.1: 34985360-34985360 on Build GRCh38
rs1006971331 Chr.1: 34985278-34985278 on Build GRCh38
rs111709408 Chr.1: 34985339-34985339 on Build GRCh38
rs889507051 Chr.1: 34985426-34985427 on Build GRCh38
rs953802485 Chr.1: 34985258-34985258 on Build GRCh38
rs973037339 Chr.1: 34985293-34985293 on Build GRCh38
rs993227161 Chr.1: 34985237-34985237 on Build GRCh38

View Full Product Details