Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41540628-41540899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR3H
Entrez Gene ID
171568
Gene Name
RNA polymerase III subunit H
Gene Aliases
RPC22.9, RPC8
UniGene
Hs.712617
Gene Chromosome Location
Chr.22: 41525799-41544606 on Build GRCh38
Transcripts
NM_001018050.3 NM_001018052.3 NM_001282884.1 NM_001282885.1 NM_138338.4 XM_011529993.1 XM_017028641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3554945 c.112-1G>T p.? Chr.22: 41540796-41540796 on Build GRCh38
6380315 c.199C>T p.H67Y Chr.22: 41540708-41540708 on Build GRCh38
6752461 c.191G>A p.G64D Chr.22: 41540716-41540716 on Build GRCh38
6752459 c.162G>A p.L54L Chr.22: 41540745-41540745 on Build GRCh38
3554946 c.112-1G>T p.? Chr.22: 41540796-41540796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376996942 Chr.22: 41540809-41540809 on Build GRCh38
rs1028045993 Chr.22: 41540780-41540780 on Build GRCh38
rs751505096 Chr.22: 41540770-41540770 on Build GRCh38
rs376017577 Chr.22: 41540805-41540805 on Build GRCh38
rs370405967 Chr.22: 41540792-41540792 on Build GRCh38
rs142800678 Chr.22: 41540725-41540725 on Build GRCh38
rs28469669 Chr.22: 41540680-41540680 on Build GRCh38
rs772552660 Chr.22: 41540801-41540801 on Build GRCh38
rs766365230 Chr.22: 41540753-41540753 on Build GRCh38
rs142795860 Chr.22: 41540854-41540854 on Build GRCh38
rs923350273 Chr.22: 41540852-41540852 on Build GRCh38
rs763023726 Chr.22: 41540847-41540848 on Build GRCh38
rs761617738 Chr.22: 41540737-41540737 on Build GRCh38
rs1012694709 Chr.22: 41540777-41540777 on Build GRCh38
rs755109778 Chr.22: 41540660-41540660 on Build GRCh38
rs199809623 Chr.22: 41540706-41540706 on Build GRCh38
rs764783735 Chr.22: 41540740-41540740 on Build GRCh38
rs751404461 Chr.22: 41540655-41540655 on Build GRCh38
rs201601310 Chr.22: 41540714-41540714 on Build GRCh38
rs770214794 Chr.22: 41540709-41540709 on Build GRCh38
rs762761702 Chr.22: 41540746-41540746 on Build GRCh38
rs760837242 Chr.22: 41540759-41540762 on Build GRCh38
rs766516697 Chr.22: 41540806-41540809 on Build GRCh38
rs373950442 Chr.22: 41540662-41540662 on Build GRCh38
rs754776458 Chr.22: 41540771-41540771 on Build GRCh38
rs772550402 Chr.22: 41540829-41540829 on Build GRCh38
rs772614082 Chr.22: 41540727-41540727 on Build GRCh38
rs767363401 Chr.22: 41540772-41540772 on Build GRCh38
rs776419185 Chr.22: 41540734-41540734 on Build GRCh38
rs377376349 Chr.22: 41540776-41540776 on Build GRCh38
rs138200565 Chr.22: 41540795-41540795 on Build GRCh38
rs753944316 Chr.22: 41540838-41540839 on Build GRCh38
rs779452694 Chr.22: 41540797-41540797 on Build GRCh38
rs769856766 Chr.22: 41540672-41540672 on Build GRCh38
rs746401124 Chr.22: 41540800-41540800 on Build GRCh38
rs771472417 Chr.22: 41540715-41540715 on Build GRCh38
rs370993475 Chr.22: 41540815-41540815 on Build GRCh38
rs897879648 Chr.22: 41540723-41540723 on Build GRCh38
rs561734411 Chr.22: 41540820-41540820 on Build GRCh38
rs756339721 Chr.22: 41540783-41540783 on Build GRCh38
rs778309126 Chr.22: 41540873-41540873 on Build GRCh38
rs769602998 Chr.22: 41540824-41540824 on Build GRCh38
rs765851017 Chr.22: 41540831-41540831 on Build GRCh38
rs748405557 Chr.22: 41540664-41540664 on Build GRCh38
rs772855372 Chr.22: 41540743-41540743 on Build GRCh38
rs962044846 Chr.22: 41540817-41540817 on Build GRCh38
rs201966659 Chr.22: 41540716-41540716 on Build GRCh38
rs749414419 Chr.22: 41540793-41540793 on Build GRCh38

View Full Product Details