Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75313115-75313364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6901056 c.2564G>A p.R855K Chr.6: 75313268-75313268 on Build GRCh38
1661861 c.2539G>A p.V847M Chr.6: 75313293-75313293 on Build GRCh38
3875555 c.2634C>T p.N878N Chr.6: 75313198-75313198 on Build GRCh38
6876904 c.2481C>T p.S827S Chr.6: 75313351-75313351 on Build GRCh38
6471386 c.2666G>A p.R889Q Chr.6: 75313166-75313166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760288859 Chr.6: 75313269-75313269 on Build GRCh38
rs763857606 Chr.6: 75313169-75313169 on Build GRCh38
rs745722891 Chr.6: 75313261-75313261 on Build GRCh38
rs144188799 Chr.6: 75313166-75313166 on Build GRCh38
rs184534406 Chr.6: 75313213-75313213 on Build GRCh38
rs780053107 Chr.6: 75313205-75313205 on Build GRCh38
rs751607164 Chr.6: 75313325-75313325 on Build GRCh38
rs140528948 Chr.6: 75313146-75313146 on Build GRCh38
rs141090895 Chr.6: 75313316-75313316 on Build GRCh38
rs750947467 Chr.6: 75313197-75313197 on Build GRCh38
rs766903756 Chr.6: 75313317-75313317 on Build GRCh38
rs763807669 Chr.6: 75313279-75313279 on Build GRCh38
rs775088298 Chr.6: 75313268-75313268 on Build GRCh38
rs750815354 Chr.6: 75313297-75313297 on Build GRCh38
rs757601616 Chr.6: 75313185-75313185 on Build GRCh38
rs146644501 Chr.6: 75313238-75313238 on Build GRCh38
rs372347173 Chr.6: 75313174-75313174 on Build GRCh38
rs774205816 Chr.6: 75313152-75313152 on Build GRCh38
rs576355587 Chr.6: 75313267-75313267 on Build GRCh38
rs150255059 Chr.6: 75313335-75313335 on Build GRCh38
rs765584991 Chr.6: 75313295-75313295 on Build GRCh38
rs750454653 Chr.6: 75313294-75313294 on Build GRCh38
rs199510768 Chr.6: 75313162-75313162 on Build GRCh38
rs867896706 Chr.6: 75313221-75313221 on Build GRCh38
rs1027673508 Chr.6: 75313186-75313186 on Build GRCh38
rs867134216 Chr.6: 75313157-75313157 on Build GRCh38
rs775141616 Chr.6: 75313167-75313167 on Build GRCh38
rs758784720 Chr.6: 75313314-75313314 on Build GRCh38
rs760342188 Chr.6: 75313168-75313168 on Build GRCh38
rs758951374 Chr.6: 75313203-75313203 on Build GRCh38
rs765750626 Chr.6: 75313194-75313194 on Build GRCh38
rs770693128 Chr.6: 75313245-75313245 on Build GRCh38
rs749090836 Chr.6: 75313242-75313242 on Build GRCh38
rs774343848 Chr.6: 75313260-75313260 on Build GRCh38
rs201741737 Chr.6: 75313148-75313148 on Build GRCh38
rs755154067 Chr.6: 75313227-75313227 on Build GRCh38
rs776471964 Chr.6: 75313291-75313291 on Build GRCh38

View Full Product Details