Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47573404-47573673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD4
Entrez Gene ID
55709
Gene Name
kelch repeat and BTB domain containing 4
Gene Aliases
BKLHD4, HSPC252
UniGene
Hs.718483
Gene Chromosome Location
Chr.11: 47572197-47579015 on Build GRCh38
Transcripts
NM_001318716.1 NM_001318717.1 NM_001318718.1 NM_001318719.1 NM_001318720.1 NM_001318721.1 NM_001318722.1 NM_001318723.1 NM_001318724.1 NM_001318725.1 NM_016506.6 NM_018095.5 XM_017018009.1 NM_001143984.1 NM_175732.2

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
UniGene
Hs.656205
Gene Chromosome Location
Chr.11: 47565430-47573461 on Build GRCh38
Transcripts
NM_001318716.1 NM_001318717.1 NM_001318718.1 NM_001318719.1 NM_001318720.1 NM_001318721.1 NM_001318722.1 NM_001318723.1 NM_001318724.1 NM_001318725.1 NM_016506.6 NM_018095.5 XM_017018009.1 NM_001143984.1 NM_175732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4821334 c.856G>C p.D286H Chr.11: 47573631-47573631 on Build GRCh38
7416886 c.930G>A p.W310* Chr.11: 47573557-47573557 on Build GRCh38
4574404 c.981C>T p.P327P Chr.11: 47573506-47573506 on Build GRCh38
4866569 c.1103G>A p.R368H Chr.11: 47573432-47573432 on Build GRCh38
5955354 c.1049C>A p.S350* Chr.11: 47573486-47573486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748140900 Chr.11: 47573615-47573615 on Build GRCh38
rs768581356 Chr.11: 47573620-47573620 on Build GRCh38
rs909392785 Chr.11: 47573456-47573456 on Build GRCh38
rs765630705 Chr.11: 47573485-47573485 on Build GRCh38
rs139512073 Chr.11: 47573506-47573506 on Build GRCh38
rs753654999 Chr.11: 47573476-47573476 on Build GRCh38
rs776295137 Chr.11: 47573625-47573625 on Build GRCh38
rs773599306 Chr.11: 47573429-47573429 on Build GRCh38
rs781403257 Chr.11: 47573528-47573528 on Build GRCh38
rs1036617814 Chr.11: 47573505-47573505 on Build GRCh38
rs759804817 Chr.11: 47573651-47573651 on Build GRCh38
rs372051017 Chr.11: 47573650-47573650 on Build GRCh38
rs775355395 Chr.11: 47573449-47573449 on Build GRCh38
rs758741599 Chr.11: 47573537-47573537 on Build GRCh38
rs754116406 Chr.11: 47573448-47573448 on Build GRCh38
rs143529938 Chr.11: 47573431-47573431 on Build GRCh38
rs375208548 Chr.11: 47573509-47573509 on Build GRCh38
rs367925558 Chr.11: 47573632-47573632 on Build GRCh38
rs748955906 Chr.11: 47573430-47573430 on Build GRCh38
rs149622767 Chr.11: 47573530-47573530 on Build GRCh38
rs867676589 Chr.11: 47573460-47573460 on Build GRCh38
rs774949831 Chr.11: 47573597-47573597 on Build GRCh38
rs144325348 Chr.11: 47573579-47573579 on Build GRCh38
rs745403900 Chr.11: 47573582-47573582 on Build GRCh38
rs767776935 Chr.11: 47573652-47573652 on Build GRCh38
rs749606273 Chr.11: 47573540-47573540 on Build GRCh38
rs758977537 Chr.11: 47573434-47573434 on Build GRCh38
rs766534364 Chr.11: 47573641-47573641 on Build GRCh38
rs755063316 Chr.11: 47573511-47573511 on Build GRCh38
rs760663862 Chr.11: 47573598-47573598 on Build GRCh38
rs756681498 Chr.11: 47573533-47573533 on Build GRCh38
rs931437850 Chr.11: 47573541-47573541 on Build GRCh38
rs770492766 Chr.11: 47573572-47573572 on Build GRCh38
rs11039302 Chr.11: 47573499-47573499 on Build GRCh38
rs1046517505 Chr.11: 47573571-47573571 on Build GRCh38
rs767471949 Chr.11: 47573447-47573447 on Build GRCh38
rs760584018 Chr.11: 47573459-47573459 on Build GRCh38
rs773744022 Chr.11: 47573432-47573432 on Build GRCh38
rs147179417 Chr.11: 47573461-47573461 on Build GRCh38

View Full Product Details