Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55564222-55564461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R3B
Entrez Gene ID
57223
Gene Name
protein phosphatase 4 regulatory subunit 3B
Gene Aliases
FLFL2, PP4R3B, PSY2, SMEK2, smk1
UniGene
Hs.516182
Gene Chromosome Location
Chr.2: 55547292-55617757 on Build GRCh38
Transcripts
NM_001122964.2 NM_001282850.1 NM_020463.3 XM_005264442.2 XM_005264444.2 XM_005264445.2 XM_005264446.2 XM_005264447.2 XM_017004531.1 XM_017004532.1 XM_017004533.1 XM_017004534.1 XM_017004535.1 XM_017004536.1 XM_017004537.1 XM_017004538.1 XM_017004539.1 XM_017004540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094916 c.2178A>C p.K726N Chr.2: 55564395-55564395 on Build GRCh38
4094917 c.1923A>C p.K641N Chr.2: 55564395-55564395 on Build GRCh38
1483158 c.2155G>A p.E719K Chr.2: 55564418-55564418 on Build GRCh38
5661575 c.1943A>G p.E648G Chr.2: 55564375-55564375 on Build GRCh38
1306902 c.1904A>T p.D635V Chr.2: 55564414-55564414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374496252 Chr.2: 55564298-55564298 on Build GRCh38
rs760364591 Chr.2: 55564321-55564321 on Build GRCh38
rs756214017 Chr.2: 55564416-55564416 on Build GRCh38
rs762665530 Chr.2: 55564373-55564373 on Build GRCh38
rs1000011209 Chr.2: 55564322-55564322 on Build GRCh38
rs978638203 Chr.2: 55564435-55564435 on Build GRCh38
rs779373535 Chr.2: 55564433-55564433 on Build GRCh38
rs775482180 Chr.2: 55564364-55564364 on Build GRCh38
rs751721216 Chr.2: 55564286-55564286 on Build GRCh38
rs756768748 Chr.2: 55564435-55564437 on Build GRCh38
rs889815634 Chr.2: 55564288-55564288 on Build GRCh38
rs893789335 Chr.2: 55564429-55564429 on Build GRCh38
rs370731321 Chr.2: 55564406-55564406 on Build GRCh38
rs989738948 Chr.2: 55564407-55564407 on Build GRCh38
rs114827448 Chr.2: 55564303-55564303 on Build GRCh38
rs752479091 Chr.2: 55564273-55564273 on Build GRCh38
rs748504654 Chr.2: 55564437-55564437 on Build GRCh38
rs759059901 Chr.2: 55564344-55564344 on Build GRCh38
rs1033956185 Chr.2: 55564358-55564358 on Build GRCh38
rs755332412 Chr.2: 55564430-55564430 on Build GRCh38
rs143223785 Chr.2: 55564391-55564391 on Build GRCh38
rs749674250 Chr.2: 55564301-55564301 on Build GRCh38
rs780439305 Chr.2: 55564418-55564418 on Build GRCh38
rs1031041217 Chr.2: 55564404-55564404 on Build GRCh38
rs758204890 Chr.2: 55564281-55564281 on Build GRCh38
rs979838195 Chr.2: 55564355-55564355 on Build GRCh38
rs1005455602 Chr.2: 55564294-55564294 on Build GRCh38
rs767702898 Chr.2: 55564396-55564396 on Build GRCh38
rs746394398 Chr.2: 55564299-55564299 on Build GRCh38
rs763879823 Chr.2: 55564383-55564383 on Build GRCh38
rs770253599 Chr.2: 55564300-55564300 on Build GRCh38
rs757330269 Chr.2: 55564296-55564296 on Build GRCh38
rs533630759 Chr.2: 55564329-55564329 on Build GRCh38
rs777910806 Chr.2: 55564282-55564282 on Build GRCh38
rs765202362 Chr.2: 55564263-55564263 on Build GRCh38
rs138438998 Chr.2: 55564420-55564420 on Build GRCh38
rs994367384 Chr.2: 55564302-55564302 on Build GRCh38
rs765158627 Chr.2: 55564359-55564359 on Build GRCh38
rs370443835 Chr.2: 55564289-55564289 on Build GRCh38
rs145292231 Chr.2: 55564332-55564332 on Build GRCh38
rs751807506 Chr.2: 55564390-55564390 on Build GRCh38

View Full Product Details