Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113996634-113996877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASA3
Entrez Gene ID
22821
Gene Name
RAS p21 protein activator 3
Gene Aliases
GAP1IP4BP, GAPIII
UniGene
Hs.593075
Gene Chromosome Location
Chr.13: 113977783-114132623 on Build GRCh38
Transcripts
NM_001320821.1 NM_001320822.1 NM_007368.3 XM_011534841.2 XM_011534842.2 XM_017020438.1 XM_017020439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4046263 c.1962G>A p.A654A Chr.13: 113996710-113996710 on Build GRCh38
6360763 c.2009A>T p.D670V Chr.13: 113996663-113996663 on Build GRCh38
945673 c.1961C>T p.A654V Chr.13: 113996711-113996711 on Build GRCh38
4440353 c.2028C>T p.S676S Chr.13: 113996644-113996644 on Build GRCh38
5454207 c.2036A>G p.N679S Chr.13: 113996636-113996636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021112309 Chr.13: 113996833-113996833 on Build GRCh38
rs377432039 Chr.13: 113996665-113996665 on Build GRCh38
rs371159351 Chr.13: 113996658-113996658 on Build GRCh38
rs778265586 Chr.13: 113996685-113996685 on Build GRCh38
rs775215983 Chr.13: 113996693-113996693 on Build GRCh38
rs139536887 Chr.13: 113996696-113996696 on Build GRCh38
rs893099627 Chr.13: 113996835-113996835 on Build GRCh38
rs185925768 Chr.13: 113996829-113996829 on Build GRCh38
rs555338628 Chr.13: 113996793-113996793 on Build GRCh38
rs750178304 Chr.13: 113996720-113996720 on Build GRCh38
rs1017727670 Chr.13: 113996802-113996802 on Build GRCh38
rs765180652 Chr.13: 113996711-113996711 on Build GRCh38
rs116482043 Chr.13: 113996788-113996788 on Build GRCh38
rs559714955 Chr.13: 113996826-113996826 on Build GRCh38
rs1030045003 Chr.13: 113996821-113996821 on Build GRCh38
rs748331268 Chr.13: 113996847-113996847 on Build GRCh38
rs147384608 Chr.13: 113996662-113996662 on Build GRCh38
rs1106101 Chr.13: 113996722-113996722 on Build GRCh38
rs781476890 Chr.13: 113996675-113996675 on Build GRCh38
rs781385097 Chr.13: 113996751-113996751 on Build GRCh38
rs766452063 Chr.13: 113996725-113996725 on Build GRCh38
rs1047650918 Chr.13: 113996791-113996791 on Build GRCh38
rs768398309 Chr.13: 113996769-113996770 on Build GRCh38
rs778459878 Chr.13: 113996769-113996769 on Build GRCh38
rs758499365 Chr.13: 113996659-113996659 on Build GRCh38
rs751525550 Chr.13: 113996664-113996664 on Build GRCh38
rs755220076 Chr.13: 113996750-113996750 on Build GRCh38
rs955541487 Chr.13: 113996827-113996827 on Build GRCh38
rs771586756 Chr.13: 113996692-113996692 on Build GRCh38
rs368147603 Chr.13: 113996782-113996782 on Build GRCh38
rs964399213 Chr.13: 113996814-113996814 on Build GRCh38
rs749689177 Chr.13: 113996686-113996686 on Build GRCh38
rs545196911 Chr.13: 113996830-113996830 on Build GRCh38
rs374142199 Chr.13: 113996703-113996703 on Build GRCh38
rs926628403 Chr.13: 113996756-113996756 on Build GRCh38
rs368364962 Chr.13: 113996744-113996744 on Build GRCh38
rs761751729 Chr.13: 113996774-113996775 on Build GRCh38
rs373745428 Chr.13: 113996794-113996794 on Build GRCh38
rs143664399 Chr.13: 113996707-113996707 on Build GRCh38
rs1028190545 Chr.13: 113996842-113996842 on Build GRCh38
rs751789471 Chr.13: 113996742-113996742 on Build GRCh38
rs564375106 Chr.13: 113996836-113996836 on Build GRCh38
rs111972201 Chr.13: 113996781-113996781 on Build GRCh38
rs540632406 Chr.13: 113996709-113996709 on Build GRCh38
rs557535624 Chr.13: 113996772-113996772 on Build GRCh38
rs752869210 Chr.13: 113996767-113996767 on Build GRCh38
rs979706264 Chr.13: 113996843-113996843 on Build GRCh38
rs7998472 Chr.13: 113996820-113996820 on Build GRCh38
rs182137769 Chr.13: 113996777-113996777 on Build GRCh38
rs770459445 Chr.13: 113996680-113996680 on Build GRCh38
rs372051906 Chr.13: 113996768-113996768 on Build GRCh38
rs748657787 Chr.13: 113996676-113996676 on Build GRCh38
rs761708403 Chr.13: 113996710-113996710 on Build GRCh38
rs776571055 Chr.13: 113996772-113996773 on Build GRCh38

View Full Product Details