Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123885194-123885467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM225
Entrez Gene ID
338661
Gene Name
transmembrane protein 225
Gene Aliases
PMP22CD, PPP1R154
UniGene
Hs.98377
Gene Chromosome Location
Chr.11: 123882920-123886763 on Build GRCh38
Transcripts
NM_001013743.2 XM_006718832.3 XM_011542802.2 XM_017017653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6803889 c.103G>T p.E35* Chr.11: 123885323-123885323 on Build GRCh38
4734479 c.64G>T p.V22F Chr.11: 123885362-123885362 on Build GRCh38
6803888 c.52T>C p.S18P Chr.11: 123885374-123885374 on Build GRCh38
686735 c.78G>A p.M26I Chr.11: 123885348-123885348 on Build GRCh38
686736 c.79G>A p.G27R Chr.11: 123885347-123885347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116452853 Chr.11: 123885350-123885350 on Build GRCh38
rs373708314 Chr.11: 123885423-123885423 on Build GRCh38
rs1035509901 Chr.11: 123885396-123885396 on Build GRCh38
rs781422550 Chr.11: 123885398-123885398 on Build GRCh38
rs200182919 Chr.11: 123885219-123885219 on Build GRCh38
rs755815456 Chr.11: 123885236-123885236 on Build GRCh38
rs769699845 Chr.11: 123885406-123885412 on Build GRCh38
rs779312082 Chr.11: 123885267-123885267 on Build GRCh38
rs375111550 Chr.11: 123885241-123885241 on Build GRCh38
rs115828609 Chr.11: 123885425-123885425 on Build GRCh38
rs988736808 Chr.11: 123885232-123885232 on Build GRCh38
rs149108305 Chr.11: 123885395-123885395 on Build GRCh38
rs758935223 Chr.11: 123885385-123885385 on Build GRCh38
rs948088998 Chr.11: 123885380-123885380 on Build GRCh38
rs753406271 Chr.11: 123885259-123885259 on Build GRCh38
rs906616254 Chr.11: 123885394-123885394 on Build GRCh38
rs756738262 Chr.11: 123885264-123885264 on Build GRCh38
rs370041522 Chr.11: 123885218-123885218 on Build GRCh38
rs893758522 Chr.11: 123885375-123885375 on Build GRCh38
rs963095028 Chr.11: 123885216-123885216 on Build GRCh38
rs1001808686 Chr.11: 123885275-123885276 on Build GRCh38
rs781332898 Chr.11: 123885329-123885329 on Build GRCh38
rs780380919 Chr.11: 123885297-123885297 on Build GRCh38
rs1044564126 Chr.11: 123885313-123885313 on Build GRCh38
rs761263754 Chr.11: 123885370-123885370 on Build GRCh38
rs116693665 Chr.11: 123885397-123885397 on Build GRCh38
rs775514173 Chr.11: 123885437-123885437 on Build GRCh38
rs145210692 Chr.11: 123885387-123885387 on Build GRCh38
rs771821395 Chr.11: 123885353-123885353 on Build GRCh38
rs750947267 Chr.11: 123885383-123885383 on Build GRCh38
rs748370038 Chr.11: 123885418-123885418 on Build GRCh38
rs769876191 Chr.11: 123885330-123885330 on Build GRCh38
rs769815254 Chr.11: 123885422-123885422 on Build GRCh38
rs760483112 Chr.11: 123885365-123885365 on Build GRCh38
rs760573106 Chr.11: 123885439-123885439 on Build GRCh38
rs753796924 Chr.11: 123885217-123885217 on Build GRCh38
rs766241363 Chr.11: 123885215-123885215 on Build GRCh38
rs142202385 Chr.11: 123885419-123885419 on Build GRCh38
rs564866134 Chr.11: 123885382-123885382 on Build GRCh38
rs914261550 Chr.11: 123885257-123885257 on Build GRCh38
rs750905300 Chr.11: 123885279-123885279 on Build GRCh38
rs866899641 Chr.11: 123885277-123885277 on Build GRCh38
rs774193780 Chr.11: 123885348-123885348 on Build GRCh38
rs747293317 Chr.11: 123885314-123885314 on Build GRCh38
rs752448179 Chr.11: 123885231-123885231 on Build GRCh38
rs772052922 Chr.11: 123885431-123885431 on Build GRCh38
rs768866977 Chr.11: 123885322-123885322 on Build GRCh38
rs775422018 Chr.11: 123885355-123885355 on Build GRCh38
rs745859884 Chr.11: 123885427-123885427 on Build GRCh38
rs369900040 Chr.11: 123885366-123885366 on Build GRCh38
rs868739873 Chr.11: 123885433-123885433 on Build GRCh38
rs758735814 Chr.11: 123885294-123885294 on Build GRCh38
rs994863631 Chr.11: 123885421-123885421 on Build GRCh38

View Full Product Details