Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 114845616-114845867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDST4
Entrez Gene ID
64579
Gene Name
N-deacetylase and N-sulfotransferase 4
Gene Aliases
N-HSST, N-HSST 4, NDST-4, NHSST4
UniGene
Hs.591700
Gene Chromosome Location
Chr.4: 114824122-115114098 on Build GRCh38
Transcripts
NM_022569.2 XM_017008545.1 XM_017008546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1732437 c.2072C>A p.T691N Chr.4: 114845866-114845866 on Build GRCh38
5535287 c.2096G>A p.R699K Chr.4: 114845842-114845842 on Build GRCh38
3825023 c.2091A>G p.S697S Chr.4: 114845847-114845847 on Build GRCh38
3993407 c.2076C>T p.I692I Chr.4: 114845862-114845862 on Build GRCh38
3409015 c.2078T>C p.L693P Chr.4: 114845860-114845860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59091642 Chr.4: 114845803-114845803 on Build GRCh38
rs10003387 Chr.4: 114845703-114845703 on Build GRCh38
rs35449662 Chr.4: 114845835-114845835 on Build GRCh38
rs938283148 Chr.4: 114845679-114845679 on Build GRCh38
rs779070902 Chr.4: 114845844-114845844 on Build GRCh38
rs374688610 Chr.4: 114845790-114845790 on Build GRCh38
rs766002967 Chr.4: 114845792-114845792 on Build GRCh38
rs373498943 Chr.4: 114845802-114845802 on Build GRCh38
rs531968719 Chr.4: 114845743-114845743 on Build GRCh38
rs1047073558 Chr.4: 114845677-114845677 on Build GRCh38
rs777754119 Chr.4: 114845807-114845807 on Build GRCh38
rs764757142 Chr.4: 114845774-114845774 on Build GRCh38
rs749226214 Chr.4: 114845817-114845817 on Build GRCh38
rs918250348 Chr.4: 114845655-114845655 on Build GRCh38
rs781646266 Chr.4: 114845826-114845827 on Build GRCh38
rs1029974879 Chr.4: 114845744-114845744 on Build GRCh38
rs770364005 Chr.4: 114845712-114845712 on Build GRCh38
rs1010955002 Chr.4: 114845702-114845702 on Build GRCh38
rs1055272185 Chr.4: 114845693-114845693 on Build GRCh38
rs745906541 Chr.4: 114845845-114845845 on Build GRCh38
rs768855557 Chr.4: 114845804-114845805 on Build GRCh38
rs968433586 Chr.4: 114845775-114845775 on Build GRCh38
rs543526207 Chr.4: 114845747-114845747 on Build GRCh38
rs1030128337 Chr.4: 114845694-114845695 on Build GRCh38
rs747565469 Chr.4: 114845787-114845788 on Build GRCh38
rs949782203 Chr.4: 114845656-114845656 on Build GRCh38
rs985793693 Chr.4: 114845787-114845787 on Build GRCh38
rs777275295 Chr.4: 114845782-114845782 on Build GRCh38
rs145578814 Chr.4: 114845664-114845664 on Build GRCh38
rs906809635 Chr.4: 114845678-114845678 on Build GRCh38
rs200004349 Chr.4: 114845798-114845798 on Build GRCh38
rs774635464 Chr.4: 114845810-114845810 on Build GRCh38
rs893989193 Chr.4: 114845699-114845699 on Build GRCh38
rs750989469 Chr.4: 114845793-114845793 on Build GRCh38

View Full Product Details