Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85349261-85349534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RETSAT
Entrez Gene ID
54884
Gene Name
retinol saturase
Gene Aliases
-
UniGene
Hs.440401
Gene Chromosome Location
Chr.2: 85341955-85354698 on Build GRCh38
Transcripts
NM_017750.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5177137 c.884delG p.G295fs*12 Chr.2: 85349497-85349497 on Build GRCh38
6765098 c.925C>T p.R309W Chr.2: 85349456-85349456 on Build GRCh38
4511713 c.877C>T p.R293* Chr.2: 85349504-85349504 on Build GRCh38
6019812 c.963G>C p.Q321H Chr.2: 85349418-85349418 on Build GRCh38
6159036 c.878G>T p.R293L Chr.2: 85349503-85349503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755144692 Chr.2: 85349365-85349365 on Build GRCh38
rs750548924 Chr.2: 85349337-85349337 on Build GRCh38
rs770494557 Chr.2: 85349389-85349389 on Build GRCh38
rs572229299 Chr.2: 85349305-85349305 on Build GRCh38
rs779454072 Chr.2: 85349451-85349451 on Build GRCh38
rs952996219 Chr.2: 85349428-85349428 on Build GRCh38
rs373458754 Chr.2: 85349333-85349333 on Build GRCh38
rs762604028 Chr.2: 85349429-85349429 on Build GRCh38
rs756201218 Chr.2: 85349351-85349351 on Build GRCh38
rs1008219765 Chr.2: 85349284-85349284 on Build GRCh38
rs372113449 Chr.2: 85349441-85349441 on Build GRCh38
rs747573438 Chr.2: 85349478-85349478 on Build GRCh38
rs769507148 Chr.2: 85349501-85349501 on Build GRCh38
rs774278780 Chr.2: 85349505-85349505 on Build GRCh38
rs755270394 Chr.2: 85349445-85349445 on Build GRCh38
rs138110856 Chr.2: 85349456-85349456 on Build GRCh38
rs778965663 Chr.2: 85349448-85349448 on Build GRCh38
rs867466240 Chr.2: 85349473-85349473 on Build GRCh38
rs143604495 Chr.2: 85349455-85349455 on Build GRCh38
rs375130450 Chr.2: 85349444-85349444 on Build GRCh38
rs761893901 Chr.2: 85349506-85349506 on Build GRCh38
rs768718814 Chr.2: 85349503-85349503 on Build GRCh38
rs761842652 Chr.2: 85349439-85349439 on Build GRCh38
rs919998503 Chr.2: 85349353-85349353 on Build GRCh38
rs372505970 Chr.2: 85349421-85349421 on Build GRCh38
rs923590852 Chr.2: 85349474-85349474 on Build GRCh38
rs148892494 Chr.2: 85349367-85349367 on Build GRCh38
rs780432401 Chr.2: 85349355-85349355 on Build GRCh38
rs780793300 Chr.2: 85349493-85349493 on Build GRCh38
rs181584954 Chr.2: 85349499-85349499 on Build GRCh38
rs757739295 Chr.2: 85349483-85349483 on Build GRCh38
rs143670128 Chr.2: 85349504-85349504 on Build GRCh38
rs986574163 Chr.2: 85349496-85349496 on Build GRCh38
rs373847804 Chr.2: 85349397-85349397 on Build GRCh38
rs778204911 Chr.2: 85349384-85349384 on Build GRCh38
rs961512775 Chr.2: 85349324-85349324 on Build GRCh38
rs559661769 Chr.2: 85349508-85349508 on Build GRCh38
rs755034233 Chr.2: 85349359-85349368 on Build GRCh38
rs753129634 Chr.2: 85349449-85349449 on Build GRCh38
rs764137514 Chr.2: 85349430-85349430 on Build GRCh38
rs112528661 Chr.2: 85349461-85349461 on Build GRCh38
rs978352379 Chr.2: 85349386-85349386 on Build GRCh38
rs772667649 Chr.2: 85349371-85349371 on Build GRCh38
rs769297429 Chr.2: 85349417-85349417 on Build GRCh38
rs202200883 Chr.2: 85349396-85349396 on Build GRCh38
rs10590946 Chr.2: 85349329-85349330 on Build GRCh38
rs376359072 Chr.2: 85349360-85349360 on Build GRCh38
rs955268640 Chr.2: 85349468-85349468 on Build GRCh38
rs775384205 Chr.2: 85349424-85349424 on Build GRCh38
rs748432530 Chr.2: 85349368-85349368 on Build GRCh38
rs751471213 Chr.2: 85349434-85349434 on Build GRCh38

View Full Product Details