Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56994868-56995137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCPEP1
Entrez Gene ID
59342
Gene Name
serine carboxypeptidase 1
Gene Aliases
HSCP1, RISC
UniGene
Hs.514950
Gene Chromosome Location
Chr.17: 56978107-57006768 on Build GRCh38
Transcripts
NM_021626.2 XM_005257557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3194718 c.625G>T p.V209L Chr.17: 56994986-56994986 on Build GRCh38
5451450 c.623C>T p.S208L Chr.17: 56994984-56994984 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs549822059 Chr.17: 56994900-56994900 on Build GRCh38
rs766900856 Chr.17: 56994936-56994936 on Build GRCh38
rs140957167 Chr.17: 56994988-56994988 on Build GRCh38
rs969304311 Chr.17: 56994918-56994918 on Build GRCh38
rs536508221 Chr.17: 56995051-56995051 on Build GRCh38
rs756927792 Chr.17: 56994954-56994954 on Build GRCh38
rs763072102 Chr.17: 56995028-56995028 on Build GRCh38
rs922122396 Chr.17: 56995017-56995017 on Build GRCh38
rs781543538 Chr.17: 56994946-56994946 on Build GRCh38
rs766386580 Chr.17: 56995030-56995030 on Build GRCh38
rs78357658 Chr.17: 56994967-56994967 on Build GRCh38
rs756511445 Chr.17: 56995059-56995059 on Build GRCh38
rs1019260258 Chr.17: 56994975-56994975 on Build GRCh38
rs913496744 Chr.17: 56995095-56995095 on Build GRCh38
rs1023395817 Chr.17: 56994903-56994903 on Build GRCh38
rs765522167 Chr.17: 56995016-56995016 on Build GRCh38
rs746924387 Chr.17: 56994985-56994985 on Build GRCh38
rs185432213 Chr.17: 56994948-56994948 on Build GRCh38
rs768456536 Chr.17: 56994986-56994986 on Build GRCh38
rs951352830 Chr.17: 56994941-56994941 on Build GRCh38
rs77100318 Chr.17: 56995053-56995053 on Build GRCh38
rs745353594 Chr.17: 56994978-56994978 on Build GRCh38
rs755472721 Chr.17: 56995040-56995040 on Build GRCh38
rs773631202 Chr.17: 56995026-56995026 on Build GRCh38
rs767823649 Chr.17: 56995042-56995042 on Build GRCh38
rs752042308 Chr.17: 56995039-56995039 on Build GRCh38
rs151035266 Chr.17: 56994914-56994914 on Build GRCh38
rs192931764 Chr.17: 56994937-56994937 on Build GRCh38
rs201685402 Chr.17: 56994974-56994974 on Build GRCh38
rs755315457 Chr.17: 56994942-56994942 on Build GRCh38
rs559901448 Chr.17: 56994898-56994898 on Build GRCh38
rs775439550 Chr.17: 56994984-56994984 on Build GRCh38
rs150266367 Chr.17: 56995074-56995074 on Build GRCh38
rs373428304 Chr.17: 56995062-56995062 on Build GRCh38
rs745520219 Chr.17: 56995066-56995066 on Build GRCh38
rs774260876 Chr.17: 56994949-56994950 on Build GRCh38
rs761746587 Chr.17: 56995001-56995001 on Build GRCh38
rs771549769 Chr.17: 56994980-56994980 on Build GRCh38

View Full Product Details