Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121038825-121039070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OASL
Entrez Gene ID
8638
Gene Name
2'-5'-oligoadenylate synthetase like
Gene Aliases
OASLd, TRIP-14, TRIP14, p59 OASL, p59-OASL, p59OASL
UniGene
Hs.118633
Gene Chromosome Location
Chr.12: 121018882-121039242 on Build GRCh38
Transcripts
NM_001261825.1 NM_003733.3 NM_198213.2 XM_017020140.1 XM_017020141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
935993 c.30A>G p.T10T Chr.12: 121038942-121038942 on Build GRCh38
1605662 c.23A>G p.Y8C Chr.12: 121038949-121038949 on Build GRCh38
4039436 c.27C>T p.S9S Chr.12: 121038945-121038945 on Build GRCh38
3811244 c.121G>C p.E41Q Chr.12: 121038851-121038851 on Build GRCh38
7099663 c.54C>T p.F18F Chr.12: 121038918-121038918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779402321 Chr.12: 121038990-121038990 on Build GRCh38
rs779410044 Chr.12: 121038864-121038864 on Build GRCh38
rs951355987 Chr.12: 121038909-121038909 on Build GRCh38
rs778378386 Chr.12: 121038980-121038980 on Build GRCh38
rs768609474 Chr.12: 121039015-121039015 on Build GRCh38
rs746049557 Chr.12: 121038907-121038907 on Build GRCh38
rs776432126 Chr.12: 121038930-121038930 on Build GRCh38
rs140436299 Chr.12: 121038922-121038922 on Build GRCh38
rs77356806 Chr.12: 121038989-121038989 on Build GRCh38
rs989453589 Chr.12: 121038914-121038914 on Build GRCh38
rs559152850 Chr.12: 121039034-121039034 on Build GRCh38
rs201649356 Chr.12: 121038854-121038854 on Build GRCh38
rs200293050 Chr.12: 121038867-121038867 on Build GRCh38
rs186524186 Chr.12: 121038880-121038880 on Build GRCh38
rs771303424 Chr.12: 121038892-121038892 on Build GRCh38
rs754289456 Chr.12: 121038986-121038986 on Build GRCh38
rs372213456 Chr.12: 121038971-121038971 on Build GRCh38
rs138743509 Chr.12: 121038890-121038890 on Build GRCh38
rs757965757 Chr.12: 121038857-121038857 on Build GRCh38
rs189679788 Chr.12: 121039014-121039014 on Build GRCh38
rs199714834 Chr.12: 121038925-121038925 on Build GRCh38
rs753560565 Chr.12: 121038848-121038848 on Build GRCh38
rs934147632 Chr.12: 121038918-121038918 on Build GRCh38
rs767245758 Chr.12: 121038945-121038945 on Build GRCh38
rs566075133 Chr.12: 121038946-121038946 on Build GRCh38
rs201238602 Chr.12: 121038866-121038866 on Build GRCh38
rs775423659 Chr.12: 121038917-121038917 on Build GRCh38
rs144785999 Chr.12: 121038855-121038855 on Build GRCh38
rs368849271 Chr.12: 121038951-121038951 on Build GRCh38
rs765981728 Chr.12: 121038956-121038956 on Build GRCh38
rs774679549 Chr.12: 121038893-121038893 on Build GRCh38
rs75191997 Chr.12: 121039007-121039007 on Build GRCh38
rs921453145 Chr.12: 121039024-121039024 on Build GRCh38
rs35632880 Chr.12: 121038953-121038953 on Build GRCh38
rs975118227 Chr.12: 121039010-121039010 on Build GRCh38
rs201179685 Chr.12: 121038860-121038860 on Build GRCh38
rs746164611 Chr.12: 121039001-121039001 on Build GRCh38
rs1047415721 Chr.12: 121039040-121039040 on Build GRCh38
rs761554855 Chr.12: 121038940-121038940 on Build GRCh38
rs772115794 Chr.12: 121039005-121039005 on Build GRCh38
rs756660060 Chr.12: 121038972-121038972 on Build GRCh38
rs377444845 Chr.12: 121038849-121038849 on Build GRCh38
rs768785257 Chr.12: 121038981-121038981 on Build GRCh38
rs71454673 Chr.12: 121039043-121039043 on Build GRCh38
rs929687971 Chr.12: 121039028-121039028 on Build GRCh38
rs74436726 Chr.12: 121038884-121038884 on Build GRCh38
rs772249079 Chr.12: 121038911-121038911 on Build GRCh38

View Full Product Details