Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144129357-144129630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2A14
Entrez Gene ID
135941
Gene Name
olfactory receptor family 2 subfamily A member 14
Gene Aliases
OR2A14P, OR2A6, OST182
UniGene
Hs.534547
Gene Chromosome Location
Chr.7: 144129113-144130045 on Build GRCh38
Transcripts
NM_001001659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673626 c.287G>A p.C96Y Chr.7: 144129399-144129399 on Build GRCh38
4415797 c.492C>G p.S164R Chr.7: 144129604-144129604 on Build GRCh38
5699651 c.248A>G p.N83S Chr.7: 144129360-144129360 on Build GRCh38
293657 c.381delC p.L129fs*7 Chr.7: 144129493-144129493 on Build GRCh38
745087 c.334C>T p.L112L Chr.7: 144129446-144129446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756734262 Chr.7: 144129468-144129468 on Build GRCh38
rs745789380 Chr.7: 144129575-144129575 on Build GRCh38
rs756578197 Chr.7: 144129472-144129472 on Build GRCh38
rs781721094 Chr.7: 144129413-144129413 on Build GRCh38
rs201784448 Chr.7: 144129494-144129494 on Build GRCh38
rs780530168 Chr.7: 144129541-144129542 on Build GRCh38
rs199549254 Chr.7: 144129437-144129437 on Build GRCh38
rs769869666 Chr.7: 144129580-144129580 on Build GRCh38
rs778169852 Chr.7: 144129397-144129397 on Build GRCh38
rs375928210 Chr.7: 144129500-144129500 on Build GRCh38
rs369022856 Chr.7: 144129481-144129481 on Build GRCh38
rs750769131 Chr.7: 144129467-144129467 on Build GRCh38
rs767730530 Chr.7: 144129595-144129595 on Build GRCh38
rs200056281 Chr.7: 144129444-144129444 on Build GRCh38
rs746385754 Chr.7: 144129420-144129420 on Build GRCh38
rs1027163962 Chr.7: 144129386-144129386 on Build GRCh38
rs767983743 Chr.7: 144129461-144129461 on Build GRCh38
rs559068119 Chr.7: 144129436-144129436 on Build GRCh38
rs995009672 Chr.7: 144129450-144129451 on Build GRCh38
rs756457793 Chr.7: 144129530-144129530 on Build GRCh38
rs77933816 Chr.7: 144129585-144129585 on Build GRCh38
rs925886303 Chr.7: 144129559-144129559 on Build GRCh38
rs908180081 Chr.7: 144129421-144129421 on Build GRCh38
rs200949224 Chr.7: 144129607-144129607 on Build GRCh38
rs767641163 Chr.7: 144129527-144129527 on Build GRCh38
rs895824157 Chr.7: 144129605-144129605 on Build GRCh38
rs766638381 Chr.7: 144129610-144129610 on Build GRCh38
rs754463801 Chr.7: 144129483-144129483 on Build GRCh38
rs372050427 Chr.7: 144129440-144129440 on Build GRCh38
rs770222923 Chr.7: 144129426-144129427 on Build GRCh38
rs771723186 Chr.7: 144129493-144129493 on Build GRCh38
rs561589216 Chr.7: 144129561-144129561 on Build GRCh38
rs777309629 Chr.7: 144129556-144129556 on Build GRCh38
rs764268400 Chr.7: 144129514-144129514 on Build GRCh38
rs751804602 Chr.7: 144129457-144129457 on Build GRCh38
rs757623646 Chr.7: 144129460-144129460 on Build GRCh38
rs2961161 Chr.7: 144129604-144129604 on Build GRCh38
rs978613997 Chr.7: 144129548-144129548 on Build GRCh38
rs766228978 Chr.7: 144129445-144129445 on Build GRCh38
rs770597478 Chr.7: 144129560-144129560 on Build GRCh38
rs746722616 Chr.7: 144129557-144129557 on Build GRCh38
rs762073663 Chr.7: 144129517-144129517 on Build GRCh38
rs372060689 Chr.7: 144129425-144129425 on Build GRCh38
rs945266518 Chr.7: 144129465-144129465 on Build GRCh38
rs750760277 Chr.7: 144129528-144129528 on Build GRCh38
rs773286195 Chr.7: 144129598-144129598 on Build GRCh38
rs778460650 Chr.7: 144129489-144129489 on Build GRCh38
rs545498324 Chr.7: 144129454-144129454 on Build GRCh38
rs950981031 Chr.7: 144129458-144129458 on Build GRCh38
rs267601377 Chr.7: 144129455-144129455 on Build GRCh38
rs747785648 Chr.7: 144129490-144129490 on Build GRCh38
rs377028134 Chr.7: 144129429-144129429 on Build GRCh38
rs376721058 Chr.7: 144129473-144129473 on Build GRCh38
rs749707842 Chr.7: 144129571-144129571 on Build GRCh38
rs894518945 Chr.7: 144129542-144129542 on Build GRCh38
rs759583275 Chr.7: 144129512-144129512 on Build GRCh38
rs186823699 Chr.7: 144129555-144129555 on Build GRCh38
rs2961160 Chr.7: 144129510-144129510 on Build GRCh38
rs757670540 Chr.7: 144129401-144129401 on Build GRCh38
rs755535845 Chr.7: 144129547-144129547 on Build GRCh38
rs780459337 Chr.7: 144129480-144129480 on Build GRCh38
rs779561390 Chr.7: 144129427-144129427 on Build GRCh38
rs774632747 Chr.7: 144129516-144129516 on Build GRCh38
rs751920461 Chr.7: 144129398-144129398 on Build GRCh38
rs764315381 Chr.7: 144129456-144129456 on Build GRCh38
rs772936096 Chr.7: 144129443-144129443 on Build GRCh38
rs778336071 Chr.7: 144129554-144129554 on Build GRCh38
rs754337358 Chr.7: 144129545-144129545 on Build GRCh38

View Full Product Details