Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20362353-20362624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDILT
Entrez Gene ID
204474
Gene Name
protein disulfide isomerase like, testis expressed
Gene Aliases
PDIA7
UniGene
Hs.376025
Gene Chromosome Location
Chr.16: 20359170-20404742 on Build GRCh38
Transcripts
NM_174924.1 XM_011545764.1 XM_011545765.1 XM_011545766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7230112 c.1336G>A p.D446N Chr.16: 20362484-20362484 on Build GRCh38
5434084 c.1357C>G p.Q453E Chr.16: 20362463-20362463 on Build GRCh38
369912 c.1371G>A p.L457L Chr.16: 20362449-20362449 on Build GRCh38
5906030 c.1400C>T p.P467L Chr.16: 20362420-20362420 on Build GRCh38
257702 c.1386C>A p.F462L Chr.16: 20362434-20362434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024699788 Chr.16: 20362436-20362436 on Build GRCh38
rs11648131 Chr.16: 20362484-20362484 on Build GRCh38
rs772390274 Chr.16: 20362602-20362602 on Build GRCh38
rs779028833 Chr.16: 20362598-20362598 on Build GRCh38
rs753450940 Chr.16: 20362440-20362440 on Build GRCh38
rs746056355 Chr.16: 20362599-20362599 on Build GRCh38
rs763137688 Chr.16: 20362405-20362405 on Build GRCh38
rs758283687 Chr.16: 20362449-20362449 on Build GRCh38
rs201187995 Chr.16: 20362415-20362415 on Build GRCh38
rs748202857 Chr.16: 20362396-20362396 on Build GRCh38
rs750176775 Chr.16: 20362445-20362445 on Build GRCh38
rs370774858 Chr.16: 20362533-20362533 on Build GRCh38
rs769750254 Chr.16: 20362397-20362397 on Build GRCh38
rs1023305091 Chr.16: 20362512-20362512 on Build GRCh38
rs770981068 Chr.16: 20362596-20362596 on Build GRCh38
rs367692063 Chr.16: 20362555-20362555 on Build GRCh38
rs772316367 Chr.16: 20362492-20362492 on Build GRCh38
rs768685735 Chr.16: 20362471-20362471 on Build GRCh38
rs753361388 Chr.16: 20362515-20362515 on Build GRCh38
rs775966813 Chr.16: 20362499-20362499 on Build GRCh38
rs139748181 Chr.16: 20362577-20362577 on Build GRCh38
rs139805278 Chr.16: 20362444-20362444 on Build GRCh38
rs372901536 Chr.16: 20362386-20362386 on Build GRCh38
rs139363566 Chr.16: 20362485-20362485 on Build GRCh38
rs775294775 Chr.16: 20362390-20362390 on Build GRCh38
rs987530983 Chr.16: 20362520-20362520 on Build GRCh38
rs898443160 Chr.16: 20362583-20362586 on Build GRCh38
rs192834336 Chr.16: 20362413-20362413 on Build GRCh38
rs779690891 Chr.16: 20362542-20362542 on Build GRCh38
rs150026846 Chr.16: 20362519-20362519 on Build GRCh38
rs189278427 Chr.16: 20362504-20362504 on Build GRCh38
rs541369003 Chr.16: 20362579-20362579 on Build GRCh38
rs763484471 Chr.16: 20362438-20362438 on Build GRCh38
rs751390432 Chr.16: 20362548-20362548 on Build GRCh38
rs753129109 Chr.16: 20362424-20362424 on Build GRCh38
rs758137761 Chr.16: 20362534-20362534 on Build GRCh38
rs779958356 Chr.16: 20362466-20362466 on Build GRCh38
rs112178426 Chr.16: 20362385-20362385 on Build GRCh38
rs11865916 Chr.16: 20362481-20362481 on Build GRCh38
rs759809629 Chr.16: 20362488-20362488 on Build GRCh38
rs761308279 Chr.16: 20362427-20362427 on Build GRCh38
rs988589133 Chr.16: 20362509-20362509 on Build GRCh38
rs764924755 Chr.16: 20362530-20362530 on Build GRCh38
rs145948559 Chr.16: 20362416-20362416 on Build GRCh38
rs746889157 Chr.16: 20362468-20362468 on Build GRCh38
rs761067553 Chr.16: 20362501-20362501 on Build GRCh38
rs764688950 Chr.16: 20362508-20362508 on Build GRCh38
rs756925042 Chr.16: 20362443-20362443 on Build GRCh38
rs944950477 Chr.16: 20362428-20362428 on Build GRCh38

View Full Product Details