Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78655932-78656443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TENM4
Entrez Gene ID
26011
Gene Name
teneurin transmembrane protein 4
Gene Aliases
Doc4, ETM5, ODZ4, TNM4, Ten-M4
UniGene
Hs.213087
Gene Chromosome Location
Chr.11: 78653283-79441019 on Build GRCh38
Transcripts
NM_001098816.2 XM_011544933.2 XM_017017525.1 XM_017017526.1

SNP Detail

SNP ID Chromosome Location
rs1032948316 Chr.11: 78656349-78656349 on Build GRCh38
rs990567038 Chr.11: 78656018-78656018 on Build GRCh38
rs938208110 Chr.11: 78656017-78656017 on Build GRCh38
rs899226732 Chr.11: 78656358-78656358 on Build GRCh38
rs374634871 Chr.11: 78656087-78656087 on Build GRCh38
rs1029580621 Chr.11: 78656404-78656404 on Build GRCh38
rs1038615102 Chr.11: 78656350-78656350 on Build GRCh38
rs886421699 Chr.11: 78656373-78656373 on Build GRCh38
rs76567808 Chr.11: 78656369-78656369 on Build GRCh38
rs541827780 Chr.11: 78656242-78656242 on Build GRCh38
rs1051003479 Chr.11: 78656296-78656296 on Build GRCh38
rs571267434 Chr.11: 78656033-78656033 on Build GRCh38
rs547831187 Chr.11: 78656340-78656340 on Build GRCh38
rs950917174 Chr.11: 78656105-78656105 on Build GRCh38
rs905683595 Chr.11: 78656307-78656307 on Build GRCh38
rs959565832 Chr.11: 78656146-78656146 on Build GRCh38
rs77611857 Chr.11: 78656096-78656096 on Build GRCh38
rs1051342425 Chr.11: 78656071-78656071 on Build GRCh38
rs891687518 Chr.11: 78656359-78656359 on Build GRCh38
rs1022763038 Chr.11: 78656061-78656061 on Build GRCh38
rs771098158 Chr.11: 78656395-78656395 on Build GRCh38
rs920280113 Chr.11: 78656228-78656228 on Build GRCh38
rs774312834 Chr.11: 78656100-78656100 on Build GRCh38
rs1017682758 Chr.11: 78656092-78656092 on Build GRCh38
rs12270560 Chr.11: 78656203-78656203 on Build GRCh38
rs561705452 Chr.11: 78656272-78656272 on Build GRCh38
rs1037810038 Chr.11: 78656008-78656008 on Build GRCh38
rs928973663 Chr.11: 78656231-78656231 on Build GRCh38
rs34086370 Chr.11: 78656275-78656276 on Build GRCh38
rs941640609 Chr.11: 78656199-78656199 on Build GRCh38
rs77841734 Chr.11: 78656018-78656018 on Build GRCh38
rs772563492 Chr.11: 78656159-78656159 on Build GRCh38
rs955496837 Chr.11: 78656407-78656407 on Build GRCh38
rs370397073 Chr.11: 78656134-78656134 on Build GRCh38
rs552795927 Chr.11: 78655962-78655962 on Build GRCh38
rs147573744 Chr.11: 78656039-78656039 on Build GRCh38
rs895575727 Chr.11: 78656249-78656249 on Build GRCh38
rs911472815 Chr.11: 78655968-78655968 on Build GRCh38
rs982666404 Chr.11: 78655997-78655997 on Build GRCh38
rs899257656 Chr.11: 78656095-78656095 on Build GRCh38
rs576924916 Chr.11: 78656088-78656088 on Build GRCh38
rs771171558 Chr.11: 78656076-78656076 on Build GRCh38
rs982372724 Chr.11: 78656107-78656107 on Build GRCh38
rs923620826 Chr.11: 78656235-78656235 on Build GRCh38
rs1029717959 Chr.11: 78656097-78656097 on Build GRCh38
rs928168380 Chr.11: 78656017-78656018 on Build GRCh38
rs1026536505 Chr.11: 78655957-78655957 on Build GRCh38
rs889738995 Chr.11: 78656329-78656329 on Build GRCh38
rs895544422 Chr.11: 78656043-78656043 on Build GRCh38
rs1000544521 Chr.11: 78656332-78656332 on Build GRCh38
rs911390345 Chr.11: 78656153-78656153 on Build GRCh38
rs562740748 Chr.11: 78656164-78656164 on Build GRCh38
rs569223923 Chr.11: 78655996-78655996 on Build GRCh38
rs183816671 Chr.11: 78656049-78656049 on Build GRCh38
rs949790444 Chr.11: 78656252-78656252 on Build GRCh38
rs955424605 Chr.11: 78656115-78656115 on Build GRCh38
rs553683402 Chr.11: 78656079-78656079 on Build GRCh38
rs902398376 Chr.11: 78656048-78656048 on Build GRCh38
rs991428505 Chr.11: 78656158-78656158 on Build GRCh38

View Full Product Details