Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6621154-6621600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAR5
Entrez Gene ID
57121
Gene Name
lysophosphatidic acid receptor 5
Gene Aliases
GPR92, GPR93, KPG_010, LPA5
UniGene
Hs.155538
Gene Chromosome Location
Chr.12: 6618835-6636131 on Build GRCh38
Transcripts
NM_001142961.1 NM_020400.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770322 c.56G>A p.R19Q Chr.12: 6621193-6621193 on Build GRCh38
4968225 c.34G>T p.V12F Chr.12: 6621215-6621215 on Build GRCh38
1256626 c.55C>T p.R19* Chr.12: 6621194-6621194 on Build GRCh38
7426767 c.79G>C p.V27L Chr.12: 6621170-6621170 on Build GRCh38
942778 c.29G>T p.S10I Chr.12: 6621220-6621220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12303174 Chr.12: 6621271-6621271 on Build GRCh38
rs780724716 Chr.12: 6621220-6621220 on Build GRCh38
rs1001884936 Chr.12: 6621513-6621513 on Build GRCh38
rs752400173 Chr.12: 6621194-6621194 on Build GRCh38
rs766491986 Chr.12: 6621225-6621225 on Build GRCh38
rs1014828257 Chr.12: 6621417-6621417 on Build GRCh38
rs767795581 Chr.12: 6621438-6621438 on Build GRCh38
rs935753140 Chr.12: 6621554-6621554 on Build GRCh38
rs766349066 Chr.12: 6621180-6621180 on Build GRCh38
rs766255531 Chr.12: 6621281-6621281 on Build GRCh38
rs754791280 Chr.12: 6621182-6621182 on Build GRCh38
rs140954912 Chr.12: 6621238-6621238 on Build GRCh38
rs987104754 Chr.12: 6621520-6621520 on Build GRCh38
rs769981302 Chr.12: 6621241-6621241 on Build GRCh38
rs116264317 Chr.12: 6621268-6621268 on Build GRCh38
rs773315144 Chr.12: 6621264-6621264 on Build GRCh38
rs755778258 Chr.12: 6621200-6621200 on Build GRCh38
rs974390253 Chr.12: 6621532-6621532 on Build GRCh38
rs552596152 Chr.12: 6621378-6621378 on Build GRCh38
rs896023264 Chr.12: 6621409-6621409 on Build GRCh38
rs151060687 Chr.12: 6621226-6621226 on Build GRCh38
rs568397306 Chr.12: 6621529-6621529 on Build GRCh38
rs773276160 Chr.12: 6621278-6621278 on Build GRCh38
rs777331197 Chr.12: 6621203-6621203 on Build GRCh38
rs528462096 Chr.12: 6621430-6621430 on Build GRCh38
rs113415869 Chr.12: 6621464-6621464 on Build GRCh38
rs751525562 Chr.12: 6621325-6621325 on Build GRCh38
rs932686048 Chr.12: 6621371-6621371 on Build GRCh38
rs772683804 Chr.12: 6621210-6621210 on Build GRCh38
rs989866092 Chr.12: 6621567-6621567 on Build GRCh38
rs951594777 Chr.12: 6621454-6621454 on Build GRCh38
rs976898163 Chr.12: 6621314-6621314 on Build GRCh38
rs569551081 Chr.12: 6621279-6621279 on Build GRCh38
rs184483155 Chr.12: 6621250-6621250 on Build GRCh38
rs780207538 Chr.12: 6621381-6621381 on Build GRCh38
rs907242933 Chr.12: 6621351-6621351 on Build GRCh38
rs940586299 Chr.12: 6621395-6621396 on Build GRCh38
rs912803458 Chr.12: 6621526-6621526 on Build GRCh38
rs1036300615 Chr.12: 6621396-6621411 on Build GRCh38
rs943866667 Chr.12: 6621310-6621310 on Build GRCh38
rs748788177 Chr.12: 6621208-6621208 on Build GRCh38
rs1034425658 Chr.12: 6621384-6621384 on Build GRCh38
rs548497441 Chr.12: 6621446-6621446 on Build GRCh38
rs368795459 Chr.12: 6621181-6621181 on Build GRCh38
rs747333675 Chr.12: 6621224-6621224 on Build GRCh38
rs752940917 Chr.12: 6621471-6621471 on Build GRCh38
rs1046054905 Chr.12: 6621336-6621336 on Build GRCh38
rs776657231 Chr.12: 6621232-6621232 on Build GRCh38
rs142475411 Chr.12: 6621398-6621398 on Build GRCh38
rs762527361 Chr.12: 6621494-6621495 on Build GRCh38
rs200022683 Chr.12: 6621553-6621553 on Build GRCh38

View Full Product Details