Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52592305-52592808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT72
Entrez Gene ID
140807
Gene Name
keratin 72
Gene Aliases
K6IRS2, K6irs, KRT6, KRT6IRS2
UniGene
Hs.662013
Gene Chromosome Location
Chr.12: 52585589-52602900 on Build GRCh38
Transcripts
NM_001146225.1 NM_001146226.1 NM_080747.2 XM_017018833.1 XM_017018834.1 XM_017018835.1 XM_017018836.1 XM_017018837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5618264 c.791A>G p.Y264C Chr.12: 52592403-52592403 on Build GRCh38
7085198 c.793G>A p.E265K Chr.12: 52592401-52592401 on Build GRCh38
237643 c.798+6G>A p.? Chr.12: 52592390-52592390 on Build GRCh38
4792928 c.712G>A p.A238T Chr.12: 52592482-52592482 on Build GRCh38
4696865 c.727A>G p.K243E Chr.12: 52592467-52592467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006458210 Chr.12: 52592643-52592643 on Build GRCh38
rs935100767 Chr.12: 52592561-52592561 on Build GRCh38
rs369263271 Chr.12: 52592515-52592515 on Build GRCh38
rs199651611 Chr.12: 52592410-52592410 on Build GRCh38
rs761427108 Chr.12: 52592475-52592475 on Build GRCh38
rs377500272 Chr.12: 52592488-52592488 on Build GRCh38
rs772283468 Chr.12: 52592538-52592538 on Build GRCh38
rs534777680 Chr.12: 52592570-52592570 on Build GRCh38
rs766999091 Chr.12: 52592395-52592395 on Build GRCh38
rs537053362 Chr.12: 52592784-52592784 on Build GRCh38
rs747538608 Chr.12: 52592674-52592674 on Build GRCh38
rs74572393 Chr.12: 52592487-52592487 on Build GRCh38
rs773921666 Chr.12: 52592384-52592384 on Build GRCh38
rs373893434 Chr.12: 52592651-52592651 on Build GRCh38
rs1003513413 Chr.12: 52592527-52592527 on Build GRCh38
rs886318029 Chr.12: 52592477-52592477 on Build GRCh38
rs775921357 Chr.12: 52592352-52592352 on Build GRCh38
rs1034855703 Chr.12: 52592508-52592508 on Build GRCh38
rs374695100 Chr.12: 52592658-52592658 on Build GRCh38
rs776011221 Chr.12: 52592447-52592447 on Build GRCh38
rs764467430 Chr.12: 52592376-52592376 on Build GRCh38
rs617820 Chr.12: 52592348-52592348 on Build GRCh38
rs1035812611 Chr.12: 52592680-52592680 on Build GRCh38
rs141997666 Chr.12: 52592706-52592706 on Build GRCh38
rs769801679 Chr.12: 52592445-52592445 on Build GRCh38
rs957100728 Chr.12: 52592360-52592360 on Build GRCh38
rs781191778 Chr.12: 52592433-52592433 on Build GRCh38
rs778749490 Chr.12: 52592422-52592422 on Build GRCh38
rs1029624372 Chr.12: 52592454-52592454 on Build GRCh38
rs188560376 Chr.12: 52592565-52592565 on Build GRCh38
rs979537176 Chr.12: 52592544-52592544 on Build GRCh38
rs12315878 Chr.12: 52592489-52592489 on Build GRCh38
rs888048025 Chr.12: 52592629-52592629 on Build GRCh38
rs755627285 Chr.12: 52592398-52592398 on Build GRCh38
rs749939528 Chr.12: 52592396-52592396 on Build GRCh38
rs147557252 Chr.12: 52592650-52592650 on Build GRCh38
rs758073847 Chr.12: 52592497-52592497 on Build GRCh38
rs756233345 Chr.12: 52592506-52592506 on Build GRCh38
rs746445264 Chr.12: 52592347-52592347 on Build GRCh38
rs747236825 Chr.12: 52592429-52592429 on Build GRCh38
rs749771207 Chr.12: 52592463-52592463 on Build GRCh38
rs746052399 Chr.12: 52592503-52592503 on Build GRCh38
rs1037563108 Chr.12: 52592752-52592752 on Build GRCh38
rs138477551 Chr.12: 52592480-52592480 on Build GRCh38
rs932184769 Chr.12: 52592591-52592591 on Build GRCh38
rs370287826 Chr.12: 52592531-52592531 on Build GRCh38
rs763314058 Chr.12: 52592367-52592367 on Build GRCh38
rs774770393 Chr.12: 52592471-52592471 on Build GRCh38
rs757189180 Chr.12: 52592343-52592343 on Build GRCh38
rs958777212 Chr.12: 52592693-52592693 on Build GRCh38
rs902026819 Chr.12: 52592575-52592575 on Build GRCh38
rs202119816 Chr.12: 52592516-52592516 on Build GRCh38
rs757497302 Chr.12: 52592432-52592432 on Build GRCh38
rs760291117 Chr.12: 52592484-52592484 on Build GRCh38
rs12833456 Chr.12: 52592403-52592403 on Build GRCh38
rs147981249 Chr.12: 52592491-52592491 on Build GRCh38
rs769188524 Chr.12: 52592525-52592525 on Build GRCh38
rs962147531 Chr.12: 52592501-52592501 on Build GRCh38
rs772931718 Chr.12: 52592481-52592481 on Build GRCh38
rs767843394 Chr.12: 52592414-52592416 on Build GRCh38
rs989609474 Chr.12: 52592373-52592373 on Build GRCh38
rs989661711 Chr.12: 52592547-52592547 on Build GRCh38
rs974530949 Chr.12: 52592619-52592619 on Build GRCh38
rs59458788 Chr.12: 52592526-52592526 on Build GRCh38
rs773618452 Chr.12: 52592628-52592628 on Build GRCh38
rs745964525 Chr.12: 52592439-52592439 on Build GRCh38
rs771410676 Chr.12: 52592675-52592675 on Build GRCh38
rs200584411 Chr.12: 52592402-52592402 on Build GRCh38
rs200192876 Chr.12: 52592469-52592469 on Build GRCh38
rs779334061 Chr.12: 52592548-52592548 on Build GRCh38
rs373308271 Chr.12: 52592393-52592393 on Build GRCh38
rs773019338 Chr.12: 52592541-52592541 on Build GRCh38
rs373177559 Chr.12: 52592502-52592502 on Build GRCh38
rs368017456 Chr.12: 52592543-52592543 on Build GRCh38

View Full Product Details