Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57550299-57550802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF5A
Entrez Gene ID
3798
Gene Name
kinesin family member 5A
Gene Aliases
D12S1889, MY050, NKHC, SPG10
UniGene
Hs.151219
Gene Chromosome Location
Chr.12: 57550064-57584771 on Build GRCh38
Transcripts
NM_004984.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4853705 c.73C>T p.L25L Chr.12: 57550344-57550344 on Build GRCh38
1363362 c.98delT p.F34fs*72 Chr.12: 57550369-57550369 on Build GRCh38
282252 c.116G>T p.S39I Chr.12: 57550387-57550387 on Build GRCh38
942104 c.52C>T p.P18S Chr.12: 57550323-57550323 on Build GRCh38
4608463 c.84C>A p.D28E Chr.12: 57550355-57550355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005883145 Chr.12: 57550526-57550526 on Build GRCh38
rs981831569 Chr.12: 57550626-57550626 on Build GRCh38
rs199955108 Chr.12: 57550363-57550363 on Build GRCh38
rs367708472 Chr.12: 57550638-57550638 on Build GRCh38
rs551728784 Chr.12: 57550609-57550609 on Build GRCh38
rs202202417 Chr.12: 57550439-57550439 on Build GRCh38
rs745558435 Chr.12: 57550336-57550336 on Build GRCh38
rs540463538 Chr.12: 57550368-57550368 on Build GRCh38
rs893377842 Chr.12: 57550614-57550614 on Build GRCh38
rs999138312 Chr.12: 57550499-57550499 on Build GRCh38
rs771847226 Chr.12: 57550345-57550345 on Build GRCh38
rs750467987 Chr.12: 57550417-57550417 on Build GRCh38
rs923503057 Chr.12: 57550364-57550364 on Build GRCh38
rs770302674 Chr.12: 57550323-57550323 on Build GRCh38
rs901288929 Chr.12: 57550491-57550491 on Build GRCh38
rs73346031 Chr.12: 57550710-57550710 on Build GRCh38
rs1023589192 Chr.12: 57550531-57550531 on Build GRCh38
rs768642667 Chr.12: 57550391-57550391 on Build GRCh38
rs956463320 Chr.12: 57550655-57550655 on Build GRCh38
rs573410126 Chr.12: 57550359-57550359 on Build GRCh38
rs149569914 Chr.12: 57550397-57550397 on Build GRCh38
rs1002160775 Chr.12: 57550746-57550746 on Build GRCh38
rs994188066 Chr.12: 57550494-57550494 on Build GRCh38
rs532943668 Chr.12: 57550521-57550521 on Build GRCh38
rs544250204 Chr.12: 57550456-57550456 on Build GRCh38
rs889133161 Chr.12: 57550528-57550528 on Build GRCh38
rs1053819082 Chr.12: 57550576-57550576 on Build GRCh38
rs756616721 Chr.12: 57550447-57550447 on Build GRCh38
rs562433318 Chr.12: 57550468-57550468 on Build GRCh38
rs372131378 Chr.12: 57550409-57550409 on Build GRCh38
rs969699971 Chr.12: 57550612-57550612 on Build GRCh38
rs200454498 Chr.12: 57550394-57550394 on Build GRCh38
rs751931806 Chr.12: 57550431-57550431 on Build GRCh38
rs746810755 Chr.12: 57550431-57550431 on Build GRCh38
rs903313352 Chr.12: 57550493-57550493 on Build GRCh38
rs527519747 Chr.12: 57550749-57550749 on Build GRCh38
rs892271773 Chr.12: 57550613-57550613 on Build GRCh38
rs1025444769 Chr.12: 57550508-57550508 on Build GRCh38
rs200876187 Chr.12: 57550331-57550331 on Build GRCh38
rs778633954 Chr.12: 57550449-57550449 on Build GRCh38
rs763226331 Chr.12: 57550420-57550420 on Build GRCh38
rs1024788090 Chr.12: 57550709-57550709 on Build GRCh38
rs766570565 Chr.12: 57550429-57550429 on Build GRCh38
rs755262908 Chr.12: 57550435-57550435 on Build GRCh38

View Full Product Details