Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68765298-68765803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35E3
Entrez Gene ID
55508
Gene Name
solute carrier family 35 member E3
Gene Aliases
BLOV1
UniGene
Hs.506011
Gene Chromosome Location
Chr.12: 68746148-68768677 on Build GRCh38
Transcripts
NM_018656.2 XM_017019584.1

SNP Detail

SNP ID Chromosome Location
rs925270316 Chr.12: 68765444-68765444 on Build GRCh38
rs146162108 Chr.12: 68765386-68765386 on Build GRCh38
rs779590744 Chr.12: 68765621-68765624 on Build GRCh38
rs557670325 Chr.12: 68765394-68765394 on Build GRCh38
rs71091555 Chr.12: 68765652-68765653 on Build GRCh38
rs7980064 Chr.12: 68765636-68765636 on Build GRCh38
rs576182052 Chr.12: 68765412-68765412 on Build GRCh38
rs563552878 Chr.12: 68765747-68765747 on Build GRCh38
rs896152209 Chr.12: 68765391-68765391 on Build GRCh38
rs540641637 Chr.12: 68765518-68765518 on Build GRCh38
rs910556151 Chr.12: 68765633-68765638 on Build GRCh38
rs753886562 Chr.12: 68765640-68765640 on Build GRCh38
rs748130682 Chr.12: 68765469-68765469 on Build GRCh38
rs925352705 Chr.12: 68765754-68765754 on Build GRCh38
rs886874031 Chr.12: 68765579-68765579 on Build GRCh38
rs967198309 Chr.12: 68765698-68765698 on Build GRCh38
rs187124298 Chr.12: 68765741-68765741 on Build GRCh38
rs914005148 Chr.12: 68765418-68765418 on Build GRCh38
rs1013705572 Chr.12: 68765400-68765400 on Build GRCh38
rs200429800 Chr.12: 68765429-68765429 on Build GRCh38
rs757521835 Chr.12: 68765528-68765529 on Build GRCh38
rs10682214 Chr.12: 68765622-68765622 on Build GRCh38
rs945413488 Chr.12: 68765427-68765427 on Build GRCh38
rs140181314 Chr.12: 68765550-68765550 on Build GRCh38
rs969506730 Chr.12: 68765435-68765435 on Build GRCh38
rs201323679 Chr.12: 68765542-68765542 on Build GRCh38
rs61099436 Chr.12: 68765565-68765565 on Build GRCh38
rs770559801 Chr.12: 68765730-68765731 on Build GRCh38
rs986502987 Chr.12: 68765624-68765624 on Build GRCh38
rs200321501 Chr.12: 68765620-68765620 on Build GRCh38
rs757984997 Chr.12: 68765635-68765638 on Build GRCh38
rs1020764805 Chr.12: 68765685-68765685 on Build GRCh38
rs921172316 Chr.12: 68765370-68765370 on Build GRCh38
rs777593955 Chr.12: 68765637-68765640 on Build GRCh38
rs904868612 Chr.12: 68765729-68765736 on Build GRCh38
rs929851495 Chr.12: 68765674-68765674 on Build GRCh38
rs145289164 Chr.12: 68765729-68765730 on Build GRCh38
rs938008740 Chr.12: 68765457-68765457 on Build GRCh38
rs1044122840 Chr.12: 68765443-68765443 on Build GRCh38
rs956854405 Chr.12: 68765756-68765756 on Build GRCh38
rs1003933448 Chr.12: 68765590-68765590 on Build GRCh38
rs1000602571 Chr.12: 68765738-68765738 on Build GRCh38
rs768706173 Chr.12: 68765635-68765640 on Build GRCh38
rs1009771938 Chr.12: 68765772-68765772 on Build GRCh38
rs896729663 Chr.12: 68765522-68765522 on Build GRCh38
rs182509628 Chr.12: 68765652-68765652 on Build GRCh38
rs368059660 Chr.12: 68765620-68765621 on Build GRCh38
rs574011853 Chr.12: 68765541-68765541 on Build GRCh38
rs61449205 Chr.12: 68765621-68765622 on Build GRCh38
rs942091317 Chr.12: 68765648-68765648 on Build GRCh38
rs1037779209 Chr.12: 68765668-68765669 on Build GRCh38
rs866411241 Chr.12: 68765634-68765634 on Build GRCh38
rs896361311 Chr.12: 68765694-68765694 on Build GRCh38
rs543388420 Chr.12: 68765431-68765431 on Build GRCh38
rs571781946 Chr.12: 68765385-68765385 on Build GRCh38
rs919742913 Chr.12: 68765671-68765671 on Build GRCh38
rs747998029 Chr.12: 68765624-68765625 on Build GRCh38
rs898347481 Chr.12: 68765332-68765332 on Build GRCh38
rs769741052 Chr.12: 68765698-68765699 on Build GRCh38
rs938055712 Chr.12: 68765764-68765764 on Build GRCh38
rs191335602 Chr.12: 68765471-68765471 on Build GRCh38
rs10878867 Chr.12: 68765608-68765608 on Build GRCh38
rs762585446 Chr.12: 68765708-68765708 on Build GRCh38
rs369887330 Chr.12: 68765608-68765609 on Build GRCh38
rs200183748 Chr.12: 68765606-68765607 on Build GRCh38
rs996901226 Chr.12: 68765638-68765638 on Build GRCh38
rs555615765 Chr.12: 68765458-68765458 on Build GRCh38
rs1032480595 Chr.12: 68765493-68765493 on Build GRCh38
rs954724289 Chr.12: 68765597-68765597 on Build GRCh38
rs76925627 Chr.12: 68765565-68765565 on Build GRCh38
rs776437120 Chr.12: 68765670-68765679 on Build GRCh38
rs1038144457 Chr.12: 68765329-68765330 on Build GRCh38
rs372783294 Chr.12: 68765668-68765668 on Build GRCh38
rs768582507 Chr.12: 68765361-68765361 on Build GRCh38

View Full Product Details