Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95485748-95486284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METAP2
Entrez Gene ID
10988
Gene Name
methionyl aminopeptidase 2
Gene Aliases
MAP2, MNPEP, p67eIF2
UniGene
Hs.444986
Gene Chromosome Location
Chr.12: 95474046-95515839 on Build GRCh38
Transcripts
NM_001317182.1 NM_001317183.1 NM_006838.3 NR_133673.1 XM_005268583.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466591 c.367C>A p.L123M Chr.12: 95485920-95485920 on Build GRCh38
5905908 c.374C>T p.P125L Chr.12: 95485927-95485927 on Build GRCh38
3466590 c.343C>T p.P115S Chr.12: 95485896-95485896 on Build GRCh38
4790912 c.392A>G p.K131R Chr.12: 95485945-95485945 on Build GRCh38
944663 c.328A>G p.K110E Chr.12: 95485881-95485881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935903641 Chr.12: 95486116-95486116 on Build GRCh38
rs771573036 Chr.12: 95485980-95485980 on Build GRCh38
rs779365414 Chr.12: 95485988-95485988 on Build GRCh38
rs368848423 Chr.12: 95486205-95486205 on Build GRCh38
rs773125375 Chr.12: 95486032-95486032 on Build GRCh38
rs945535193 Chr.12: 95486183-95486183 on Build GRCh38
rs971820616 Chr.12: 95486035-95486035 on Build GRCh38
rs185587799 Chr.12: 95486179-95486179 on Build GRCh38
rs1041159802 Chr.12: 95486201-95486203 on Build GRCh38
rs749193121 Chr.12: 95486101-95486101 on Build GRCh38
rs749649078 Chr.12: 95485861-95485861 on Build GRCh38
rs775611546 Chr.12: 95486009-95486009 on Build GRCh38
rs1050037175 Chr.12: 95486089-95486089 on Build GRCh38
rs556738635 Chr.12: 95486043-95486043 on Build GRCh38
rs770285178 Chr.12: 95485849-95485849 on Build GRCh38
rs771481518 Chr.12: 95485864-95485864 on Build GRCh38
rs2305293 Chr.12: 95485958-95485958 on Build GRCh38
rs932681064 Chr.12: 95486248-95486249 on Build GRCh38
rs944938140 Chr.12: 95486126-95486126 on Build GRCh38
rs775594889 Chr.12: 95485882-95485882 on Build GRCh38
rs371107641 Chr.12: 95485961-95485961 on Build GRCh38
rs375637960 Chr.12: 95486005-95486005 on Build GRCh38
rs746817286 Chr.12: 95485973-95485973 on Build GRCh38
rs746245394 Chr.12: 95485990-95485990 on Build GRCh38
rs903928749 Chr.12: 95486250-95486250 on Build GRCh38
rs760781980 Chr.12: 95486021-95486021 on Build GRCh38
rs201631697 Chr.12: 95485895-95485895 on Build GRCh38
rs746156472 Chr.12: 95485901-95485901 on Build GRCh38
rs1043859201 Chr.12: 95486199-95486199 on Build GRCh38
rs558509110 Chr.12: 95486162-95486162 on Build GRCh38
rs910813994 Chr.12: 95486109-95486109 on Build GRCh38
rs200917784 Chr.12: 95485881-95485881 on Build GRCh38
rs1053354683 Chr.12: 95485791-95485791 on Build GRCh38
rs901220330 Chr.12: 95486215-95486215 on Build GRCh38
rs186720359 Chr.12: 95485798-95485798 on Build GRCh38
rs768544048 Chr.12: 95486141-95486141 on Build GRCh38
rs201895862 Chr.12: 95485904-95485904 on Build GRCh38
rs751359091 Chr.12: 95485930-95485930 on Build GRCh38
rs373520276 Chr.12: 95485857-95485857 on Build GRCh38
rs143687752 Chr.12: 95485911-95485911 on Build GRCh38
rs778980103 Chr.12: 95485847-95485847 on Build GRCh38
rs574815007 Chr.12: 95486075-95486075 on Build GRCh38
rs751792121 Chr.12: 95486111-95486112 on Build GRCh38
rs766226459 Chr.12: 95485920-95485920 on Build GRCh38
rs1016166036 Chr.12: 95485977-95485977 on Build GRCh38
rs920341697 Chr.12: 95486045-95486045 on Build GRCh38
rs968082027 Chr.12: 95485790-95485790 on Build GRCh38
rs913075667 Chr.12: 95486137-95486137 on Build GRCh38
rs925832235 Chr.12: 95485786-95485786 on Build GRCh38
rs1040951607 Chr.12: 95486148-95486148 on Build GRCh38
rs988684435 Chr.12: 95486132-95486132 on Build GRCh38
rs199882893 Chr.12: 95486018-95486018 on Build GRCh38
rs1041127577 Chr.12: 95485894-95485894 on Build GRCh38
rs80029408 Chr.12: 95486083-95486083 on Build GRCh38
rs754599001 Chr.12: 95485943-95485943 on Build GRCh38
rs567046568 Chr.12: 95486252-95486252 on Build GRCh38
rs747625125 Chr.12: 95485829-95485829 on Build GRCh38
rs768673713 Chr.12: 95486025-95486025 on Build GRCh38
rs201267991 Chr.12: 95485964-95485964 on Build GRCh38
rs925905579 Chr.12: 95486113-95486113 on Build GRCh38
rs749878450 Chr.12: 95485971-95485971 on Build GRCh38
rs1022382499 Chr.12: 95485782-95485782 on Build GRCh38
rs200936264 Chr.12: 95485859-95485859 on Build GRCh38
rs969696217 Chr.12: 95486106-95486106 on Build GRCh38
rs1034298854 Chr.12: 95485893-95485893 on Build GRCh38
rs759768309 Chr.12: 95485869-95485869 on Build GRCh38
rs377641984 Chr.12: 95485866-95485866 on Build GRCh38
rs148941453 Chr.12: 95485905-95485905 on Build GRCh38

View Full Product Details