Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94323130-94323624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINA6
Entrez Gene ID
866
Gene Name
serpin family A member 6
Gene Aliases
CBG
UniGene
Hs.532635
Gene Chromosome Location
Chr.14: 94304248-94323351 on Build GRCh38
Transcripts
NM_001756.3

SNP Detail

SNP ID Chromosome Location
rs553091197 Chr.14: 94323350-94323350 on Build GRCh38
rs2281517 Chr.14: 94323450-94323450 on Build GRCh38
rs930514934 Chr.14: 94323274-94323274 on Build GRCh38
rs868763365 Chr.14: 94323300-94323300 on Build GRCh38
rs1033888933 Chr.14: 94323431-94323431 on Build GRCh38
rs772054522 Chr.14: 94323571-94323571 on Build GRCh38
rs552398508 Chr.14: 94323233-94323233 on Build GRCh38
rs146611849 Chr.14: 94323284-94323284 on Build GRCh38
rs1028785545 Chr.14: 94323551-94323551 on Build GRCh38
rs1025050183 Chr.14: 94323210-94323210 on Build GRCh38
rs372177821 Chr.14: 94323379-94323379 on Build GRCh38
rs995903645 Chr.14: 94323314-94323314 on Build GRCh38
rs762377943 Chr.14: 94323231-94323231 on Build GRCh38
rs181877796 Chr.14: 94323179-94323179 on Build GRCh38
rs763536443 Chr.14: 94323280-94323280 on Build GRCh38
rs541132551 Chr.14: 94323345-94323345 on Build GRCh38
rs927365128 Chr.14: 94323435-94323435 on Build GRCh38
rs1008570583 Chr.14: 94323567-94323567 on Build GRCh38
rs1000833631 Chr.14: 94323189-94323189 on Build GRCh38
rs750232689 Chr.14: 94323420-94323420 on Build GRCh38
rs951857124 Chr.14: 94323546-94323546 on Build GRCh38
rs1998056 Chr.14: 94323158-94323158 on Build GRCh38
rs910460866 Chr.14: 94323569-94323569 on Build GRCh38
rs188371806 Chr.14: 94323377-94323377 on Build GRCh38
rs968501308 Chr.14: 94323311-94323311 on Build GRCh38
rs113577834 Chr.14: 94323265-94323265 on Build GRCh38
rs895184268 Chr.14: 94323433-94323433 on Build GRCh38
rs551456304 Chr.14: 94323340-94323340 on Build GRCh38
rs866844860 Chr.14: 94323257-94323257 on Build GRCh38
rs77990214 Chr.14: 94323556-94323556 on Build GRCh38
rs904089024 Chr.14: 94323187-94323187 on Build GRCh38
rs962234716 Chr.14: 94323364-94323364 on Build GRCh38
rs115833113 Chr.14: 94323405-94323405 on Build GRCh38
rs945556432 Chr.14: 94323375-94323375 on Build GRCh38
rs938725433 Chr.14: 94323435-94323436 on Build GRCh38
rs752357337 Chr.14: 94323195-94323195 on Build GRCh38
rs186322756 Chr.14: 94323198-94323198 on Build GRCh38
rs186384700 Chr.14: 94323512-94323512 on Build GRCh38
rs905841422 Chr.14: 94323211-94323211 on Build GRCh38
rs992690614 Chr.14: 94323209-94323209 on Build GRCh38
rs973261929 Chr.14: 94323365-94323365 on Build GRCh38
rs971665642 Chr.14: 94323228-94323228 on Build GRCh38
rs142418468 Chr.14: 94323151-94323151 on Build GRCh38
rs565715807 Chr.14: 94323157-94323157 on Build GRCh38
rs532557078 Chr.14: 94323202-94323202 on Build GRCh38
rs180685032 Chr.14: 94323495-94323495 on Build GRCh38
rs112182835 Chr.14: 94323264-94323264 on Build GRCh38
rs1013575472 Chr.14: 94323440-94323440 on Build GRCh38
rs1021650944 Chr.14: 94323173-94323173 on Build GRCh38
rs1001031150 Chr.14: 94323424-94323424 on Build GRCh38
rs980181807 Chr.14: 94323421-94323421 on Build GRCh38
rs1002814361 Chr.14: 94323248-94323248 on Build GRCh38
rs115091995 Chr.14: 94323316-94323316 on Build GRCh38
rs901577369 Chr.14: 94323357-94323357 on Build GRCh38
rs959449688 Chr.14: 94323199-94323199 on Build GRCh38
rs796593451 Chr.14: 94323288-94323288 on Build GRCh38
rs904663722 Chr.14: 94323500-94323500 on Build GRCh38
rs984459205 Chr.14: 94323279-94323279 on Build GRCh38
rs1956177 Chr.14: 94323473-94323473 on Build GRCh38
rs528993857 Chr.14: 94323513-94323513 on Build GRCh38

View Full Product Details