Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28983694-28984189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPNS1
Entrez Gene ID
83985
Gene Name
sphingolipid transporter 1 (putative)
Gene Aliases
HSpin1, LAT, PP2030, SPIN1, SPINL, nrs
UniGene
Hs.617449
Gene Chromosome Location
Chr.16: 28974767-28984769 on Build GRCh38
Transcripts
NM_001142448.1 NM_001142449.1 NM_001142450.1 NM_001142451.1 NM_032038.2 XM_006721096.3 XM_017023757.1 XM_017023758.1 XM_017023759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5621907 c.1468C>T p.R490W Chr.16: 28983933-28983933 on Build GRCh38
6789078 c.1366G>A p.E456K Chr.16: 28983831-28983831 on Build GRCh38
5663310 c.1335G>T p.L445L Chr.16: 28983800-28983800 on Build GRCh38
5421062 c.1398C>T p.L466L Chr.16: 28983863-28983863 on Build GRCh38
241742 c.1426G>A p.A476T Chr.16: 28983891-28983891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022204233 Chr.16: 28983817-28983817 on Build GRCh38
rs373796041 Chr.16: 28983933-28983933 on Build GRCh38
rs900895558 Chr.16: 28983720-28983720 on Build GRCh38
rs11862178 Chr.16: 28983723-28983723 on Build GRCh38
rs4788114 Chr.16: 28983824-28983824 on Build GRCh38
rs142722234 Chr.16: 28983818-28983818 on Build GRCh38
rs765011819 Chr.16: 28983968-28983968 on Build GRCh38
rs939643295 Chr.16: 28983779-28983779 on Build GRCh38
rs141085007 Chr.16: 28983838-28983838 on Build GRCh38
rs780566473 Chr.16: 28983823-28983823 on Build GRCh38
rs755239709 Chr.16: 28983998-28984001 on Build GRCh38
rs188613201 Chr.16: 28984055-28984055 on Build GRCh38
rs759856408 Chr.16: 28984029-28984029 on Build GRCh38
rs144866787 Chr.16: 28983845-28983845 on Build GRCh38
rs767629651 Chr.16: 28983867-28983867 on Build GRCh38
rs999852543 Chr.16: 28984047-28984047 on Build GRCh38
rs759530239 Chr.16: 28983866-28983866 on Build GRCh38
rs770664577 Chr.16: 28983854-28983854 on Build GRCh38
rs1022794339 Chr.16: 28984154-28984154 on Build GRCh38
rs777395936 Chr.16: 28983996-28983996 on Build GRCh38
rs79457245 Chr.16: 28983906-28983906 on Build GRCh38
rs1034828510 Chr.16: 28984050-28984050 on Build GRCh38
rs781180363 Chr.16: 28984035-28984035 on Build GRCh38
rs566613285 Chr.16: 28983801-28983801 on Build GRCh38
rs768758578 Chr.16: 28983771-28983771 on Build GRCh38
rs755475291 Chr.16: 28983902-28983902 on Build GRCh38
rs371131211 Chr.16: 28983908-28983908 on Build GRCh38
rs946873671 Chr.16: 28984057-28984057 on Build GRCh38
rs753755127 Chr.16: 28983810-28983810 on Build GRCh38
rs780068029 Chr.16: 28983998-28983998 on Build GRCh38
rs751983098 Chr.16: 28983895-28983895 on Build GRCh38
rs36014837 Chr.16: 28983954-28983955 on Build GRCh38
rs779326526 Chr.16: 28983987-28983987 on Build GRCh38
rs778017891 Chr.16: 28983929-28983929 on Build GRCh38
rs753460582 Chr.16: 28983884-28983884 on Build GRCh38
rs781239865 Chr.16: 28983831-28983831 on Build GRCh38
rs775587482 Chr.16: 28983936-28983936 on Build GRCh38
rs747058489 Chr.16: 28983768-28983768 on Build GRCh38
rs533907477 Chr.16: 28984132-28984132 on Build GRCh38
rs768699762 Chr.16: 28984012-28984012 on Build GRCh38
rs750776330 Chr.16: 28983805-28983805 on Build GRCh38
rs777699822 Chr.16: 28983980-28983980 on Build GRCh38
rs769309598 Chr.16: 28984024-28984024 on Build GRCh38
rs770063032 Chr.16: 28983788-28983788 on Build GRCh38
rs748297241 Chr.16: 28983837-28983837 on Build GRCh38
rs934210279 Chr.16: 28983951-28983951 on Build GRCh38
rs761360134 Chr.16: 28983961-28983961 on Build GRCh38
rs922917666 Chr.16: 28984141-28984141 on Build GRCh38
rs970126950 Chr.16: 28984071-28984071 on Build GRCh38
rs780560669 Chr.16: 28984095-28984095 on Build GRCh38
rs140236840 Chr.16: 28983931-28983931 on Build GRCh38
rs370343624 Chr.16: 28983991-28983991 on Build GRCh38
rs564540512 Chr.16: 28984040-28984040 on Build GRCh38
rs993926469 Chr.16: 28984115-28984115 on Build GRCh38
rs375000927 Chr.16: 28983795-28983795 on Build GRCh38
rs763749687 Chr.16: 28983874-28983874 on Build GRCh38
rs199764231 Chr.16: 28983971-28983971 on Build GRCh38
rs776771216 Chr.16: 28983782-28983782 on Build GRCh38
rs960817089 Chr.16: 28984054-28984054 on Build GRCh38
rs1006035099 Chr.16: 28984150-28984150 on Build GRCh38
rs1047611547 Chr.16: 28984122-28984122 on Build GRCh38
rs541879892 Chr.16: 28983974-28983974 on Build GRCh38
rs766318180 Chr.16: 28983796-28983796 on Build GRCh38
rs879020759 Chr.16: 28983772-28983772 on Build GRCh38
rs150322036 Chr.16: 28983950-28983950 on Build GRCh38
rs750786482 Chr.16: 28984062-28984062 on Build GRCh38
rs138506930 Chr.16: 28983909-28983909 on Build GRCh38
rs757390248 Chr.16: 28983984-28983984 on Build GRCh38
rs568797681 Chr.16: 28984075-28984075 on Build GRCh38
rs762037718 Chr.16: 28983783-28983783 on Build GRCh38
rs773355607 Chr.16: 28983840-28983840 on Build GRCh38
rs761430686 Chr.16: 28984016-28984016 on Build GRCh38
rs753116139 Chr.16: 28984169-28984169 on Build GRCh38
rs368322414 Chr.16: 28983804-28983804 on Build GRCh38
rs536309173 Chr.16: 28983728-28983728 on Build GRCh38
rs116848701 Chr.16: 28984097-28984097 on Build GRCh38
rs748205194 Chr.16: 28983913-28983913 on Build GRCh38
rs371271382 Chr.16: 28983930-28983930 on Build GRCh38
rs763513365 Chr.16: 28983940-28983940 on Build GRCh38
rs113165798 Chr.16: 28983890-28983890 on Build GRCh38
rs776004267 Chr.16: 28984015-28984015 on Build GRCh38
rs780470001 Chr.16: 28983758-28983758 on Build GRCh38
rs879197737 Chr.16: 28983959-28983959 on Build GRCh38
rs1026655132 Chr.16: 28983887-28983887 on Build GRCh38
rs369963623 Chr.16: 28983830-28983830 on Build GRCh38
rs201490940 Chr.16: 28983859-28983859 on Build GRCh38
rs148875869 Chr.16: 28983802-28983802 on Build GRCh38
rs200095367 Chr.16: 28983937-28983937 on Build GRCh38
rs550212603 Chr.16: 28984069-28984069 on Build GRCh38
rs772598185 Chr.16: 28983742-28983742 on Build GRCh38
rs928336576 Chr.16: 28983976-28983976 on Build GRCh38
rs1040491499 Chr.16: 28984105-28984105 on Build GRCh38
rs11859822 Chr.16: 28983736-28983736 on Build GRCh38
rs368195802 Chr.16: 28983769-28983769 on Build GRCh38
rs746284233 Chr.16: 28983741-28983741 on Build GRCh38
rs756210979 Chr.16: 28983924-28983924 on Build GRCh38
rs772874528 Chr.16: 28984027-28984027 on Build GRCh38
rs560203035 Chr.16: 28983977-28983977 on Build GRCh38
rs772109038 Chr.16: 28983935-28983935 on Build GRCh38
rs754014715 Chr.16: 28983981-28983981 on Build GRCh38
rs745697231 Chr.16: 28983934-28983934 on Build GRCh38
rs566548790 Chr.16: 28984129-28984129 on Build GRCh38
rs767874913 Chr.16: 28984160-28984160 on Build GRCh38
rs760307159 Chr.16: 28983869-28983869 on Build GRCh38
rs984060861 Chr.16: 28984077-28984077 on Build GRCh38
rs969771252 Chr.16: 28984156-28984159 on Build GRCh38
rs756153037 Chr.16: 28983979-28983979 on Build GRCh38
rs181365946 Chr.16: 28984089-28984089 on Build GRCh38
rs764766781 Chr.16: 28983888-28983888 on Build GRCh38
rs373424807 Chr.16: 28983847-28983847 on Build GRCh38
rs118086285 Chr.16: 28983978-28983978 on Build GRCh38
rs759048740 Chr.16: 28983798-28983798 on Build GRCh38
rs201680708 Chr.16: 28983868-28983868 on Build GRCh38
rs374340826 Chr.16: 28984008-28984008 on Build GRCh38

View Full Product Details