Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15615647-15616154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
UniGene
Hs.548021
Gene Chromosome Location
Chr.17: 15565482-15619704 on Build GRCh38
Transcripts
NM_001282540.1 NM_006382.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667950 c.566C>G p.S189C Chr.17: 15615749-15615749 on Build GRCh38
4551340 c.526G>A p.D176N Chr.17: 15615789-15615789 on Build GRCh38
1290636 c.538delT p.S180fs*58 Chr.17: 15615777-15615777 on Build GRCh38
6330282 c.506-1G>T p.? Chr.17: 15615810-15615810 on Build GRCh38
3514508 c.616C>T p.P206S Chr.17: 15615699-15615699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023816183 Chr.17: 15615973-15615973 on Build GRCh38
rs549826227 Chr.17: 15616049-15616049 on Build GRCh38
rs543887330 Chr.17: 15615833-15615833 on Build GRCh38
rs982257879 Chr.17: 15615992-15615992 on Build GRCh38
rs766990929 Chr.17: 15615819-15615819 on Build GRCh38
rs560903732 Chr.17: 15615827-15615827 on Build GRCh38
rs531510415 Chr.17: 15615899-15615899 on Build GRCh38
rs935265783 Chr.17: 15615848-15615848 on Build GRCh38
rs10432021 Chr.17: 15615860-15615860 on Build GRCh38
rs750755512 Chr.17: 15615720-15615720 on Build GRCh38
rs567230696 Chr.17: 15615976-15615976 on Build GRCh38
rs1005124602 Chr.17: 15615938-15615938 on Build GRCh38
rs762681852 Chr.17: 15615782-15615782 on Build GRCh38
rs770698143 Chr.17: 15615793-15615793 on Build GRCh38
rs570189689 Chr.17: 15616093-15616093 on Build GRCh38
rs758695103 Chr.17: 15615835-15615835 on Build GRCh38
rs370886881 Chr.17: 15615736-15615736 on Build GRCh38
rs774958112 Chr.17: 15615823-15615823 on Build GRCh38
rs867756712 Chr.17: 15616062-15616062 on Build GRCh38
rs927817128 Chr.17: 15615999-15615999 on Build GRCh38
rs369772327 Chr.17: 15615673-15615673 on Build GRCh38
rs947371517 Chr.17: 15615884-15615884 on Build GRCh38
rs559515043 Chr.17: 15616073-15616073 on Build GRCh38
rs900555626 Chr.17: 15615914-15615914 on Build GRCh38
rs756217794 Chr.17: 15615845-15615845 on Build GRCh38
rs781464763 Chr.17: 15615843-15615843 on Build GRCh38
rs551418976 Chr.17: 15615906-15615906 on Build GRCh38
rs779298883 Chr.17: 15615709-15615709 on Build GRCh38
rs373640136 Chr.17: 15615692-15615692 on Build GRCh38
rs988060255 Chr.17: 15616015-15616015 on Build GRCh38
rs536008345 Chr.17: 15616011-15616011 on Build GRCh38
rs766761768 Chr.17: 15615836-15615836 on Build GRCh38
rs572371649 Chr.17: 15615731-15615731 on Build GRCh38
rs184045735 Chr.17: 15615950-15615950 on Build GRCh38
rs780440925 Chr.17: 15615728-15615728 on Build GRCh38
rs187406714 Chr.17: 15616094-15616094 on Build GRCh38
rs913276559 Chr.17: 15615850-15615850 on Build GRCh38
rs565193427 Chr.17: 15615926-15615926 on Build GRCh38
rs752066262 Chr.17: 15615839-15615839 on Build GRCh38
rs973598983 Chr.17: 15615704-15615704 on Build GRCh38
rs748271774 Chr.17: 15615844-15615844 on Build GRCh38
rs1043542738 Chr.17: 15615898-15615898 on Build GRCh38
rs969191327 Chr.17: 15615987-15615987 on Build GRCh38
rs370221413 Chr.17: 15615832-15615832 on Build GRCh38
rs778157654 Chr.17: 15615846-15615846 on Build GRCh38
rs147264211 Chr.17: 15615667-15615667 on Build GRCh38
rs996767233 Chr.17: 15615923-15615923 on Build GRCh38
rs981946059 Chr.17: 15615816-15615816 on Build GRCh38
rs372782740 Chr.17: 15615746-15615746 on Build GRCh38
rs562585905 Chr.17: 15615866-15615866 on Build GRCh38
rs755489179 Chr.17: 15615841-15615841 on Build GRCh38
rs759508281 Chr.17: 15615809-15615809 on Build GRCh38
rs879749155 Chr.17: 15616057-15616057 on Build GRCh38
rs994476711 Chr.17: 15615957-15615957 on Build GRCh38
rs1015125393 Chr.17: 15615945-15615945 on Build GRCh38
rs558810490 Chr.17: 15615729-15615729 on Build GRCh38
rs547418957 Chr.17: 15615955-15615955 on Build GRCh38
rs10432022 Chr.17: 15615875-15615875 on Build GRCh38
rs748433280 Chr.17: 15615735-15615735 on Build GRCh38
rs890609668 Chr.17: 15615937-15615937 on Build GRCh38
rs956525100 Chr.17: 15616009-15616009 on Build GRCh38
rs758747777 Chr.17: 15615722-15615722 on Build GRCh38
rs915128554 Chr.17: 15616031-15616031 on Build GRCh38
rs199671807 Chr.17: 15615801-15615801 on Build GRCh38

View Full Product Details