Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15717375-15717875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF286A
Entrez Gene ID
57335
Gene Name
zinc finger protein 286A
Gene Aliases
ZNF286
UniGene
Hs.585799
Gene Chromosome Location
Chr.17: 15699577-15720787 on Build GRCh38
Transcripts
NM_001130842.1 NM_001288642.1 NM_001288643.1 NM_001288644.1 NM_001288645.1 NM_001288646.1 NM_001288647.1 NM_001288648.1 NM_001288649.1 NM_020652.2

SNP Detail

SNP ID Chromosome Location
rs770039385 Chr.17: 15717506-15717506 on Build GRCh38
rs913434286 Chr.17: 15717600-15717600 on Build GRCh38
rs940485408 Chr.17: 15717700-15717700 on Build GRCh38
rs898883149 Chr.17: 15717747-15717747 on Build GRCh38
rs1051408379 Chr.17: 15717688-15717688 on Build GRCh38
rs895811796 Chr.17: 15717802-15717802 on Build GRCh38
rs969299110 Chr.17: 15717603-15717603 on Build GRCh38
rs550778478 Chr.17: 15717514-15717514 on Build GRCh38
rs1043693952 Chr.17: 15717442-15717442 on Build GRCh38
rs72821704 Chr.17: 15717561-15717561 on Build GRCh38
rs1035677772 Chr.17: 15717563-15717563 on Build GRCh38
rs922174194 Chr.17: 15717450-15717450 on Build GRCh38
rs911540822 Chr.17: 15717696-15717696 on Build GRCh38
rs566390201 Chr.17: 15717631-15717631 on Build GRCh38
rs1036064381 Chr.17: 15717711-15717711 on Build GRCh38
rs531683663 Chr.17: 15717497-15717497 on Build GRCh38
rs62071728 Chr.17: 15717765-15717765 on Build GRCh38
rs180819876 Chr.17: 15717790-15717790 on Build GRCh38
rs960133914 Chr.17: 15717580-15717580 on Build GRCh38
rs780547373 Chr.17: 15717802-15717802 on Build GRCh38
rs376967329 Chr.17: 15717517-15717517 on Build GRCh38
rs529041872 Chr.17: 15717571-15717571 on Build GRCh38
rs555035011 Chr.17: 15717739-15717739 on Build GRCh38
rs748451492 Chr.17: 15717773-15717777 on Build GRCh38
rs560516458 Chr.17: 15717440-15717440 on Build GRCh38
rs546494500 Chr.17: 15717588-15717588 on Build GRCh38
rs1004577471 Chr.17: 15717490-15717490 on Build GRCh38
rs1010205679 Chr.17: 15717818-15717818 on Build GRCh38
rs529306482 Chr.17: 15717458-15717458 on Build GRCh38
rs978913264 Chr.17: 15717608-15717608 on Build GRCh38
rs563011782 Chr.17: 15717491-15717491 on Build GRCh38
rs932224038 Chr.17: 15717669-15717669 on Build GRCh38
rs540920506 Chr.17: 15717419-15717419 on Build GRCh38
rs878936400 Chr.17: 15717515-15717515 on Build GRCh38
rs535096856 Chr.17: 15717662-15717662 on Build GRCh38
rs570538004 Chr.17: 15717523-15717523 on Build GRCh38
rs1051355284 Chr.17: 15717459-15717459 on Build GRCh38
rs1003442205 Chr.17: 15717791-15717791 on Build GRCh38
rs566040586 Chr.17: 15717819-15717819 on Build GRCh38
rs111947351 Chr.17: 15717686-15717688 on Build GRCh38
rs879250403 Chr.17: 15717799-15717799 on Build GRCh38
rs543000747 Chr.17: 15717460-15717460 on Build GRCh38

View Full Product Details