Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17995034-17995541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRC3
Entrez Gene ID
83450
Gene Name
dynein regulatory complex subunit 3
Gene Aliases
CFAP134, LRRC48
UniGene
Hs.579264
Gene Chromosome Location
Chr.17: 17972766-18016889 on Build GRCh38
Transcripts
NM_001130090.1 NM_001130091.1 NM_001130092.1 NM_031294.3 XM_006721583.3 XM_011524018.2 XM_011524019.2 XM_011524020.2 XM_011524021.2 XM_011524022.2 XM_011524023.2 XM_011524025.2 XM_011524026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064429 c.757A>G p.M253V Chr.17: 17995044-17995044 on Build GRCh38
5910934 c.824+5C>T p.? Chr.17: 17995116-17995116 on Build GRCh38
4064428 c.757A>G p.M253V Chr.17: 17995044-17995044 on Build GRCh38
5910933 c.824+5C>T p.? Chr.17: 17995116-17995116 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779776072 Chr.17: 17995220-17995220 on Build GRCh38
rs760734762 Chr.17: 17995074-17995074 on Build GRCh38
rs553336000 Chr.17: 17995389-17995389 on Build GRCh38
rs982307102 Chr.17: 17995246-17995246 on Build GRCh38
rs781657886 Chr.17: 17995423-17995423 on Build GRCh38
rs753600788 Chr.17: 17995082-17995082 on Build GRCh38
rs535856387 Chr.17: 17995471-17995471 on Build GRCh38
rs111309590 Chr.17: 17995065-17995065 on Build GRCh38
rs962824500 Chr.17: 17995224-17995224 on Build GRCh38
rs768792328 Chr.17: 17995518-17995518 on Build GRCh38
rs986062217 Chr.17: 17995148-17995148 on Build GRCh38
rs761543661 Chr.17: 17995092-17995092 on Build GRCh38
rs753473585 Chr.17: 17995253-17995253 on Build GRCh38
rs767633482 Chr.17: 17995066-17995066 on Build GRCh38
rs533510758 Chr.17: 17995343-17995343 on Build GRCh38
rs778146635 Chr.17: 17995308-17995308 on Build GRCh38
rs897273539 Chr.17: 17995357-17995357 on Build GRCh38
rs775615688 Chr.17: 17995162-17995175 on Build GRCh38
rs945090360 Chr.17: 17995241-17995241 on Build GRCh38
rs549599488 Chr.17: 17995108-17995108 on Build GRCh38
rs750149905 Chr.17: 17995095-17995095 on Build GRCh38
rs774591992 Chr.17: 17995064-17995064 on Build GRCh38
rs753129002 Chr.17: 17995115-17995115 on Build GRCh38
rs749627432 Chr.17: 17995127-17995127 on Build GRCh38
rs118118204 Chr.17: 17995217-17995217 on Build GRCh38
rs769801869 Chr.17: 17995464-17995464 on Build GRCh38
rs763186411 Chr.17: 17995492-17995492 on Build GRCh38
rs550474872 Chr.17: 17995124-17995124 on Build GRCh38
rs981458612 Chr.17: 17995301-17995301 on Build GRCh38
rs764932947 Chr.17: 17995094-17995094 on Build GRCh38
rs928741071 Chr.17: 17995302-17995302 on Build GRCh38
rs775375793 Chr.17: 17995072-17995072 on Build GRCh38
rs772135761 Chr.17: 17995147-17995147 on Build GRCh38
rs763629865 Chr.17: 17995089-17995089 on Build GRCh38
rs763786025 Chr.17: 17995079-17995079 on Build GRCh38
rs753825599 Chr.17: 17995495-17995496 on Build GRCh38
rs995725556 Chr.17: 17995233-17995233 on Build GRCh38
rs746238738 Chr.17: 17995416-17995416 on Build GRCh38
rs957960750 Chr.17: 17995479-17995479 on Build GRCh38
rs746128692 Chr.17: 17995134-17995134 on Build GRCh38
rs569342179 Chr.17: 17995179-17995179 on Build GRCh38
rs771310302 Chr.17: 17995131-17995131 on Build GRCh38
rs746443273 Chr.17: 17995456-17995456 on Build GRCh38
rs1036972602 Chr.17: 17995356-17995356 on Build GRCh38
rs1051327481 Chr.17: 17995178-17995178 on Build GRCh38
rs888455603 Chr.17: 17995304-17995304 on Build GRCh38
rs754595964 Chr.17: 17995262-17995262 on Build GRCh38
rs756584085 Chr.17: 17995117-17995117 on Build GRCh38
rs142781572 Chr.17: 17995273-17995273 on Build GRCh38
rs11656675 Chr.17: 17995394-17995394 on Build GRCh38
rs747361761 Chr.17: 17995335-17995335 on Build GRCh38
rs771078867 Chr.17: 17995058-17995058 on Build GRCh38
rs779089812 Chr.17: 17995132-17995132 on Build GRCh38
rs989278430 Chr.17: 17995321-17995321 on Build GRCh38
rs577241821 Chr.17: 17995292-17995292 on Build GRCh38
rs1023134654 Chr.17: 17995272-17995272 on Build GRCh38
rs913631135 Chr.17: 17995236-17995236 on Build GRCh38
rs772135861 Chr.17: 17995142-17995169 on Build GRCh38
rs779773820 Chr.17: 17995111-17995111 on Build GRCh38
rs1017491799 Chr.17: 17995476-17995476 on Build GRCh38

View Full Product Details