Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32376181-32376679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1 XM_005270986.4 XM_011541677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4383304 c.821A>G p.Q274R Chr.1: 32376648-32376648 on Build GRCh38
4383305 c.821A>G p.Q274R Chr.1: 32376648-32376648 on Build GRCh38
1584476 c.1116G>A p.E372E Chr.1: 32376353-32376353 on Build GRCh38
2078010 c.820delC p.Q274fs*9 Chr.1: 32376649-32376649 on Build GRCh38
4031493 c.874A>G p.T292A Chr.1: 32376595-32376595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767166821 Chr.1: 32376610-32376610 on Build GRCh38
rs534002148 Chr.1: 32376453-32376453 on Build GRCh38
rs539863664 Chr.1: 32376329-32376329 on Build GRCh38
rs753294266 Chr.1: 32376263-32376263 on Build GRCh38
rs1018341810 Chr.1: 32376531-32376531 on Build GRCh38
rs113956658 Chr.1: 32376225-32376225 on Build GRCh38
rs997688250 Chr.1: 32376573-32376573 on Build GRCh38
rs765957048 Chr.1: 32376510-32376510 on Build GRCh38
rs768131064 Chr.1: 32376262-32376262 on Build GRCh38
rs766120043 Chr.1: 32376411-32376411 on Build GRCh38
rs200768243 Chr.1: 32376253-32376253 on Build GRCh38
rs919388297 Chr.1: 32376416-32376416 on Build GRCh38
rs369320327 Chr.1: 32376216-32376216 on Build GRCh38
rs774777636 Chr.1: 32376378-32376378 on Build GRCh38
rs1056858521 Chr.1: 32376425-32376425 on Build GRCh38
rs939373843 Chr.1: 32376424-32376424 on Build GRCh38
rs967540445 Chr.1: 32376231-32376231 on Build GRCh38
rs137963263 Chr.1: 32376551-32376551 on Build GRCh38
rs770608172 Chr.1: 32376598-32376598 on Build GRCh38
rs751315201 Chr.1: 32376420-32376420 on Build GRCh38
rs140093699 Chr.1: 32376322-32376322 on Build GRCh38
rs930767236 Chr.1: 32376291-32376291 on Build GRCh38
rs777074928 Chr.1: 32376468-32376468 on Build GRCh38
rs768507023 Chr.1: 32376357-32376357 on Build GRCh38
rs199507469 Chr.1: 32376431-32376431 on Build GRCh38
rs764778612 Chr.1: 32376492-32376492 on Build GRCh38
rs746040723 Chr.1: 32376436-32376436 on Build GRCh38
rs756140270 Chr.1: 32376542-32376542 on Build GRCh38
rs773915545 Chr.1: 32376599-32376599 on Build GRCh38
rs999276364 Chr.1: 32376505-32376505 on Build GRCh38
rs373221193 Chr.1: 32376335-32376335 on Build GRCh38
rs1030885629 Chr.1: 32376514-32376514 on Build GRCh38
rs772737804 Chr.1: 32376506-32376506 on Build GRCh38
rs963859893 Chr.1: 32376532-32376532 on Build GRCh38
rs780024884 Chr.1: 32376221-32376221 on Build GRCh38
rs754886873 Chr.1: 32376426-32376426 on Build GRCh38
rs756871320 Chr.1: 32376303-32376303 on Build GRCh38
rs542990623 Chr.1: 32376386-32376386 on Build GRCh38
rs754796139 Chr.1: 32376517-32376517 on Build GRCh38
rs777841286 Chr.1: 32376358-32376358 on Build GRCh38
rs772612008 Chr.1: 32376385-32376385 on Build GRCh38
rs770288004 Chr.1: 32376570-32376570 on Build GRCh38
rs757449292 Chr.1: 32376428-32376428 on Build GRCh38
rs749341450 Chr.1: 32376364-32376364 on Build GRCh38
rs762522583 Chr.1: 32376508-32376508 on Build GRCh38
rs908152499 Chr.1: 32376552-32376552 on Build GRCh38
rs373949372 Chr.1: 32376562-32376562 on Build GRCh38
rs758636700 Chr.1: 32376550-32376550 on Build GRCh38
rs145768557 Chr.1: 32376563-32376563 on Build GRCh38
rs1016559065 Chr.1: 32376623-32376623 on Build GRCh38
rs559169736 Chr.1: 32376281-32376281 on Build GRCh38
rs376399921 Chr.1: 32376398-32376398 on Build GRCh38
rs376175900 Chr.1: 32376405-32376405 on Build GRCh38
rs752572832 Chr.1: 32376618-32376618 on Build GRCh38
rs774798291 Chr.1: 32376244-32376244 on Build GRCh38
rs147609045 Chr.1: 32376282-32376282 on Build GRCh38
rs532220214 Chr.1: 32376325-32376325 on Build GRCh38
rs911009571 Chr.1: 32376214-32376214 on Build GRCh38
rs752624440 Chr.1: 32376534-32376534 on Build GRCh38
rs11540980 Chr.1: 32376604-32376604 on Build GRCh38
rs867515163 Chr.1: 32376271-32376271 on Build GRCh38
rs186550400 Chr.1: 32376370-32376370 on Build GRCh38
rs772455480 Chr.1: 32376438-32376438 on Build GRCh38
rs138488087 Chr.1: 32376579-32376579 on Build GRCh38
rs760084708 Chr.1: 32376274-32376274 on Build GRCh38
rs769959824 Chr.1: 32376224-32376224 on Build GRCh38
rs778550453 Chr.1: 32376307-32376307 on Build GRCh38
rs759945487 Chr.1: 32376384-32376384 on Build GRCh38
rs776152956 Chr.1: 32376389-32376389 on Build GRCh38
rs1043762372 Chr.1: 32376447-32376447 on Build GRCh38
rs748173227 Chr.1: 32376222-32376222 on Build GRCh38
rs375037008 Chr.1: 32376217-32376217 on Build GRCh38
rs775858534 Chr.1: 32376446-32376446 on Build GRCh38
rs1007934435 Chr.1: 32376523-32376523 on Build GRCh38
rs767458494 Chr.1: 32376522-32376522 on Build GRCh38
rs747835366 Chr.1: 32376607-32376607 on Build GRCh38
rs764141724 Chr.1: 32376648-32376648 on Build GRCh38
rs779097844 Chr.1: 32376545-32376545 on Build GRCh38
rs772648029 Chr.1: 32376574-32376574 on Build GRCh38
rs372990632 Chr.1: 32376399-32376399 on Build GRCh38
rs1030068531 Chr.1: 32376591-32376591 on Build GRCh38
rs748607896 Chr.1: 32376565-32376565 on Build GRCh38
rs201712402 Chr.1: 32376427-32376427 on Build GRCh38
rs372793606 Chr.1: 32376537-32376537 on Build GRCh38
rs771282804 Chr.1: 32376239-32376239 on Build GRCh38
rs372365622 Chr.1: 32376302-32376302 on Build GRCh38
rs761285170 Chr.1: 32376477-32376477 on Build GRCh38
rs865907637 Chr.1: 32376544-32376544 on Build GRCh38
rs769196804 Chr.1: 32376457-32376457 on Build GRCh38
rs903715652 Chr.1: 32376501-32376501 on Build GRCh38
rs758569110 Chr.1: 32376388-32376388 on Build GRCh38
rs750118665 Chr.1: 32376403-32376403 on Build GRCh38
rs763614691 Chr.1: 32376301-32376301 on Build GRCh38
rs143046222 Chr.1: 32376226-32376226 on Build GRCh38
rs760624182 Chr.1: 32376625-32376625 on Build GRCh38
rs555559494 Chr.1: 32376511-32376511 on Build GRCh38
rs370603325 Chr.1: 32376546-32376546 on Build GRCh38
rs973829875 Chr.1: 32376533-32376533 on Build GRCh38

View Full Product Details