Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61456325-61456831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX4
Entrez Gene ID
9496
Gene Name
T-box 4
Gene Aliases
ICPPS, SPS
UniGene
Hs.143907
Gene Chromosome Location
Chr.17: 61452418-61485110 on Build GRCh38
Transcripts
NM_001321120.1 NM_018488.3 XM_011525490.2 XM_011525491.2 XM_011525495.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4821419 c.30C>T p.S10S Chr.17: 61456520-61456520 on Build GRCh38
5725096 c.65G>A p.G22E Chr.17: 61456555-61456555 on Build GRCh38
3755734 c.17G>C p.G6A Chr.17: 61456507-61456507 on Build GRCh38
6412725 c.17G>A p.G6D Chr.17: 61456507-61456507 on Build GRCh38
5517497 c.67G>A p.E23K Chr.17: 61456557-61456557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573485618 Chr.17: 61456561-61456561 on Build GRCh38
rs764516902 Chr.17: 61456473-61456473 on Build GRCh38
rs748755835 Chr.17: 61456515-61456515 on Build GRCh38
rs374081904 Chr.17: 61456377-61456377 on Build GRCh38
rs750615061 Chr.17: 61456484-61456484 on Build GRCh38
rs879172188 Chr.17: 61456632-61456632 on Build GRCh38
rs571382932 Chr.17: 61456376-61456376 on Build GRCh38
rs756802394 Chr.17: 61456524-61456524 on Build GRCh38
rs972113931 Chr.17: 61456385-61456385 on Build GRCh38
rs924569049 Chr.17: 61456583-61456583 on Build GRCh38
rs776660359 Chr.17: 61456513-61456513 on Build GRCh38
rs763175971 Chr.17: 61456640-61456640 on Build GRCh38
rs1053508476 Chr.17: 61456501-61456501 on Build GRCh38
rs774561065 Chr.17: 61456437-61456437 on Build GRCh38
rs886053187 Chr.17: 61456600-61456600 on Build GRCh38
rs771639735 Chr.17: 61456533-61456533 on Build GRCh38
rs906087336 Chr.17: 61456744-61456744 on Build GRCh38
rs971863326 Chr.17: 61456576-61456576 on Build GRCh38
rs778604942 Chr.17: 61456531-61456531 on Build GRCh38
rs755880238 Chr.17: 61456649-61456649 on Build GRCh38
rs868766804 Chr.17: 61456527-61456527 on Build GRCh38
rs924403939 Chr.17: 61456425-61456425 on Build GRCh38
rs567745835 Chr.17: 61456356-61456356 on Build GRCh38
rs188957952 Chr.17: 61456426-61456426 on Build GRCh38
rs951749304 Chr.17: 61456587-61456587 on Build GRCh38
rs766513791 Chr.17: 61456644-61456644 on Build GRCh38
rs923153057 Chr.17: 61456628-61456628 on Build GRCh38
rs898060595 Chr.17: 61456643-61456643 on Build GRCh38
rs3744448 Chr.17: 61456507-61456507 on Build GRCh38
rs575407008 Chr.17: 61456662-61456662 on Build GRCh38
rs976025717 Chr.17: 61456414-61456414 on Build GRCh38
rs985777915 Chr.17: 61456375-61456375 on Build GRCh38
rs886053186 Chr.17: 61456598-61456598 on Build GRCh38
rs775860775 Chr.17: 61456553-61456553 on Build GRCh38
rs558901097 Chr.17: 61456636-61456636 on Build GRCh38
rs926593125 Chr.17: 61456616-61456616 on Build GRCh38
rs1017376942 Chr.17: 61456771-61456771 on Build GRCh38
rs887945237 Chr.17: 61456685-61456685 on Build GRCh38
rs765350383 Chr.17: 61456483-61456483 on Build GRCh38
rs951670338 Chr.17: 61456364-61456364 on Build GRCh38
rs769433167 Chr.17: 61456503-61456503 on Build GRCh38
rs115059628 Chr.17: 61456692-61456692 on Build GRCh38
rs943166340 Chr.17: 61456440-61456440 on Build GRCh38
rs764025450 Chr.17: 61456651-61456651 on Build GRCh38
rs753878539 Chr.17: 61456481-61456481 on Build GRCh38
rs914572255 Chr.17: 61456654-61456655 on Build GRCh38
rs772891363 Chr.17: 61456655-61456657 on Build GRCh38
rs761904474 Chr.17: 61456482-61456482 on Build GRCh38
rs749406551 Chr.17: 61456358-61456358 on Build GRCh38
rs748638630 Chr.17: 61456372-61456372 on Build GRCh38
rs918948901 Chr.17: 61456408-61456408 on Build GRCh38
rs148424252 Chr.17: 61456594-61456594 on Build GRCh38
rs761189656 Chr.17: 61456468-61456468 on Build GRCh38
rs946799652 Chr.17: 61456603-61456603 on Build GRCh38
rs775779053 Chr.17: 61456463-61456463 on Build GRCh38
rs117410176 Chr.17: 61456506-61456506 on Build GRCh38
rs752496226 Chr.17: 61456646-61456646 on Build GRCh38
rs747369610 Chr.17: 61456554-61456554 on Build GRCh38
rs762348865 Chr.17: 61456584-61456584 on Build GRCh38
rs1057096239 Chr.17: 61456641-61456641 on Build GRCh38
rs104894648 Chr.17: 61456674-61456674 on Build GRCh38
rs947332474 Chr.17: 61456667-61456667 on Build GRCh38
rs868469301 Chr.17: 61456493-61456493 on Build GRCh38
rs965946950 Chr.17: 61456396-61456396 on Build GRCh38
rs1035145047 Chr.17: 61456357-61456357 on Build GRCh38
rs1045129746 Chr.17: 61456615-61456615 on Build GRCh38
rs567672041 Chr.17: 61456465-61456465 on Build GRCh38
rs752448330 Chr.17: 61456508-61456508 on Build GRCh38
rs1052923226 Chr.17: 61456449-61456449 on Build GRCh38
rs1023259344 Chr.17: 61456512-61456512 on Build GRCh38
rs80164240 Chr.17: 61456665-61456665 on Build GRCh38
rs745523181 Chr.17: 61456532-61456532 on Build GRCh38
rs1040213179 Chr.17: 61456448-61456448 on Build GRCh38
rs537562420 Chr.17: 61456389-61456389 on Build GRCh38
rs929578916 Chr.17: 61456645-61456645 on Build GRCh38
rs910435096 Chr.17: 61456588-61456588 on Build GRCh38
rs777880490 Chr.17: 61456514-61456514 on Build GRCh38
rs1007353431 Chr.17: 61456538-61456538 on Build GRCh38
rs984702351 Chr.17: 61456415-61456415 on Build GRCh38
rs769157857 Chr.17: 61456555-61456555 on Build GRCh38
rs765299000 Chr.17: 61456593-61456593 on Build GRCh38
rs956174365 Chr.17: 61456589-61456589 on Build GRCh38

View Full Product Details