Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50070766-50071265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
472977 c.695G>T p.W232L Chr.17: 50070874-50070874 on Build GRCh38
3989123 c.710A>G p.Y237C Chr.17: 50070889-50070889 on Build GRCh38
77909 c.703T>C p.S235P Chr.17: 50070882-50070882 on Build GRCh38
3518976 c.711T>C p.Y237Y Chr.17: 50070890-50070890 on Build GRCh38
1135957 c.695G>T p.W232L Chr.17: 50070874-50070874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747560253 Chr.17: 50070827-50070827 on Build GRCh38
rs758927070 Chr.17: 50070797-50070797 on Build GRCh38
rs73330539 Chr.17: 50071092-50071092 on Build GRCh38
rs557617782 Chr.17: 50070904-50070904 on Build GRCh38
rs368380539 Chr.17: 50070929-50070929 on Build GRCh38
rs1008005015 Chr.17: 50070870-50070870 on Build GRCh38
rs1054897358 Chr.17: 50071197-50071197 on Build GRCh38
rs137894674 Chr.17: 50071098-50071098 on Build GRCh38
rs2301628 Chr.17: 50071049-50071049 on Build GRCh38
rs192243328 Chr.17: 50071031-50071031 on Build GRCh38
rs777082984 Chr.17: 50070957-50070957 on Build GRCh38
rs560251647 Chr.17: 50071132-50071132 on Build GRCh38
rs765579449 Chr.17: 50070911-50070911 on Build GRCh38
rs750490348 Chr.17: 50071057-50071057 on Build GRCh38
rs34149622 Chr.17: 50070998-50070999 on Build GRCh38
rs370414445 Chr.17: 50070839-50070839 on Build GRCh38
rs934374521 Chr.17: 50070796-50070796 on Build GRCh38
rs772513802 Chr.17: 50070946-50070957 on Build GRCh38
rs943934161 Chr.17: 50071170-50071170 on Build GRCh38
rs758804602 Chr.17: 50070920-50070920 on Build GRCh38
rs369606985 Chr.17: 50071219-50071219 on Build GRCh38
rs115842631 Chr.17: 50070965-50070965 on Build GRCh38
rs577615099 Chr.17: 50071102-50071102 on Build GRCh38
rs752000829 Chr.17: 50070941-50070941 on Build GRCh38
rs775371538 Chr.17: 50070875-50070875 on Build GRCh38
rs994296608 Chr.17: 50071232-50071232 on Build GRCh38
rs201400074 Chr.17: 50070812-50070812 on Build GRCh38
rs539540896 Chr.17: 50070882-50070882 on Build GRCh38
rs982287658 Chr.17: 50071048-50071048 on Build GRCh38
rs61730089 Chr.17: 50070866-50070866 on Build GRCh38
rs769430191 Chr.17: 50070925-50070925 on Build GRCh38
rs888472286 Chr.17: 50071234-50071234 on Build GRCh38
rs990317077 Chr.17: 50071014-50071014 on Build GRCh38
rs896948038 Chr.17: 50071224-50071224 on Build GRCh38
rs758093831 Chr.17: 50071218-50071219 on Build GRCh38
rs566628333 Chr.17: 50071013-50071013 on Build GRCh38
rs1049663768 Chr.17: 50071152-50071152 on Build GRCh38
rs762848462 Chr.17: 50070982-50070982 on Build GRCh38
rs988960170 Chr.17: 50071070-50071070 on Build GRCh38
rs72840624 Chr.17: 50071155-50071155 on Build GRCh38
rs755579979 Chr.17: 50070943-50070943 on Build GRCh38
rs777403418 Chr.17: 50070851-50070851 on Build GRCh38
rs1048953480 Chr.17: 50071233-50071233 on Build GRCh38
rs140829923 Chr.17: 50070865-50070865 on Build GRCh38
rs970913075 Chr.17: 50071045-50071045 on Build GRCh38
rs756850578 Chr.17: 50070964-50070964 on Build GRCh38
rs766592990 Chr.17: 50071147-50071147 on Build GRCh38
rs770624378 Chr.17: 50070862-50070862 on Build GRCh38
rs748890642 Chr.17: 50070854-50070854 on Build GRCh38
rs764066781 Chr.17: 50070890-50070890 on Build GRCh38
rs748846687 Chr.17: 50070963-50070963 on Build GRCh38
rs898370013 Chr.17: 50070836-50070836 on Build GRCh38
rs78253610 Chr.17: 50071058-50071058 on Build GRCh38
rs1019838935 Chr.17: 50070888-50070888 on Build GRCh38
rs1053085327 Chr.17: 50071241-50071241 on Build GRCh38
rs750779502 Chr.17: 50070914-50070914 on Build GRCh38
rs137929880 Chr.17: 50070863-50070863 on Build GRCh38
rs539853180 Chr.17: 50071117-50071117 on Build GRCh38
rs1023875725 Chr.17: 50071035-50071035 on Build GRCh38
rs776835697 Chr.17: 50070903-50070903 on Build GRCh38
rs957563043 Chr.17: 50070987-50070987 on Build GRCh38
rs972549559 Chr.17: 50071141-50071141 on Build GRCh38

View Full Product Details