Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26192514-26193059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMA8
Entrez Gene ID
143471
Gene Name
proteasome subunit alpha 8
Gene Aliases
PSMA7L
UniGene
Hs.464813
Gene Chromosome Location
Chr.18: 26133847-26193355 on Build GRCh38
Transcripts
NM_001025096.1 NM_001025097.1 NM_001308188.1 NM_144662.2 XM_011525818.1

SNP Detail

SNP ID Chromosome Location
rs2069310 Chr.18: 26192609-26192609 on Build GRCh38
rs545247104 Chr.18: 26192945-26192945 on Build GRCh38
rs974355056 Chr.18: 26192755-26192755 on Build GRCh38
rs964520613 Chr.18: 26192595-26192595 on Build GRCh38
rs955128778 Chr.18: 26192994-26192994 on Build GRCh38
rs1000863004 Chr.18: 26192847-26192847 on Build GRCh38
rs1040077194 Chr.18: 26192779-26192779 on Build GRCh38
rs539287968 Chr.18: 26192723-26192723 on Build GRCh38
rs971690297 Chr.18: 26192891-26192891 on Build GRCh38
rs973767560 Chr.18: 26192980-26192980 on Build GRCh38
rs956817847 Chr.18: 26192866-26192866 on Build GRCh38
rs541636681 Chr.18: 26192938-26192938 on Build GRCh38
rs3794196 Chr.18: 26192977-26192977 on Build GRCh38
rs1055155662 Chr.18: 26192941-26192941 on Build GRCh38
rs926957910 Chr.18: 26192642-26192642 on Build GRCh38
rs1007156385 Chr.18: 26192756-26192756 on Build GRCh38
rs1031038518 Chr.18: 26192859-26192859 on Build GRCh38
rs937880023 Chr.18: 26192804-26192804 on Build GRCh38
rs192674845 Chr.18: 26192722-26192722 on Build GRCh38
rs1056213184 Chr.18: 26192820-26192820 on Build GRCh38
rs1000827971 Chr.18: 26192568-26192568 on Build GRCh38
rs997048778 Chr.18: 26192969-26192969 on Build GRCh38
rs755621879 Chr.18: 26192547-26192547 on Build GRCh38
rs941314058 Chr.18: 26192879-26192879 on Build GRCh38
rs1041733017 Chr.18: 26192908-26192908 on Build GRCh38
rs765673824 Chr.18: 26192594-26192594 on Build GRCh38
rs1029453624 Chr.18: 26192682-26192682 on Build GRCh38
rs962664301 Chr.18: 26192728-26192728 on Build GRCh38
rs955097603 Chr.18: 26192717-26192717 on Build GRCh38
rs1024157477 Chr.18: 26192883-26192883 on Build GRCh38
rs903453034 Chr.18: 26192555-26192555 on Build GRCh38
rs950113562 Chr.18: 26192876-26192876 on Build GRCh38
rs918326560 Chr.18: 26193034-26193034 on Build GRCh38
rs1010469928 Chr.18: 26192880-26192880 on Build GRCh38
rs926555945 Chr.18: 26192784-26192784 on Build GRCh38
rs1038127614 Chr.18: 26192697-26192697 on Build GRCh38
rs559269934 Chr.18: 26192839-26192839 on Build GRCh38
rs577734129 Chr.18: 26192929-26192929 on Build GRCh38

View Full Product Details