Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31630863-31631404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388381 c.742C>T p.H248Y Chr.18: 31630993-31630993 on Build GRCh38
987650 c.720C>T p.Y240Y Chr.18: 31631015-31631015 on Build GRCh38
5790044 c.619C>T p.L207F Chr.18: 31631116-31631116 on Build GRCh38
6609475 c.720C>A p.Y240* Chr.18: 31631015-31631015 on Build GRCh38
438048 c.633C>T p.G211G Chr.18: 31631102-31631102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753373002 Chr.18: 31631198-31631199 on Build GRCh38
rs749390292 Chr.18: 31631089-31631089 on Build GRCh38
rs893959892 Chr.18: 31631210-31631211 on Build GRCh38
rs374256252 Chr.18: 31630933-31630933 on Build GRCh38
rs767895710 Chr.18: 31631163-31631163 on Build GRCh38
rs755773107 Chr.18: 31630922-31630922 on Build GRCh38
rs769873090 Chr.18: 31630948-31630948 on Build GRCh38
rs10657836 Chr.18: 31631201-31631202 on Build GRCh38
rs901456078 Chr.18: 31631121-31631121 on Build GRCh38
rs746887661 Chr.18: 31631108-31631108 on Build GRCh38
rs139281930 Chr.18: 31630927-31630927 on Build GRCh38
rs1011041264 Chr.18: 31631211-31631212 on Build GRCh38
rs529792758 Chr.18: 31631360-31631360 on Build GRCh38
rs978458848 Chr.18: 31631325-31631325 on Build GRCh38
rs535479774 Chr.18: 31631256-31631256 on Build GRCh38
rs771234493 Chr.18: 31631057-31631057 on Build GRCh38
rs571747123 Chr.18: 31630888-31630888 on Build GRCh38
rs895863823 Chr.18: 31631112-31631112 on Build GRCh38
rs370684224 Chr.18: 31630996-31630996 on Build GRCh38
rs926194656 Chr.18: 31631341-31631341 on Build GRCh38
rs528822847 Chr.18: 31631344-31631344 on Build GRCh38
rs575895503 Chr.18: 31631200-31631201 on Build GRCh38
rs757620997 Chr.18: 31631029-31631029 on Build GRCh38
rs943045028 Chr.18: 31630981-31630981 on Build GRCh38
rs762503282 Chr.18: 31631025-31631025 on Build GRCh38
rs910327983 Chr.18: 31631291-31631291 on Build GRCh38
rs776527478 Chr.18: 31631122-31631122 on Build GRCh38
rs145579731 Chr.18: 31631039-31631039 on Build GRCh38
rs750823164 Chr.18: 31631171-31631171 on Build GRCh38
rs756491951 Chr.18: 31631175-31631175 on Build GRCh38
rs3794882 Chr.18: 31631353-31631353 on Build GRCh38
rs771956540 Chr.18: 31630970-31630970 on Build GRCh38
rs879094712 Chr.18: 31631177-31631177 on Build GRCh38
rs56030446 Chr.18: 31631215-31631216 on Build GRCh38
rs971882934 Chr.18: 31631184-31631184 on Build GRCh38
rs115587085 Chr.18: 31631066-31631066 on Build GRCh38
rs375543822 Chr.18: 31631322-31631322 on Build GRCh38
rs958172607 Chr.18: 31630898-31630898 on Build GRCh38
rs764464352 Chr.18: 31631024-31631024 on Build GRCh38
rs927903009 Chr.18: 31630949-31630949 on Build GRCh38
rs370233035 Chr.18: 31631013-31631013 on Build GRCh38
rs755029993 Chr.18: 31631195-31631195 on Build GRCh38
rs777579205 Chr.18: 31631096-31631096 on Build GRCh38
rs539168456 Chr.18: 31630892-31630892 on Build GRCh38
rs1016173058 Chr.18: 31631174-31631174 on Build GRCh38
rs373088056 Chr.18: 31630926-31630926 on Build GRCh38
rs764783684 Chr.18: 31630914-31630914 on Build GRCh38
rs779003247 Chr.18: 31631196-31631196 on Build GRCh38
rs759124163 Chr.18: 31630910-31630910 on Build GRCh38
rs943015273 Chr.18: 31631324-31631324 on Build GRCh38
rs201457689 Chr.18: 31630952-31630952 on Build GRCh38
rs759217309 Chr.18: 31631014-31631014 on Build GRCh38
rs773191966 Chr.18: 31630976-31630976 on Build GRCh38
rs764566089 Chr.18: 31631027-31631027 on Build GRCh38
rs575804631 Chr.18: 31631161-31631161 on Build GRCh38
rs550492236 Chr.18: 31630905-31630905 on Build GRCh38
rs565318215 Chr.18: 31631351-31631351 on Build GRCh38
rs954415194 Chr.18: 31631188-31631188 on Build GRCh38
rs1024141341 Chr.18: 31631213-31631214 on Build GRCh38
rs181651811 Chr.18: 31631028-31631028 on Build GRCh38
rs762435135 Chr.18: 31631151-31631151 on Build GRCh38
rs776439295 Chr.18: 31631002-31631002 on Build GRCh38
rs768779660 Chr.18: 31631092-31631092 on Build GRCh38
rs763592255 Chr.18: 31631156-31631156 on Build GRCh38
rs752003560 Chr.18: 31631150-31631150 on Build GRCh38
rs749683444 Chr.18: 31630942-31630942 on Build GRCh38
rs760617720 Chr.18: 31630984-31630984 on Build GRCh38
rs551205879 Chr.18: 31631181-31631181 on Build GRCh38
rs781475875 Chr.18: 31631031-31631031 on Build GRCh38
rs879108763 Chr.18: 31631080-31631080 on Build GRCh38
rs756940141 Chr.18: 31631300-31631300 on Build GRCh38
rs909144418 Chr.18: 31630906-31630906 on Build GRCh38
rs561649679 Chr.18: 31631342-31631342 on Build GRCh38
rs1040036030 Chr.18: 31631103-31631103 on Build GRCh38
rs987110172 Chr.18: 31630943-31630943 on Build GRCh38
rs759374765 Chr.18: 31631135-31631135 on Build GRCh38
rs752189145 Chr.18: 31631024-31631024 on Build GRCh38
rs746021486 Chr.18: 31630944-31630944 on Build GRCh38
rs190821754 Chr.18: 31631087-31631087 on Build GRCh38
rs1003512835 Chr.18: 31631311-31631311 on Build GRCh38
rs781613295 Chr.18: 31631193-31631195 on Build GRCh38
rs753270681 Chr.18: 31630921-31630921 on Build GRCh38
rs779944668 Chr.18: 31630951-31630951 on Build GRCh38
rs569080540 Chr.18: 31630909-31630909 on Build GRCh38
rs961906336 Chr.18: 31630993-31630993 on Build GRCh38
rs769380898 Chr.18: 31631150-31631150 on Build GRCh38
rs563686348 Chr.18: 31631157-31631157 on Build GRCh38
rs757785939 Chr.18: 31631191-31631193 on Build GRCh38
rs368857613 Chr.18: 31630923-31630923 on Build GRCh38
rs770874188 Chr.18: 31631120-31631120 on Build GRCh38
rs546999932 Chr.18: 31631347-31631347 on Build GRCh38
rs754027203 Chr.18: 31631187-31631187 on Build GRCh38
rs764835623 Chr.18: 31631021-31631021 on Build GRCh38
rs199865050 Chr.18: 31631216-31631216 on Build GRCh38
rs114075538 Chr.18: 31631054-31631054 on Build GRCh38
rs770786176 Chr.18: 31631001-31631001 on Build GRCh38

View Full Product Details