Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44060372-44060883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF223
Entrez Gene ID
7766
Gene Name
zinc finger protein 223
Gene Aliases
-
UniGene
Hs.720813
Gene Chromosome Location
Chr.19: 44051372-44069902 on Build GRCh38
Transcripts
NM_013361.5 XM_017027258.1 XM_017027259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4154246 c.87G>T p.Q29H Chr.19: 44060526-44060526 on Build GRCh38
3371352 c.150A>G p.Q50Q Chr.19: 44060756-44060756 on Build GRCh38
333599 c.67G>T p.G23W Chr.19: 44060506-44060506 on Build GRCh38
5451529 c.54T>C p.T18T Chr.19: 44060493-44060493 on Build GRCh38
5390027 c.160C>T p.R54* Chr.19: 44060766-44060766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754304740 Chr.19: 44060723-44060723 on Build GRCh38
rs529043701 Chr.19: 44060713-44060713 on Build GRCh38
rs761007070 Chr.19: 44060704-44060704 on Build GRCh38
rs754805317 Chr.19: 44060548-44060548 on Build GRCh38
rs770294771 Chr.19: 44060601-44060601 on Build GRCh38
rs780913422 Chr.19: 44060557-44060557 on Build GRCh38
rs769847342 Chr.19: 44060440-44060440 on Build GRCh38
rs758606924 Chr.19: 44060730-44060730 on Build GRCh38
rs774070298 Chr.19: 44060611-44060611 on Build GRCh38
rs1034263160 Chr.19: 44060407-44060407 on Build GRCh38
rs369738217 Chr.19: 44060727-44060727 on Build GRCh38
rs774281043 Chr.19: 44060623-44060624 on Build GRCh38
rs764574700 Chr.19: 44060514-44060514 on Build GRCh38
rs778982300 Chr.19: 44060415-44060415 on Build GRCh38
rs868456430 Chr.19: 44060660-44060660 on Build GRCh38
rs766538149 Chr.19: 44060832-44060832 on Build GRCh38
rs777872563 Chr.19: 44060746-44060746 on Build GRCh38
rs759965149 Chr.19: 44060472-44060472 on Build GRCh38
rs755928649 Chr.19: 44060743-44060743 on Build GRCh38
rs3826941 Chr.19: 44060828-44060828 on Build GRCh38
rs906556364 Chr.19: 44060751-44060751 on Build GRCh38
rs750509579 Chr.19: 44060728-44060728 on Build GRCh38
rs989940023 Chr.19: 44060455-44060455 on Build GRCh38
rs376647745 Chr.19: 44060584-44060584 on Build GRCh38
rs776643374 Chr.19: 44060432-44060432 on Build GRCh38
rs773833698 Chr.19: 44060465-44060465 on Build GRCh38
rs747942579 Chr.19: 44060574-44060574 on Build GRCh38
rs915367628 Chr.19: 44060695-44060695 on Build GRCh38
rs939324213 Chr.19: 44060760-44060760 on Build GRCh38
rs559215051 Chr.19: 44060683-44060683 on Build GRCh38
rs950437681 Chr.19: 44060404-44060404 on Build GRCh38
rs762525708 Chr.19: 44060453-44060453 on Build GRCh38
rs4130100 Chr.19: 44060564-44060564 on Build GRCh38
rs930496556 Chr.19: 44060705-44060705 on Build GRCh38
rs748030709 Chr.19: 44060437-44060437 on Build GRCh38
rs762292105 Chr.19: 44060804-44060804 on Build GRCh38
rs780687710 Chr.19: 44060703-44060703 on Build GRCh38
rs184945719 Chr.19: 44060633-44060633 on Build GRCh38
rs766165404 Chr.19: 44060463-44060463 on Build GRCh38
rs779228967 Chr.19: 44060537-44060537 on Build GRCh38
rs200231509 Chr.19: 44060539-44060539 on Build GRCh38
rs748845628 Chr.19: 44060682-44060682 on Build GRCh38
rs879140916 Chr.19: 44060654-44060657 on Build GRCh38
rs777647571 Chr.19: 44060859-44060859 on Build GRCh38
rs764625765 Chr.19: 44060709-44060709 on Build GRCh38
rs765758748 Chr.19: 44060816-44060816 on Build GRCh38
rs935856769 Chr.19: 44060664-44060664 on Build GRCh38
rs1004197548 Chr.19: 44060840-44060840 on Build GRCh38
rs4570979 Chr.19: 44060708-44060708 on Build GRCh38
rs777137488 Chr.19: 44060801-44060801 on Build GRCh38
rs756685152 Chr.19: 44060509-44060509 on Build GRCh38
rs753254563 Chr.19: 44060484-44060484 on Build GRCh38
rs757296070 Chr.19: 44060535-44060535 on Build GRCh38
rs775093129 Chr.19: 44060776-44060776 on Build GRCh38
rs771674484 Chr.19: 44060773-44060773 on Build GRCh38
rs371438704 Chr.19: 44060739-44060739 on Build GRCh38
rs761058579 Chr.19: 44060779-44060779 on Build GRCh38
rs748833908 Chr.19: 44060591-44060591 on Build GRCh38
rs780619926 Chr.19: 44060748-44060748 on Build GRCh38
rs1033945341 Chr.19: 44060850-44060850 on Build GRCh38
rs569036476 Chr.19: 44060798-44060798 on Build GRCh38
rs369091618 Chr.19: 44060418-44060418 on Build GRCh38
rs374214184 Chr.19: 44060427-44060427 on Build GRCh38
rs549180480 Chr.19: 44060767-44060767 on Build GRCh38
rs1018610163 Chr.19: 44060471-44060471 on Build GRCh38
rs749175823 Chr.19: 44060747-44060747 on Build GRCh38
rs751563293 Chr.19: 44060571-44060571 on Build GRCh38
rs4130101 Chr.19: 44060784-44060784 on Build GRCh38
rs945444696 Chr.19: 44060707-44060707 on Build GRCh38
rs138294962 Chr.19: 44060766-44060766 on Build GRCh38
rs772913323 Chr.19: 44060445-44060445 on Build GRCh38
rs775779464 Chr.19: 44060700-44060700 on Build GRCh38
rs370460469 Chr.19: 44060790-44060790 on Build GRCh38
rs753968765 Chr.19: 44060521-44060521 on Build GRCh38
rs373541486 Chr.19: 44060834-44060834 on Build GRCh38
rs915333159 Chr.19: 44060458-44060458 on Build GRCh38
rs763879647 Chr.19: 44060486-44060488 on Build GRCh38
rs780300517 Chr.19: 44060738-44060738 on Build GRCh38
rs770481478 Chr.19: 44060750-44060750 on Build GRCh38
rs773198821 Chr.19: 44060622-44060622 on Build GRCh38
rs149639767 Chr.19: 44060827-44060827 on Build GRCh38
rs564977135 Chr.19: 44060482-44060482 on Build GRCh38
rs773248534 Chr.19: 44060588-44060588 on Build GRCh38
rs777749418 Chr.19: 44060543-44060543 on Build GRCh38
rs775105463 Chr.19: 44060651-44060651 on Build GRCh38
rs370248865 Chr.19: 44060853-44060853 on Build GRCh38
rs202091444 Chr.19: 44060852-44060852 on Build GRCh38

View Full Product Details