Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45609320-45609830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML2
Entrez Gene ID
24139
Gene Name
echinoderm microtubule associated protein like 2
Gene Aliases
ELP70, EMAP-2, EMAP2
UniGene
Hs.24178
Gene Chromosome Location
Chr.19: 45609396-45645601 on Build GRCh38
Transcripts
NM_001193268.1 NM_001193269.1 NM_012155.2 NR_034098.1 XM_006723125.2 XM_006723126.2 XM_011526694.2 XM_011526695.2 XM_011526698.2 XM_011526700.2 XM_011526701.2 XM_017026550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274774 c.1890C>A p.D630E Chr.19: 45609723-45609723 on Build GRCh38
6287570 c.1893C>A p.S631R Chr.19: 45609720-45609720 on Build GRCh38
4127714 c.1859A>G p.H620R Chr.19: 45609754-45609754 on Build GRCh38
4913542 c.2320T>C p.L774L Chr.19: 45609734-45609734 on Build GRCh38
4680882 c.2354_2355insG p.K786fs*>12 Chr.19: 45609699-45609700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756751133 Chr.19: 45609714-45609714 on Build GRCh38
rs944964067 Chr.19: 45609790-45609790 on Build GRCh38
rs1023272338 Chr.19: 45609358-45609358 on Build GRCh38
rs1023555170 Chr.19: 45609587-45609587 on Build GRCh38
rs970356446 Chr.19: 45609411-45609411 on Build GRCh38
rs764353231 Chr.19: 45609716-45609716 on Build GRCh38
rs779017141 Chr.19: 45609648-45609648 on Build GRCh38
rs924155026 Chr.19: 45609442-45609442 on Build GRCh38
rs769550501 Chr.19: 45609664-45609664 on Build GRCh38
rs759872309 Chr.19: 45609601-45609601 on Build GRCh38
rs746114004 Chr.19: 45609764-45609764 on Build GRCh38
rs749466102 Chr.19: 45609634-45609634 on Build GRCh38
rs149432784 Chr.19: 45609703-45609703 on Build GRCh38
rs374769566 Chr.19: 45609630-45609630 on Build GRCh38
rs1012097660 Chr.19: 45609600-45609600 on Build GRCh38
rs748912148 Chr.19: 45609700-45609727 on Build GRCh38
rs911990834 Chr.19: 45609504-45609504 on Build GRCh38
rs548970502 Chr.19: 45609508-45609508 on Build GRCh38
rs749067314 Chr.19: 45609781-45609781 on Build GRCh38
rs774019016 Chr.19: 45609420-45609420 on Build GRCh38
rs781748828 Chr.19: 45609617-45609617 on Build GRCh38
rs762665531 Chr.19: 45609670-45609670 on Build GRCh38
rs899213219 Chr.19: 45609513-45609513 on Build GRCh38
rs200718928 Chr.19: 45609618-45609618 on Build GRCh38
rs910789124 Chr.19: 45609787-45609787 on Build GRCh38
rs201508885 Chr.19: 45609660-45609660 on Build GRCh38
rs555319672 Chr.19: 45609722-45609722 on Build GRCh38
rs1048338092 Chr.19: 45609548-45609548 on Build GRCh38
rs144672629 Chr.19: 45609471-45609471 on Build GRCh38
rs770650644 Chr.19: 45609786-45609786 on Build GRCh38
rs774122975 Chr.19: 45609791-45609791 on Build GRCh38
rs894951444 Chr.19: 45609557-45609557 on Build GRCh38
rs537921186 Chr.19: 45609512-45609512 on Build GRCh38
rs768009438 Chr.19: 45609412-45609412 on Build GRCh38
rs564688150 Chr.19: 45609459-45609459 on Build GRCh38
rs568895975 Chr.19: 45609509-45609509 on Build GRCh38
rs771243512 Chr.19: 45609645-45609645 on Build GRCh38
rs758883171 Chr.19: 45609608-45609608 on Build GRCh38
rs140828547 Chr.19: 45609353-45609353 on Build GRCh38
rs773023936 Chr.19: 45609777-45609777 on Build GRCh38
rs544408124 Chr.19: 45609408-45609408 on Build GRCh38
rs746540115 Chr.19: 45609709-45609709 on Build GRCh38
rs145760913 Chr.19: 45609486-45609486 on Build GRCh38
rs959271248 Chr.19: 45609388-45609388 on Build GRCh38
rs199957663 Chr.19: 45609729-45609729 on Build GRCh38
rs774949062 Chr.19: 45609407-45609407 on Build GRCh38
rs930863827 Chr.19: 45609455-45609455 on Build GRCh38
rs761241607 Chr.19: 45609461-45609461 on Build GRCh38
rs982225128 Chr.19: 45609372-45609372 on Build GRCh38
rs547174462 Chr.19: 45609473-45609473 on Build GRCh38
rs758453328 Chr.19: 45609770-45609770 on Build GRCh38
rs867490815 Chr.19: 45609530-45609530 on Build GRCh38
rs530885479 Chr.19: 45609398-45609398 on Build GRCh38
rs374687287 Chr.19: 45609659-45609659 on Build GRCh38
rs3745817 Chr.19: 45609546-45609546 on Build GRCh38
rs112361770 Chr.19: 45609409-45609409 on Build GRCh38
rs958534017 Chr.19: 45609445-45609445 on Build GRCh38
rs756522464 Chr.19: 45609625-45609625 on Build GRCh38
rs567127923 Chr.19: 45609485-45609485 on Build GRCh38
rs748076682 Chr.19: 45609774-45609774 on Build GRCh38
rs765986079 Chr.19: 45609671-45609671 on Build GRCh38
rs769833074 Chr.19: 45609775-45609775 on Build GRCh38
rs377253305 Chr.19: 45609719-45609719 on Build GRCh38
rs995313725 Chr.19: 45609369-45609369 on Build GRCh38
rs561765694 Chr.19: 45609373-45609373 on Build GRCh38
rs780236516 Chr.19: 45609562-45609563 on Build GRCh38
rs976086864 Chr.19: 45609796-45609796 on Build GRCh38
rs3745816 Chr.19: 45609649-45609649 on Build GRCh38
rs773130735 Chr.19: 45609667-45609667 on Build GRCh38
rs190700619 Chr.19: 45609673-45609673 on Build GRCh38
rs200762252 Chr.19: 45609741-45609741 on Build GRCh38
rs768054324 Chr.19: 45609804-45609807 on Build GRCh38
rs759300949 Chr.19: 45609806-45609806 on Build GRCh38
rs889334344 Chr.19: 45609564-45609564 on Build GRCh38
rs753193742 Chr.19: 45609547-45609547 on Build GRCh38
rs779629336 Chr.19: 45609628-45609628 on Build GRCh38
rs542537075 Chr.19: 45609674-45609674 on Build GRCh38
rs780151012 Chr.19: 45609771-45609771 on Build GRCh38
rs745730374 Chr.19: 45609352-45609352 on Build GRCh38

View Full Product Details