Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47842991-47843498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRX
Entrez Gene ID
1406
Gene Name
cone-rod homeobox
Gene Aliases
CORD2, CRD, LCA7, OTX3
UniGene
Hs.617342
Gene Chromosome Location
Chr.19: 47821842-47843330 on Build GRCh38
Transcripts
NM_000554.4

SNP Detail

SNP ID Chromosome Location
rs988968264 Chr.19: 47843210-47843210 on Build GRCh38
rs1027122859 Chr.19: 47843095-47843095 on Build GRCh38
rs4081725 Chr.19: 47843205-47843205 on Build GRCh38
rs191230190 Chr.19: 47843354-47843354 on Build GRCh38
rs754344803 Chr.19: 47843257-47843257 on Build GRCh38
rs897157393 Chr.19: 47843341-47843341 on Build GRCh38
rs910101081 Chr.19: 47843340-47843340 on Build GRCh38
rs1057012363 Chr.19: 47843373-47843373 on Build GRCh38
rs897276113 Chr.19: 47843035-47843035 on Build GRCh38
rs116336713 Chr.19: 47843084-47843084 on Build GRCh38
rs1044544229 Chr.19: 47843086-47843086 on Build GRCh38
rs918859491 Chr.19: 47843046-47843046 on Build GRCh38
rs771050501 Chr.19: 47843356-47843356 on Build GRCh38
rs1005673716 Chr.19: 47843027-47843039 on Build GRCh38
rs977339938 Chr.19: 47843265-47843265 on Build GRCh38
rs547147791 Chr.19: 47843197-47843197 on Build GRCh38
rs891631697 Chr.19: 47843390-47843390 on Build GRCh38
rs983100786 Chr.19: 47843075-47843075 on Build GRCh38
rs62128811 Chr.19: 47843252-47843252 on Build GRCh38
rs1030493430 Chr.19: 47843202-47843202 on Build GRCh38
rs562629791 Chr.19: 47843081-47843081 on Build GRCh38
rs976909720 Chr.19: 47843045-47843045 on Build GRCh38
rs941821072 Chr.19: 47843085-47843085 on Build GRCh38
rs570099349 Chr.19: 47843408-47843409 on Build GRCh38
rs938431015 Chr.19: 47843362-47843362 on Build GRCh38
rs1018464035 Chr.19: 47843398-47843398 on Build GRCh38
rs1010140134 Chr.19: 47843155-47843155 on Build GRCh38
rs1007075745 Chr.19: 47843128-47843128 on Build GRCh38
rs1057218040 Chr.19: 47843029-47843029 on Build GRCh38
rs11670620 Chr.19: 47843268-47843268 on Build GRCh38
rs984327527 Chr.19: 47843298-47843298 on Build GRCh38
rs905832320 Chr.19: 47843017-47843017 on Build GRCh38
rs549789683 Chr.19: 47843251-47843251 on Build GRCh38
rs112529607 Chr.19: 47843386-47843386 on Build GRCh38
rs888616924 Chr.19: 47843127-47843127 on Build GRCh38
rs11666316 Chr.19: 47843246-47843246 on Build GRCh38
rs151224936 Chr.19: 47843137-47843137 on Build GRCh38
rs560678981 Chr.19: 47843148-47843148 on Build GRCh38
rs140252108 Chr.19: 47843344-47843344 on Build GRCh38
rs753173848 Chr.19: 47843223-47843223 on Build GRCh38
rs370120429 Chr.19: 47843360-47843360 on Build GRCh38
rs904516752 Chr.19: 47843213-47843213 on Build GRCh38
rs1006858785 Chr.19: 47843238-47843238 on Build GRCh38
rs930201503 Chr.19: 47843074-47843074 on Build GRCh38
rs555613161 Chr.19: 47843149-47843149 on Build GRCh38
rs979768607 Chr.19: 47843346-47843346 on Build GRCh38
rs551578738 Chr.19: 47843105-47843105 on Build GRCh38
rs951669764 Chr.19: 47843272-47843272 on Build GRCh38
rs759844877 Chr.19: 47843111-47843111 on Build GRCh38
rs965939233 Chr.19: 47843044-47843044 on Build GRCh38
rs980000018 Chr.19: 47843256-47843256 on Build GRCh38
rs938531976 Chr.19: 47843122-47843122 on Build GRCh38

View Full Product Details