Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49958417-49958910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIGLEC11
Entrez Gene ID
114132
Gene Name
sialic acid binding Ig like lectin 11
Gene Aliases
-
UniGene
Hs.661852
Gene Chromosome Location
Chr.19: 49948993-49961172 on Build GRCh38
Transcripts
NM_001135163.1 NM_052884.2 XM_005258476.3 XM_005258477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
363525 c.1320C>A p.H440Q Chr.19: 49958686-49958686 on Build GRCh38
5390352 c.1296A>G p.G432G Chr.19: 49958710-49958710 on Build GRCh38
4448845 c.1364-1G>A p.? Chr.19: 49958571-49958571 on Build GRCh38
713350 c.1266G>C p.E422D Chr.19: 49958740-49958740 on Build GRCh38
1293808 c.1179C>A p.T393T Chr.19: 49958827-49958827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201935510 Chr.19: 49958790-49958790 on Build GRCh38
rs749897827 Chr.19: 49958454-49958454 on Build GRCh38
rs761548091 Chr.19: 49958486-49958486 on Build GRCh38
rs746150139 Chr.19: 49958789-49958789 on Build GRCh38
rs571622268 Chr.19: 49958745-49958745 on Build GRCh38
rs141171190 Chr.19: 49958543-49958543 on Build GRCh38
rs770833596 Chr.19: 49958587-49958600 on Build GRCh38
rs758267463 Chr.19: 49958830-49958830 on Build GRCh38
rs749028221 Chr.19: 49958845-49958845 on Build GRCh38
rs555532137 Chr.19: 49958599-49958599 on Build GRCh38
rs150050230 Chr.19: 49958842-49958842 on Build GRCh38
rs150811494 Chr.19: 49958866-49958866 on Build GRCh38
rs781189521 Chr.19: 49958656-49958656 on Build GRCh38
rs144054317 Chr.19: 49958663-49958663 on Build GRCh38
rs149136670 Chr.19: 49958867-49958867 on Build GRCh38
rs757457875 Chr.19: 49958748-49958748 on Build GRCh38
rs781322471 Chr.19: 49958575-49958575 on Build GRCh38
rs766070165 Chr.19: 49958884-49958884 on Build GRCh38
rs201758933 Chr.19: 49958620-49958620 on Build GRCh38
rs201076601 Chr.19: 49958716-49958716 on Build GRCh38
rs754522711 Chr.19: 49958781-49958781 on Build GRCh38
rs369053163 Chr.19: 49958604-49958604 on Build GRCh38
rs761735827 Chr.19: 49958869-49958869 on Build GRCh38
rs761012038 Chr.19: 49958481-49958481 on Build GRCh38
rs774169073 Chr.19: 49958635-49958635 on Build GRCh38
rs887981723 Chr.19: 49958838-49958858 on Build GRCh38
rs767952887 Chr.19: 49958734-49958734 on Build GRCh38
rs377029985 Chr.19: 49958668-49958668 on Build GRCh38
rs199604054 Chr.19: 49958843-49958843 on Build GRCh38
rs145434757 Chr.19: 49958844-49958844 on Build GRCh38
rs571259246 Chr.19: 49958479-49958479 on Build GRCh38
rs757057701 Chr.19: 49958704-49958704 on Build GRCh38
rs941350368 Chr.19: 49958757-49958757 on Build GRCh38
rs773147639 Chr.19: 49958487-49958487 on Build GRCh38
rs762383298 Chr.19: 49958488-49958488 on Build GRCh38
rs890913673 Chr.19: 49958780-49958780 on Build GRCh38
rs771866761 Chr.19: 49958715-49958715 on Build GRCh38
rs773041395 Chr.19: 49958630-49958630 on Build GRCh38
rs376222513 Chr.19: 49958702-49958702 on Build GRCh38
rs754126439 Chr.19: 49958703-49958703 on Build GRCh38
rs967712535 Chr.19: 49958517-49958517 on Build GRCh38
rs781409910 Chr.19: 49958622-49958622 on Build GRCh38
rs776744999 Chr.19: 49958546-49958546 on Build GRCh38
rs545039683 Chr.19: 49958666-49958666 on Build GRCh38
rs767139669 Chr.19: 49958584-49958584 on Build GRCh38
rs779491398 Chr.19: 49958525-49958525 on Build GRCh38
rs746442120 Chr.19: 49958526-49958526 on Build GRCh38
rs766335832 Chr.19: 49958834-49958834 on Build GRCh38
rs769846643 Chr.19: 49958599-49958599 on Build GRCh38
rs547243506 Chr.19: 49958725-49958725 on Build GRCh38
rs369288400 Chr.19: 49958690-49958690 on Build GRCh38
rs751230929 Chr.19: 49958473-49958473 on Build GRCh38
rs761976908 Chr.19: 49958813-49958813 on Build GRCh38
rs763314421 Chr.19: 49958684-49958684 on Build GRCh38
rs776441362 Chr.19: 49958683-49958683 on Build GRCh38
rs747066618 Chr.19: 49958609-49958609 on Build GRCh38
rs146975706 Chr.19: 49958446-49958446 on Build GRCh38
rs759965484 Chr.19: 49958733-49958733 on Build GRCh38
rs200076826 Chr.19: 49958775-49958775 on Build GRCh38
rs778620202 Chr.19: 49958705-49958705 on Build GRCh38
rs773869337 Chr.19: 49958569-49958569 on Build GRCh38
rs534540205 Chr.19: 49958875-49958875 on Build GRCh38
rs1865042 Chr.19: 49958636-49958636 on Build GRCh38
rs57682421 Chr.19: 49958478-49958478 on Build GRCh38
rs774621658 Chr.19: 49958732-49958732 on Build GRCh38
rs545385303 Chr.19: 49958881-49958882 on Build GRCh38
rs1021072788 Chr.19: 49958517-49958523 on Build GRCh38
rs779625739 Chr.19: 49958464-49958464 on Build GRCh38
rs780433809 Chr.19: 49958865-49958865 on Build GRCh38
rs1009232120 Chr.19: 49958504-49958504 on Build GRCh38
rs765942084 Chr.19: 49958490-49958490 on Build GRCh38
rs773296577 Chr.19: 49958876-49958876 on Build GRCh38
rs768316332 Chr.19: 49958483-49958483 on Build GRCh38
rs747014469 Chr.19: 49958779-49958779 on Build GRCh38
rs150740722 Chr.19: 49958572-49958572 on Build GRCh38
rs759016158 Chr.19: 49958600-49958600 on Build GRCh38
rs747171253 Chr.19: 49958538-49958538 on Build GRCh38
rs575317749 Chr.19: 49958535-49958535 on Build GRCh38
rs201895287 Chr.19: 49958675-49958675 on Build GRCh38
rs776530627 Chr.19: 49958806-49958806 on Build GRCh38
rs780074195 Chr.19: 49958778-49958778 on Build GRCh38
rs577987618 Chr.19: 49958886-49958886 on Build GRCh38
rs760450249 Chr.19: 49958645-49958645 on Build GRCh38
rs749111639 Chr.19: 49958634-49958634 on Build GRCh38
rs538978935 Chr.19: 49958827-49958827 on Build GRCh38
rs775847478 Chr.19: 49958480-49958480 on Build GRCh38
rs962969134 Chr.19: 49958853-49958853 on Build GRCh38
rs776419227 Chr.19: 49958730-49958730 on Build GRCh38
rs774799128 Chr.19: 49958812-49958813 on Build GRCh38
rs748410524 Chr.19: 49958592-49958592 on Build GRCh38
rs747805855 Chr.19: 49958785-49958785 on Build GRCh38
rs556999247 Chr.19: 49958491-49958491 on Build GRCh38
rs775191861 Chr.19: 49958643-49958643 on Build GRCh38
rs146836661 Chr.19: 49958669-49958669 on Build GRCh38
rs376377903 Chr.19: 49958838-49958838 on Build GRCh38
rs772928619 Chr.19: 49958788-49958788 on Build GRCh38
rs763484283 Chr.19: 49958631-49958632 on Build GRCh38
rs374104181 Chr.19: 49958881-49958881 on Build GRCh38
rs769565241 Chr.19: 49958628-49958628 on Build GRCh38
rs761215724 Chr.19: 49958795-49958795 on Build GRCh38
rs745784422 Chr.19: 49958707-49958707 on Build GRCh38
rs139230914 Chr.19: 49958650-49958650 on Build GRCh38
rs769581557 Chr.19: 49958563-49958563 on Build GRCh38
rs748436268 Chr.19: 49958627-49958627 on Build GRCh38
rs749024800 Chr.19: 49958667-49958667 on Build GRCh38
rs767105458 Chr.19: 49958498-49958498 on Build GRCh38
rs200767146 Chr.19: 49958477-49958477 on Build GRCh38
rs550434699 Chr.19: 49958787-49958787 on Build GRCh38
rs768949964 Chr.19: 49958791-49958791 on Build GRCh38
rs201115105 Chr.19: 49958786-49958786 on Build GRCh38
rs764698634 Chr.19: 49958453-49958453 on Build GRCh38
rs757927465 Chr.19: 49958459-49958459 on Build GRCh38
rs139706382 Chr.19: 49958682-49958682 on Build GRCh38
rs142292396 Chr.19: 49958880-49958880 on Build GRCh38
rs762937007 Chr.19: 49958566-49958566 on Build GRCh38
rs747920096 Chr.19: 49958551-49958551 on Build GRCh38
rs556913162 Chr.19: 49958839-49958839 on Build GRCh38
rs750666230 Chr.19: 49958828-49958828 on Build GRCh38
rs772490728 Chr.19: 49958792-49958792 on Build GRCh38
rs764124969 Chr.19: 49958799-49958799 on Build GRCh38
rs777762992 Chr.19: 49958651-49958651 on Build GRCh38
rs992091553 Chr.19: 49958601-49958601 on Build GRCh38
rs754698790 Chr.19: 49958492-49958492 on Build GRCh38
rs754182179 Chr.19: 49958500-49958500 on Build GRCh38
rs760739503 Chr.19: 49958738-49958738 on Build GRCh38
rs754180000 Chr.19: 49958632-49958632 on Build GRCh38
rs772399210 Chr.19: 49958848-49958848 on Build GRCh38
rs530600033 Chr.19: 49958678-49958678 on Build GRCh38
rs199892510 Chr.19: 49958793-49958793 on Build GRCh38
rs755080469 Chr.19: 49958655-49958655 on Build GRCh38
rs369116394 Chr.19: 49958649-49958649 on Build GRCh38
rs752723570 Chr.19: 49958698-49958698 on Build GRCh38
rs371066318 Chr.19: 49958694-49958694 on Build GRCh38
rs775980546 Chr.19: 49958736-49958736 on Build GRCh38
rs752293127 Chr.19: 49958596-49958596 on Build GRCh38
rs757686993 Chr.19: 49958597-49958597 on Build GRCh38
rs752895268 Chr.19: 49958661-49958661 on Build GRCh38
rs370432875 Chr.19: 49958761-49958761 on Build GRCh38
rs747814700 Chr.19: 49958731-49958731 on Build GRCh38
rs765525096 Chr.19: 49958751-49958751 on Build GRCh38
rs374158248 Chr.19: 49958695-49958695 on Build GRCh38
rs7259039 Chr.19: 49958475-49958475 on Build GRCh38
rs538668574 Chr.19: 49958485-49958485 on Build GRCh38
rs765843318 Chr.19: 49958567-49958567 on Build GRCh38
rs750138834 Chr.19: 49958752-49958752 on Build GRCh38
rs767289497 Chr.19: 49958887-49958887 on Build GRCh38
rs950062731 Chr.19: 49958467-49958467 on Build GRCh38
rs148610893 Chr.19: 49958542-49958542 on Build GRCh38
rs780898655 Chr.19: 49958783-49958783 on Build GRCh38
rs758535718 Chr.19: 49958533-49958533 on Build GRCh38
rs541281546 Chr.19: 49958638-49958638 on Build GRCh38

View Full Product Details