Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58392890-58393399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS5
Entrez Gene ID
6193
Gene Name
ribosomal protein S5
Gene Aliases
S5
UniGene
Hs.378103
Gene Chromosome Location
Chr.19: 58387269-58394804 on Build GRCh38
Transcripts
NM_001009.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6270412 c.190G>T p.A64S Chr.19: 58393057-58393057 on Build GRCh38
4722689 c.220A>T p.N74Y Chr.19: 58393087-58393087 on Build GRCh38
1002710 c.315C>T p.G105G Chr.19: 58393182-58393182 on Build GRCh38
4722688 c.210G>A p.E70E Chr.19: 58393077-58393077 on Build GRCh38
1002709 c.240C>A p.G80G Chr.19: 58393107-58393107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746231472 Chr.19: 58393234-58393234 on Build GRCh38
rs941966457 Chr.19: 58393268-58393269 on Build GRCh38
rs753707781 Chr.19: 58393166-58393166 on Build GRCh38
rs778315513 Chr.19: 58393109-58393109 on Build GRCh38
rs113252728 Chr.19: 58393270-58393270 on Build GRCh38
rs1046168621 Chr.19: 58393041-58393041 on Build GRCh38
rs907048542 Chr.19: 58393045-58393045 on Build GRCh38
rs749586251 Chr.19: 58393158-58393158 on Build GRCh38
rs781001606 Chr.19: 58392933-58392933 on Build GRCh38
rs748792780 Chr.19: 58392952-58392952 on Build GRCh38
rs776498157 Chr.19: 58393346-58393346 on Build GRCh38
rs762497331 Chr.19: 58393365-58393365 on Build GRCh38
rs759312572 Chr.19: 58393347-58393347 on Build GRCh38
rs201177783 Chr.19: 58393083-58393083 on Build GRCh38
rs941927479 Chr.19: 58392922-58392922 on Build GRCh38
rs752244586 Chr.19: 58393176-58393176 on Build GRCh38
rs765402531 Chr.19: 58393038-58393038 on Build GRCh38
rs200602413 Chr.19: 58393110-58393110 on Build GRCh38
rs112637700 Chr.19: 58393144-58393144 on Build GRCh38
rs756088743 Chr.19: 58393308-58393309 on Build GRCh38
rs769932302 Chr.19: 58393135-58393135 on Build GRCh38
rs961447783 Chr.19: 58393056-58393056 on Build GRCh38
rs149481790 Chr.19: 58393164-58393164 on Build GRCh38
rs754563972 Chr.19: 58392961-58392961 on Build GRCh38
rs776807710 Chr.19: 58393129-58393129 on Build GRCh38
rs757796615 Chr.19: 58393232-58393232 on Build GRCh38
rs752104136 Chr.19: 58393217-58393217 on Build GRCh38
rs570678601 Chr.19: 58393374-58393374 on Build GRCh38
rs989527544 Chr.19: 58393224-58393224 on Build GRCh38
rs964672327 Chr.19: 58393155-58393155 on Build GRCh38
rs368258878 Chr.19: 58393339-58393339 on Build GRCh38
rs907137981 Chr.19: 58393245-58393245 on Build GRCh38
rs533747867 Chr.19: 58393141-58393141 on Build GRCh38
rs755772448 Chr.19: 58392939-58392939 on Build GRCh38
rs771336588 Chr.19: 58393122-58393122 on Build GRCh38
rs376088776 Chr.19: 58392926-58392926 on Build GRCh38
rs778437053 Chr.19: 58393104-58393104 on Build GRCh38
rs201157224 Chr.19: 58393229-58393229 on Build GRCh38
rs562012326 Chr.19: 58393100-58393100 on Build GRCh38
rs768895074 Chr.19: 58393336-58393336 on Build GRCh38
rs771005130 Chr.19: 58393344-58393344 on Build GRCh38
rs1019256720 Chr.19: 58393353-58393353 on Build GRCh38
rs916278640 Chr.19: 58393301-58393301 on Build GRCh38
rs759592801 Chr.19: 58393034-58393034 on Build GRCh38
rs749034113 Chr.19: 58393011-58393011 on Build GRCh38
rs529055606 Chr.19: 58393244-58393244 on Build GRCh38
rs756041099 Chr.19: 58393059-58393059 on Build GRCh38
rs748648108 Chr.19: 58392996-58392996 on Build GRCh38
rs61731810 Chr.19: 58393143-58393143 on Build GRCh38
rs779857795 Chr.19: 58392943-58392943 on Build GRCh38
rs753583866 Chr.19: 58393067-58393067 on Build GRCh38
rs751568889 Chr.19: 58392925-58392925 on Build GRCh38
rs766106173 Chr.19: 58393066-58393066 on Build GRCh38
rs769992691 Chr.19: 58393325-58393325 on Build GRCh38
rs777715073 Chr.19: 58392921-58392921 on Build GRCh38
rs533211173 Chr.19: 58393206-58393206 on Build GRCh38
rs563656934 Chr.19: 58393233-58393233 on Build GRCh38
rs1046044195 Chr.19: 58393315-58393315 on Build GRCh38
rs2289143 Chr.19: 58393348-58393348 on Build GRCh38
rs760513809 Chr.19: 58393148-58393148 on Build GRCh38
rs368930275 Chr.19: 58393134-58393134 on Build GRCh38
rs764798367 Chr.19: 58393170-58393170 on Build GRCh38
rs757508690 Chr.19: 58393301-58393301 on Build GRCh38
rs542382691 Chr.19: 58392920-58392920 on Build GRCh38
rs761762077 Chr.19: 58393046-58393046 on Build GRCh38
rs750178638 Chr.19: 58393052-58393052 on Build GRCh38
rs11545062 Chr.19: 58393050-58393050 on Build GRCh38
rs367711986 Chr.19: 58393071-58393071 on Build GRCh38
rs772474879 Chr.19: 58393002-58393002 on Build GRCh38
rs6721 Chr.19: 58393029-58393029 on Build GRCh38
rs557791855 Chr.19: 58392918-58392918 on Build GRCh38
rs765671546 Chr.19: 58393330-58393330 on Build GRCh38
rs760640309 Chr.19: 58393342-58393342 on Build GRCh38
rs995208020 Chr.19: 58393084-58393084 on Build GRCh38
rs370061857 Chr.19: 58393106-58393106 on Build GRCh38
rs896989544 Chr.19: 58393324-58393324 on Build GRCh38
rs779256683 Chr.19: 58392973-58392973 on Build GRCh38
rs534915188 Chr.19: 58393328-58393328 on Build GRCh38
rs756974556 Chr.19: 58392929-58392929 on Build GRCh38
rs758870387 Chr.19: 58393108-58393108 on Build GRCh38
rs2289144 Chr.19: 58393303-58393303 on Build GRCh38
rs140308068 Chr.19: 58392934-58392934 on Build GRCh38
rs775466724 Chr.19: 58393039-58393039 on Build GRCh38
rs754596907 Chr.19: 58393167-58393167 on Build GRCh38
rs1053823036 Chr.19: 58392948-58392948 on Build GRCh38
rs113669389 Chr.19: 58393026-58393026 on Build GRCh38
rs7245665 Chr.19: 58393298-58393298 on Build GRCh38
rs374641510 Chr.19: 58393116-58393116 on Build GRCh38
rs375375343 Chr.19: 58393191-58393191 on Build GRCh38
rs181145116 Chr.19: 58393350-58393350 on Build GRCh38

View Full Product Details