Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61873015-61873529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT4
Entrez Gene ID
10575
Gene Name
chaperonin containing TCP1 subunit 4
Gene Aliases
CCT-DELTA, Cctd, SRB
UniGene
Hs.421509
Gene Chromosome Location
Chr.2: 61868127-61888671 on Build GRCh38
Transcripts
NM_001256721.1 NM_006430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445597 c.1015-3C>T p.? Chr.2: 61873115-61873115 on Build GRCh38
5344244 c.1015-4C>T p.? Chr.2: 61873116-61873116 on Build GRCh38
1408938 c.918-2A>G p.? Chr.2: 61873295-61873295 on Build GRCh38
3714296 c.1086T>A p.A362A Chr.2: 61873041-61873041 on Build GRCh38
6485782 c.1015-1G>T p.? Chr.2: 61873113-61873113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76248080 Chr.2: 61873245-61873245 on Build GRCh38
rs966484731 Chr.2: 61873134-61873134 on Build GRCh38
rs934078983 Chr.2: 61873298-61873298 on Build GRCh38
rs534161091 Chr.2: 61873374-61873374 on Build GRCh38
rs761850138 Chr.2: 61873328-61873328 on Build GRCh38
rs140356041 Chr.2: 61873244-61873244 on Build GRCh38
rs572369860 Chr.2: 61873366-61873369 on Build GRCh38
rs560370213 Chr.2: 61873079-61873079 on Build GRCh38
rs770017935 Chr.2: 61873248-61873248 on Build GRCh38
rs749950723 Chr.2: 61873091-61873091 on Build GRCh38
rs1042318210 Chr.2: 61873412-61873412 on Build GRCh38
rs1012353756 Chr.2: 61873472-61873472 on Build GRCh38
rs1016843318 Chr.2: 61873482-61873482 on Build GRCh38
rs544152631 Chr.2: 61873487-61873487 on Build GRCh38
rs765903878 Chr.2: 61873302-61873302 on Build GRCh38
rs115460183 Chr.2: 61873318-61873318 on Build GRCh38
rs775110320 Chr.2: 61873058-61873058 on Build GRCh38
rs759166372 Chr.2: 61873315-61873315 on Build GRCh38
rs902226626 Chr.2: 61873418-61873418 on Build GRCh38
rs778449826 Chr.2: 61873231-61873231 on Build GRCh38
rs772761339 Chr.2: 61873116-61873116 on Build GRCh38
rs973010455 Chr.2: 61873283-61873283 on Build GRCh38
rs771947617 Chr.2: 61873278-61873278 on Build GRCh38
rs763847917 Chr.2: 61873070-61873070 on Build GRCh38
rs773177932 Chr.2: 61873279-61873279 on Build GRCh38
rs747146630 Chr.2: 61873125-61873125 on Build GRCh38
rs909820683 Chr.2: 61873160-61873163 on Build GRCh38
rs767697848 Chr.2: 61873166-61873166 on Build GRCh38
rs918674952 Chr.2: 61873284-61873284 on Build GRCh38
rs112590269 Chr.2: 61873202-61873202 on Build GRCh38
rs904806015 Chr.2: 61873312-61873312 on Build GRCh38
rs755292873 Chr.2: 61873229-61873229 on Build GRCh38
rs767449319 Chr.2: 61873161-61873161 on Build GRCh38
rs575288657 Chr.2: 61873421-61873422 on Build GRCh38
rs200991443 Chr.2: 61873266-61873266 on Build GRCh38
rs368854623 Chr.2: 61873170-61873170 on Build GRCh38
rs370736566 Chr.2: 61873498-61873499 on Build GRCh38
rs757762953 Chr.2: 61873332-61873332 on Build GRCh38
rs372770767 Chr.2: 61873188-61873189 on Build GRCh38
rs767003656 Chr.2: 61873086-61873086 on Build GRCh38
rs755347768 Chr.2: 61873106-61873106 on Build GRCh38
rs547107601 Chr.2: 61873153-61873153 on Build GRCh38
rs374445671 Chr.2: 61873143-61873143 on Build GRCh38
rs577106563 Chr.2: 61873105-61873105 on Build GRCh38
rs918399342 Chr.2: 61873173-61873173 on Build GRCh38
rs566706824 Chr.2: 61873373-61873373 on Build GRCh38
rs541151125 Chr.2: 61873389-61873389 on Build GRCh38
rs751467696 Chr.2: 61873077-61873077 on Build GRCh38
rs569450061 Chr.2: 61873053-61873053 on Build GRCh38
rs985051971 Chr.2: 61873156-61873156 on Build GRCh38
rs546053203 Chr.2: 61873169-61873169 on Build GRCh38
rs904755375 Chr.2: 61873102-61873102 on Build GRCh38
rs780274730 Chr.2: 61873341-61873341 on Build GRCh38
rs972551424 Chr.2: 61873502-61873502 on Build GRCh38
rs759535015 Chr.2: 61873122-61873123 on Build GRCh38
rs891952556 Chr.2: 61873340-61873340 on Build GRCh38
rs776059241 Chr.2: 61873314-61873314 on Build GRCh38
rs1020690054 Chr.2: 61873111-61873111 on Build GRCh38
rs753065186 Chr.2: 61873319-61873319 on Build GRCh38
rs569498667 Chr.2: 61873321-61873321 on Build GRCh38
rs753495416 Chr.2: 61873193-61873193 on Build GRCh38
rs779786336 Chr.2: 61873097-61873097 on Build GRCh38
rs142340764 Chr.2: 61873221-61873221 on Build GRCh38
rs942934622 Chr.2: 61873355-61873355 on Build GRCh38
rs1054697713 Chr.2: 61873368-61873368 on Build GRCh38
rs184362993 Chr.2: 61873325-61873325 on Build GRCh38
rs760569395 Chr.2: 61873282-61873282 on Build GRCh38
rs765680469 Chr.2: 61873220-61873220 on Build GRCh38
rs960825905 Chr.2: 61873471-61873471 on Build GRCh38
rs11549871 Chr.2: 61873043-61873043 on Build GRCh38
rs760963243 Chr.2: 61873054-61873054 on Build GRCh38
rs762833143 Chr.2: 61873059-61873059 on Build GRCh38
rs953851016 Chr.2: 61873144-61873144 on Build GRCh38
rs754650860 Chr.2: 61873210-61873210 on Build GRCh38
rs758936362 Chr.2: 61873230-61873230 on Build GRCh38
rs774896460 Chr.2: 61873401-61873401 on Build GRCh38
rs771012319 Chr.2: 61873128-61873128 on Build GRCh38
rs539217964 Chr.2: 61873186-61873187 on Build GRCh38
rs752105942 Chr.2: 61873328-61873328 on Build GRCh38
rs144182806 Chr.2: 61873236-61873236 on Build GRCh38
rs148780970 Chr.2: 61873109-61873109 on Build GRCh38
rs774067604 Chr.2: 61873262-61873262 on Build GRCh38
rs1034660059 Chr.2: 61873447-61873447 on Build GRCh38
rs373666408 Chr.2: 61873101-61873101 on Build GRCh38
rs962986848 Chr.2: 61873486-61873486 on Build GRCh38
rs745941478 Chr.2: 61873338-61873338 on Build GRCh38
rs576867176 Chr.2: 61873397-61873397 on Build GRCh38
rs756290850 Chr.2: 61873339-61873339 on Build GRCh38
rs367852061 Chr.2: 61873108-61873108 on Build GRCh38
rs774190197 Chr.2: 61873151-61873151 on Build GRCh38
rs200182800 Chr.2: 61873334-61873334 on Build GRCh38
rs372201903 Chr.2: 61873141-61873141 on Build GRCh38
rs755653715 Chr.2: 61873094-61873094 on Build GRCh38
rs998330030 Chr.2: 61873362-61873362 on Build GRCh38
rs571816651 Chr.2: 61873179-61873179 on Build GRCh38
rs777939659 Chr.2: 61873119-61873119 on Build GRCh38
rs370544712 Chr.2: 61873057-61873057 on Build GRCh38
rs376388698 Chr.2: 61873052-61873052 on Build GRCh38
rs1003302895 Chr.2: 61873432-61873432 on Build GRCh38
rs774351359 Chr.2: 61873116-61873117 on Build GRCh38
rs561001554 Chr.2: 61873462-61873462 on Build GRCh38

View Full Product Details